Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43795341-43795842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERF2
Entrez Gene ID
10169
Gene Name
small EDRK-rich factor 2
Gene Aliases
4F5REL, FAM2C, H4F5REL, HsT17089
UniGene
Hs.424126
Gene Chromosome Location
Chr.15: 43777096-43796089 on Build GRCh38
Transcripts
NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1 NM_001258031.1 NM_001258032.1

Gene Symbol
SERF2-C15ORF63
Entrez Gene ID
100529067
Gene Name
SERF2-C15orf63 readthrough
Gene Aliases
-
UniGene
Hs.730672
Gene Chromosome Location
Chr.15: 43791976-43802589 on Build GRCh38
Transcripts
NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1 NM_001258031.1 NM_001258032.1

Gene Symbol
SERINC4
Entrez Gene ID
619189
Gene Name
serine incorporator 4
Gene Aliases
-
UniGene
Hs.424126
Gene Chromosome Location
Chr.15: 43794174-43800097 on Build GRCh38
Transcripts
NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1 NM_001258031.1 NM_001258032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6270133 c.549C>A p.A183A Chr.15: 43795450-43795450 on Build GRCh38
5948818 c.506C>T p.A169V Chr.15: 43795493-43795493 on Build GRCh38
5920881 c.1204G>A p.A402T Chr.15: 43795527-43795527 on Build GRCh38
5948819 c.1238C>T p.A413V Chr.15: 43795493-43795493 on Build GRCh38
3816159 c.1290C>T p.F430F Chr.15: 43795441-43795441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053631454 Chr.15: 43795462-43795462 on Build GRCh38
rs754731664 Chr.15: 43795747-43795747 on Build GRCh38
rs752634344 Chr.15: 43795726-43795729 on Build GRCh38
rs764891161 Chr.15: 43795652-43795652 on Build GRCh38
rs376001419 Chr.15: 43795523-43795523 on Build GRCh38
rs745858511 Chr.15: 43795561-43795561 on Build GRCh38
rs755197980 Chr.15: 43795539-43795539 on Build GRCh38
rs770807720 Chr.15: 43795455-43795455 on Build GRCh38
rs897058178 Chr.15: 43795595-43795595 on Build GRCh38
rs775893138 Chr.15: 43795578-43795578 on Build GRCh38
rs763503353 Chr.15: 43795646-43795646 on Build GRCh38
rs966267118 Chr.15: 43795771-43795771 on Build GRCh38
rs925785168 Chr.15: 43795750-43795750 on Build GRCh38
rs767483405 Chr.15: 43795722-43795723 on Build GRCh38
rs546163079 Chr.15: 43795562-43795562 on Build GRCh38
rs778999629 Chr.15: 43795550-43795550 on Build GRCh38
rs200290403 Chr.15: 43795484-43795484 on Build GRCh38
rs912016377 Chr.15: 43795817-43795817 on Build GRCh38
rs562892392 Chr.15: 43795598-43795598 on Build GRCh38
rs746775911 Chr.15: 43795574-43795574 on Build GRCh38
rs758439676 Chr.15: 43795726-43795726 on Build GRCh38
rs372346641 Chr.15: 43795651-43795651 on Build GRCh38
rs1031821762 Chr.15: 43795784-43795784 on Build GRCh38
rs74494149 Chr.15: 43795540-43795540 on Build GRCh38
rs766791350 Chr.15: 43795527-43795527 on Build GRCh38
rs944720059 Chr.15: 43795512-43795512 on Build GRCh38
rs976397453 Chr.15: 43795397-43795397 on Build GRCh38
rs764317737 Chr.15: 43795682-43795682 on Build GRCh38
rs761385821 Chr.15: 43795420-43795423 on Build GRCh38
rs767647154 Chr.15: 43795662-43795662 on Build GRCh38
rs201646905 Chr.15: 43795520-43795520 on Build GRCh38
rs150100062 Chr.15: 43795417-43795417 on Build GRCh38
rs777047886 Chr.15: 43795385-43795386 on Build GRCh38
rs756161071 Chr.15: 43795543-43795543 on Build GRCh38
rs200128740 Chr.15: 43795666-43795666 on Build GRCh38
rs142695112 Chr.15: 43795437-43795437 on Build GRCh38
rs1003110455 Chr.15: 43795579-43795579 on Build GRCh38
rs762700601 Chr.15: 43795545-43795546 on Build GRCh38
rs966608903 Chr.15: 43795390-43795390 on Build GRCh38
rs756391238 Chr.15: 43795673-43795673 on Build GRCh38
rs780841671 Chr.15: 43795758-43795758 on Build GRCh38
rs373346812 Chr.15: 43795617-43795617 on Build GRCh38
rs772863927 Chr.15: 43795543-43795543 on Build GRCh38
rs766237941 Chr.15: 43795545-43795553 on Build GRCh38
rs753936874 Chr.15: 43795708-43795708 on Build GRCh38
rs147441322 Chr.15: 43795444-43795444 on Build GRCh38
rs776032811 Chr.15: 43795491-43795491 on Build GRCh38
rs778859490 Chr.15: 43795755-43795756 on Build GRCh38
rs934734473 Chr.15: 43795445-43795445 on Build GRCh38
rs767615817 Chr.15: 43795638-43795638 on Build GRCh38
rs747895764 Chr.15: 43795763-43795763 on Build GRCh38
rs751424100 Chr.15: 43795659-43795661 on Build GRCh38
rs768321884 Chr.15: 43795485-43795485 on Build GRCh38
rs772059515 Chr.15: 43795483-43795483 on Build GRCh38
rs751850244 Chr.15: 43795440-43795440 on Build GRCh38
rs199617532 Chr.15: 43795737-43795737 on Build GRCh38
rs774611552 Chr.15: 43795653-43795653 on Build GRCh38
rs745672740 Chr.15: 43795482-43795482 on Build GRCh38
rs757303218 Chr.15: 43795746-43795747 on Build GRCh38
rs369620901 Chr.15: 43795568-43795568 on Build GRCh38
rs911878304 Chr.15: 43795506-43795506 on Build GRCh38
rs775860074 Chr.15: 43795364-43795367 on Build GRCh38
rs868632203 Chr.15: 43795778-43795778 on Build GRCh38
rs780033413 Chr.15: 43795570-43795570 on Build GRCh38
rs914549493 Chr.15: 43795450-43795450 on Build GRCh38
rs376623976 Chr.15: 43795459-43795459 on Build GRCh38
rs372519705 Chr.15: 43795735-43795735 on Build GRCh38
rs765732235 Chr.15: 43795377-43795377 on Build GRCh38
rs761334957 Chr.15: 43795492-43795492 on Build GRCh38
rs768347596 Chr.15: 43795545-43795545 on Build GRCh38
rs547487335 Chr.15: 43795792-43795792 on Build GRCh38
rs751932662 Chr.15: 43795535-43795535 on Build GRCh38
rs149510309 Chr.15: 43795629-43795629 on Build GRCh38
rs781181615 Chr.15: 43795441-43795441 on Build GRCh38
rs200741760 Chr.15: 43795460-43795460 on Build GRCh38
rs763450248 Chr.15: 43795524-43795524 on Build GRCh38
rs369243797 Chr.15: 43795591-43795591 on Build GRCh38
rs995502685 Chr.15: 43795569-43795569 on Build GRCh38
rs745469198 Chr.15: 43795776-43795777 on Build GRCh38
rs956092153 Chr.15: 43795751-43795751 on Build GRCh38
rs759897948 Chr.15: 43795656-43795656 on Build GRCh38
rs532961414 Chr.15: 43795414-43795414 on Build GRCh38
rs747612909 Chr.15: 43795593-43795593 on Build GRCh38
rs181235842 Chr.15: 43795636-43795636 on Build GRCh38
rs757004016 Chr.15: 43795475-43795475 on Build GRCh38
rs754858046 Chr.15: 43795691-43795694 on Build GRCh38
rs769363902 Chr.15: 43795642-43795642 on Build GRCh38
rs924844763 Chr.15: 43795431-43795431 on Build GRCh38
rs747463211 Chr.15: 43795370-43795371 on Build GRCh38
rs1003946517 Chr.15: 43795604-43795604 on Build GRCh38
rs750804762 Chr.15: 43795398-43795398 on Build GRCh38
rs769182206 Chr.15: 43795378-43795379 on Build GRCh38
rs886623121 Chr.15: 43795602-43795602 on Build GRCh38
rs576280150 Chr.15: 43795411-43795411 on Build GRCh38
rs768213727 Chr.15: 43795585-43795585 on Build GRCh38
rs990369286 Chr.15: 43795808-43795808 on Build GRCh38

View Full Product Details