Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51682287-51682823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCG3
Entrez Gene ID
29106
Gene Name
secretogranin III
Gene Aliases
SGIII
UniGene
Hs.232618
Gene Chromosome Location
Chr.15: 51681353-51721026 on Build GRCh38
Transcripts
NM_001165257.1 NM_013243.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2217086 c.83-1G>A p.? Chr.15: 51682516-51682516 on Build GRCh38
172080 c.103G>A p.E35K Chr.15: 51682537-51682537 on Build GRCh38
6354730 c.127A>C p.N43H Chr.15: 51682561-51682561 on Build GRCh38
5809455 c.107T>A p.L36* Chr.15: 51682541-51682541 on Build GRCh38
2217087 c.84C>T p.D28D Chr.15: 51682518-51682518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034100617 Chr.15: 51682647-51682647 on Build GRCh38
rs992923392 Chr.15: 51682641-51682641 on Build GRCh38
rs758511045 Chr.15: 51682589-51682589 on Build GRCh38
rs376881513 Chr.15: 51682535-51682535 on Build GRCh38
rs1043447073 Chr.15: 51682356-51682356 on Build GRCh38
rs957559700 Chr.15: 51682662-51682662 on Build GRCh38
rs766052539 Chr.15: 51682518-51682518 on Build GRCh38
rs953022836 Chr.15: 51682524-51682524 on Build GRCh38
rs932070156 Chr.15: 51682422-51682422 on Build GRCh38
rs911105801 Chr.15: 51682780-51682780 on Build GRCh38
rs3214015 Chr.15: 51682753-51682753 on Build GRCh38
rs913356545 Chr.15: 51682696-51682696 on Build GRCh38
rs759378241 Chr.15: 51682529-51682529 on Build GRCh38
rs913477548 Chr.15: 51682327-51682330 on Build GRCh38
rs371430571 Chr.15: 51682592-51682592 on Build GRCh38
rs990303931 Chr.15: 51682687-51682687 on Build GRCh38
rs748656544 Chr.15: 51682494-51682494 on Build GRCh38
rs896943928 Chr.15: 51682798-51682798 on Build GRCh38
rs539206227 Chr.15: 51682677-51682677 on Build GRCh38
rs946059189 Chr.15: 51682353-51682353 on Build GRCh38
rs3214014 Chr.15: 51682442-51682442 on Build GRCh38
rs758231284 Chr.15: 51682610-51682610 on Build GRCh38
rs868276225 Chr.15: 51682481-51682481 on Build GRCh38
rs761616667 Chr.15: 51682371-51682371 on Build GRCh38
rs767273685 Chr.15: 51682531-51682531 on Build GRCh38
rs555059984 Chr.15: 51682722-51682722 on Build GRCh38
rs769091338 Chr.15: 51682480-51682480 on Build GRCh38
rs755433274 Chr.15: 51682598-51682598 on Build GRCh38
rs781608195 Chr.15: 51682610-51682610 on Build GRCh38
rs551184809 Chr.15: 51682685-51682685 on Build GRCh38
rs142092548 Chr.15: 51682521-51682521 on Build GRCh38
rs748404149 Chr.15: 51682614-51682614 on Build GRCh38
rs145733878 Chr.15: 51682430-51682430 on Build GRCh38
rs960211136 Chr.15: 51682632-51682632 on Build GRCh38
rs573630982 Chr.15: 51682733-51682733 on Build GRCh38
rs762306048 Chr.15: 51682514-51682514 on Build GRCh38
rs992892080 Chr.15: 51682312-51682312 on Build GRCh38
rs780354609 Chr.15: 51682475-51682475 on Build GRCh38
rs2124591 Chr.15: 51682788-51682788 on Build GRCh38
rs747407243 Chr.15: 51682479-51682479 on Build GRCh38
rs931976328 Chr.15: 51682739-51682739 on Build GRCh38
rs2054166 Chr.15: 51682713-51682713 on Build GRCh38
rs1002174979 Chr.15: 51682533-51682533 on Build GRCh38
rs889640385 Chr.15: 51682397-51682397 on Build GRCh38
rs370323504 Chr.15: 51682420-51682420 on Build GRCh38
rs750517613 Chr.15: 51682551-51682551 on Build GRCh38
rs943856427 Chr.15: 51682783-51682783 on Build GRCh38
rs777054031 Chr.15: 51682489-51682489 on Build GRCh38
rs1050982377 Chr.15: 51682777-51682777 on Build GRCh38
rs946052913 Chr.15: 51682336-51682336 on Build GRCh38
rs769246290 Chr.15: 51682502-51682502 on Build GRCh38
rs567741373 Chr.15: 51682708-51682708 on Build GRCh38
rs528814382 Chr.15: 51682360-51682360 on Build GRCh38
rs569012710 Chr.15: 51682661-51682661 on Build GRCh38
rs936572688 Chr.15: 51682364-51682364 on Build GRCh38
rs577825066 Chr.15: 51682790-51682790 on Build GRCh38
rs529358969 Chr.15: 51682461-51682461 on Build GRCh38
rs41286490 Chr.15: 51682315-51682315 on Build GRCh38
rs752648628 Chr.15: 51682534-51682534 on Build GRCh38
rs751808851 Chr.15: 51682597-51682597 on Build GRCh38
rs367549313 Chr.15: 51682504-51682504 on Build GRCh38
rs758704434 Chr.15: 51682471-51682471 on Build GRCh38

View Full Product Details