Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65102360-65102859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBAP1L
Entrez Gene ID
390595
Gene Name
ubiquitin associated protein 1 like
Gene Aliases
UBAP-1L
UniGene
Hs.659256
Gene Chromosome Location
Chr.15: 65092760-65106656 on Build GRCh38
Transcripts
NM_001163692.1 XM_011521547.2 XM_017022172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237742 c.270G>A p.A90A Chr.15: 65102535-65102535 on Build GRCh38
4652615 c.312G>A p.R104R Chr.15: 65102493-65102493 on Build GRCh38
5546800 c.401_436del36 p.A134_L145del12 Chr.15: 65102369-65102404 on Build GRCh38
4655232 c.234C>A p.L78L Chr.15: 65102571-65102571 on Build GRCh38
6456166 c.168C>T p.A56A Chr.15: 65102637-65102637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056087667 Chr.15: 65102454-65102454 on Build GRCh38
rs577672362 Chr.15: 65102567-65102567 on Build GRCh38
rs773638566 Chr.15: 65102402-65102402 on Build GRCh38
rs572046906 Chr.15: 65102475-65102475 on Build GRCh38
rs373855851 Chr.15: 65102505-65102505 on Build GRCh38
rs534645693 Chr.15: 65102536-65102536 on Build GRCh38
rs896746560 Chr.15: 65102683-65102683 on Build GRCh38
rs747380054 Chr.15: 65102727-65102727 on Build GRCh38
rs796698167 Chr.15: 65102418-65102418 on Build GRCh38
rs553747951 Chr.15: 65102417-65102417 on Build GRCh38
rs894612393 Chr.15: 65102443-65102443 on Build GRCh38
rs953255312 Chr.15: 65102622-65102622 on Build GRCh38
rs1006783942 Chr.15: 65102659-65102659 on Build GRCh38
rs377016299 Chr.15: 65102510-65102510 on Build GRCh38
rs894685989 Chr.15: 65102457-65102457 on Build GRCh38
rs940753181 Chr.15: 65102406-65102406 on Build GRCh38
rs573549426 Chr.15: 65102763-65102763 on Build GRCh38
rs1056255774 Chr.15: 65102637-65102637 on Build GRCh38
rs907163372 Chr.15: 65102615-65102615 on Build GRCh38
rs898088702 Chr.15: 65102702-65102703 on Build GRCh38
rs997752042 Chr.15: 65102609-65102609 on Build GRCh38
rs543213099 Chr.15: 65102602-65102602 on Build GRCh38
rs913817567 Chr.15: 65102558-65102558 on Build GRCh38
rs563492310 Chr.15: 65102661-65102661 on Build GRCh38
rs928707736 Chr.15: 65102719-65102719 on Build GRCh38
rs1029970260 Chr.15: 65102760-65102760 on Build GRCh38
rs767623626 Chr.15: 65102404-65102404 on Build GRCh38
rs989435216 Chr.15: 65102553-65102553 on Build GRCh38
rs993670360 Chr.15: 65102753-65102753 on Build GRCh38
rs953412830 Chr.15: 65102386-65102386 on Build GRCh38
rs971704211 Chr.15: 65102507-65102507 on Build GRCh38
rs557531679 Chr.15: 65102542-65102542 on Build GRCh38
rs894674097 Chr.15: 65102651-65102651 on Build GRCh38
rs1016189377 Chr.15: 65102673-65102673 on Build GRCh38
rs961712188 Chr.15: 65102479-65102480 on Build GRCh38
rs756886790 Chr.15: 65102598-65102598 on Build GRCh38
rs947705293 Chr.15: 65102514-65102514 on Build GRCh38
rs768455516 Chr.15: 65102680-65102680 on Build GRCh38
rs1043389640 Chr.15: 65102526-65102526 on Build GRCh38
rs536936161 Chr.15: 65102412-65102412 on Build GRCh38
rs992029384 Chr.15: 65102791-65102791 on Build GRCh38
rs938798992 Chr.15: 65102625-65102625 on Build GRCh38
rs1028916194 Chr.15: 65102621-65102621 on Build GRCh38
rs962005533 Chr.15: 65102689-65102689 on Build GRCh38
rs893170315 Chr.15: 65102383-65102383 on Build GRCh38
rs928847933 Chr.15: 65102415-65102415 on Build GRCh38
rs960244487 Chr.15: 65102732-65102732 on Build GRCh38
rs954462322 Chr.15: 65102515-65102515 on Build GRCh38
rs984721844 Chr.15: 65102390-65102390 on Build GRCh38
rs946065640 Chr.15: 65102599-65102599 on Build GRCh38
rs954347665 Chr.15: 65102798-65102798 on Build GRCh38
rs866199632 Chr.15: 65102833-65102833 on Build GRCh38
rs113598604 Chr.15: 65102528-65102528 on Build GRCh38
rs972197091 Chr.15: 65102703-65102703 on Build GRCh38

View Full Product Details