Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27221178-27221691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM8
Entrez Gene ID
79831
Gene Name
lysine demethylase 8
Gene Aliases
JMJD5
UniGene
Hs.145717
Gene Chromosome Location
Chr.16: 27203477-27222305 on Build GRCh38
Transcripts
NM_001145348.1 NM_024773.2 XM_017023676.1 XM_017023677.1 XM_017023678.1 XM_017023679.1 XM_017023680.1

SNP Detail

SNP ID Chromosome Location
rs1023211278 Chr.16: 27221591-27221591 on Build GRCh38
rs1023516905 Chr.16: 27221307-27221307 on Build GRCh38
rs960250814 Chr.16: 27221284-27221284 on Build GRCh38
rs910410391 Chr.16: 27221487-27221487 on Build GRCh38
rs929494087 Chr.16: 27221502-27221502 on Build GRCh38
rs1001039864 Chr.16: 27221626-27221626 on Build GRCh38
rs556990824 Chr.16: 27221239-27221239 on Build GRCh38
rs568417896 Chr.16: 27221625-27221625 on Build GRCh38
rs998837781 Chr.16: 27221265-27221265 on Build GRCh38
rs112474931 Chr.16: 27221612-27221612 on Build GRCh38
rs533084480 Chr.16: 27221505-27221505 on Build GRCh38
rs902026283 Chr.16: 27221209-27221209 on Build GRCh38
rs1047838371 Chr.16: 27221529-27221529 on Build GRCh38
rs747004935 Chr.16: 27221639-27221639 on Build GRCh38
rs1015987947 Chr.16: 27221602-27221602 on Build GRCh38
rs1020695418 Chr.16: 27221430-27221430 on Build GRCh38
rs1034838419 Chr.16: 27221365-27221365 on Build GRCh38
rs867530732 Chr.16: 27221461-27221461 on Build GRCh38
rs78941332 Chr.16: 27221364-27221364 on Build GRCh38
rs998870439 Chr.16: 27221541-27221541 on Build GRCh38
rs1053074984 Chr.16: 27221560-27221560 on Build GRCh38
rs12924923 Chr.16: 27221530-27221530 on Build GRCh38
rs1051997325 Chr.16: 27221214-27221214 on Build GRCh38
rs1008095545 Chr.16: 27221230-27221230 on Build GRCh38
rs539239565 Chr.16: 27221244-27221244 on Build GRCh38
rs573930232 Chr.16: 27221208-27221208 on Build GRCh38
rs1023392451 Chr.16: 27221295-27221295 on Build GRCh38
rs751349323 Chr.16: 27221606-27221606 on Build GRCh38
rs1040515062 Chr.16: 27221200-27221200 on Build GRCh38
rs1006263524 Chr.16: 27221294-27221294 on Build GRCh38
rs943204177 Chr.16: 27221490-27221490 on Build GRCh38
rs765068981 Chr.16: 27221334-27221334 on Build GRCh38
rs960553216 Chr.16: 27221385-27221385 on Build GRCh38
rs963031086 Chr.16: 27221422-27221422 on Build GRCh38
rs970651669 Chr.16: 27221594-27221594 on Build GRCh38
rs964759833 Chr.16: 27221658-27221658 on Build GRCh38
rs368674442 Chr.16: 27221231-27221231 on Build GRCh38
rs1044377369 Chr.16: 27221570-27221570 on Build GRCh38
rs963060274 Chr.16: 27221607-27221607 on Build GRCh38
rs1134367 Chr.16: 27221562-27221562 on Build GRCh38
rs923449523 Chr.16: 27221495-27221495 on Build GRCh38
rs76839117 Chr.16: 27221448-27221448 on Build GRCh38
rs550602174 Chr.16: 27221664-27221664 on Build GRCh38
rs537057069 Chr.16: 27221201-27221201 on Build GRCh38
rs910222774 Chr.16: 27221462-27221462 on Build GRCh38
rs778515668 Chr.16: 27221424-27221424 on Build GRCh38
rs144602341 Chr.16: 27221654-27221654 on Build GRCh38
rs889208467 Chr.16: 27221215-27221215 on Build GRCh38
rs532230825 Chr.16: 27221665-27221665 on Build GRCh38
rs555705967 Chr.16: 27221427-27221427 on Build GRCh38
rs961652708 Chr.16: 27221360-27221360 on Build GRCh38
rs149257817 Chr.16: 27221460-27221460 on Build GRCh38
rs77656173 Chr.16: 27221243-27221243 on Build GRCh38
rs548433837 Chr.16: 27221600-27221600 on Build GRCh38
rs1134366 Chr.16: 27221420-27221420 on Build GRCh38
rs980965794 Chr.16: 27221375-27221375 on Build GRCh38
rs757493866 Chr.16: 27221631-27221631 on Build GRCh38
rs187645849 Chr.16: 27221283-27221283 on Build GRCh38
rs908955779 Chr.16: 27221391-27221391 on Build GRCh38
rs74016026 Chr.16: 27221584-27221584 on Build GRCh38
rs111318537 Chr.16: 27221306-27221306 on Build GRCh38
rs34179621 Chr.16: 27221459-27221459 on Build GRCh38
rs754630676 Chr.16: 27221363-27221363 on Build GRCh38
rs918957806 Chr.16: 27221634-27221634 on Build GRCh38

View Full Product Details