Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29994212-29994717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIRIP3
Entrez Gene ID
8479
Gene Name
HIRA interacting protein 3
Gene Aliases
-
UniGene
Hs.567370
Gene Chromosome Location
Chr.16: 29992321-29996096 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1478778 c.762G>T p.K254N Chr.16: 29994383-29994383 on Build GRCh38
4060063 c.587G>A p.S196N Chr.16: 29994558-29994558 on Build GRCh38
1609191 c.918G>A p.G306G Chr.16: 29994227-29994227 on Build GRCh38
7120649 c.660G>A p.L220L Chr.16: 29994485-29994485 on Build GRCh38
6144438 c.516G>T p.R172S Chr.16: 29994629-29994629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759140987 Chr.16: 29994396-29994398 on Build GRCh38
rs750398926 Chr.16: 29994273-29994273 on Build GRCh38
rs771218107 Chr.16: 29994316-29994317 on Build GRCh38
rs745680445 Chr.16: 29994662-29994662 on Build GRCh38
rs746372876 Chr.16: 29994448-29994448 on Build GRCh38
rs781726964 Chr.16: 29994632-29994632 on Build GRCh38
rs761017949 Chr.16: 29994265-29994265 on Build GRCh38
rs771607539 Chr.16: 29994595-29994595 on Build GRCh38
rs762748096 Chr.16: 29994509-29994509 on Build GRCh38
rs757729759 Chr.16: 29994325-29994325 on Build GRCh38
rs752336284 Chr.16: 29994400-29994400 on Build GRCh38
rs776409016 Chr.16: 29994694-29994694 on Build GRCh38
rs145090885 Chr.16: 29994526-29994526 on Build GRCh38
rs774327482 Chr.16: 29994588-29994588 on Build GRCh38
rs368475986 Chr.16: 29994475-29994475 on Build GRCh38
rs760470938 Chr.16: 29994529-29994529 on Build GRCh38
rs201678309 Chr.16: 29994528-29994528 on Build GRCh38
rs771510142 Chr.16: 29994239-29994239 on Build GRCh38
rs765064154 Chr.16: 29994352-29994352 on Build GRCh38
rs1026828525 Chr.16: 29994279-29994279 on Build GRCh38
rs374024211 Chr.16: 29994524-29994524 on Build GRCh38
rs778640919 Chr.16: 29994659-29994659 on Build GRCh38
rs141957940 Chr.16: 29994664-29994664 on Build GRCh38
rs1020485608 Chr.16: 29994671-29994671 on Build GRCh38
rs374834653 Chr.16: 29994629-29994629 on Build GRCh38
rs75636770 Chr.16: 29994333-29994333 on Build GRCh38
rs147563485 Chr.16: 29994635-29994635 on Build GRCh38
rs762940885 Chr.16: 29994386-29994386 on Build GRCh38
rs763792348 Chr.16: 29994268-29994268 on Build GRCh38
rs755071996 Chr.16: 29994560-29994560 on Build GRCh38
rs775074061 Chr.16: 29994603-29994603 on Build GRCh38
rs770674805 Chr.16: 29994586-29994586 on Build GRCh38
rs781192042 Chr.16: 29994564-29994564 on Build GRCh38
rs971701282 Chr.16: 29994540-29994540 on Build GRCh38
rs757678450 Chr.16: 29994431-29994431 on Build GRCh38
rs150516394 Chr.16: 29994541-29994541 on Build GRCh38
rs980042491 Chr.16: 29994585-29994585 on Build GRCh38
rs1039364913 Chr.16: 29994691-29994691 on Build GRCh38
rs370813183 Chr.16: 29994341-29994341 on Build GRCh38
rs761563175 Chr.16: 29994349-29994349 on Build GRCh38
rs748426043 Chr.16: 29994481-29994481 on Build GRCh38
rs371999310 Chr.16: 29994495-29994495 on Build GRCh38
rs749808213 Chr.16: 29994293-29994293 on Build GRCh38
rs371325743 Chr.16: 29994622-29994622 on Build GRCh38
rs781361380 Chr.16: 29994685-29994685 on Build GRCh38
rs773117037 Chr.16: 29994365-29994365 on Build GRCh38
rs77825050 Chr.16: 29994394-29994394 on Build GRCh38
rs139453333 Chr.16: 29994618-29994618 on Build GRCh38
rs760288323 Chr.16: 29994510-29994510 on Build GRCh38
rs747070567 Chr.16: 29994462-29994462 on Build GRCh38
rs774832653 Chr.16: 29994461-29994472 on Build GRCh38
rs771848295 Chr.16: 29994674-29994674 on Build GRCh38
rs758863702 Chr.16: 29994547-29994547 on Build GRCh38
rs758911005 Chr.16: 29994450-29994450 on Build GRCh38
rs754588606 Chr.16: 29994302-29994302 on Build GRCh38
rs765119373 Chr.16: 29994272-29994272 on Build GRCh38
rs753738900 Chr.16: 29994520-29994521 on Build GRCh38
rs748227621 Chr.16: 29994572-29994572 on Build GRCh38
rs779478911 Chr.16: 29994436-29994436 on Build GRCh38
rs774189583 Chr.16: 29994523-29994523 on Build GRCh38
rs775554415 Chr.16: 29994263-29994263 on Build GRCh38
rs201336607 Chr.16: 29994548-29994548 on Build GRCh38
rs771003688 Chr.16: 29994425-29994426 on Build GRCh38
rs937637691 Chr.16: 29994435-29994438 on Build GRCh38
rs561543731 Chr.16: 29994298-29994298 on Build GRCh38
rs770849284 Chr.16: 29994515-29994515 on Build GRCh38
rs899057250 Chr.16: 29994460-29994460 on Build GRCh38
rs768873561 Chr.16: 29994346-29994346 on Build GRCh38
rs896996095 Chr.16: 29994296-29994296 on Build GRCh38
rs927670509 Chr.16: 29994422-29994422 on Build GRCh38
rs929797609 Chr.16: 29994655-29994655 on Build GRCh38
rs774729176 Chr.16: 29994318-29994318 on Build GRCh38
rs752101617 Chr.16: 29994626-29994626 on Build GRCh38
rs749563175 Chr.16: 29994652-29994652 on Build GRCh38
rs750673744 Chr.16: 29994687-29994687 on Build GRCh38
rs763845357 Chr.16: 29994537-29994537 on Build GRCh38
rs757880563 Chr.16: 29994286-29994286 on Build GRCh38
rs573850419 Chr.16: 29994332-29994332 on Build GRCh38
rs771807710 Chr.16: 29994399-29994400 on Build GRCh38
rs1000638423 Chr.16: 29994476-29994476 on Build GRCh38
rs551205578 Chr.16: 29994502-29994502 on Build GRCh38
rs981819156 Chr.16: 29994413-29994413 on Build GRCh38
rs759453326 Chr.16: 29994593-29994593 on Build GRCh38
rs780440953 Chr.16: 29994457-29994457 on Build GRCh38
rs761851139 Chr.16: 29994551-29994553 on Build GRCh38
rs756214996 Chr.16: 29994638-29994638 on Build GRCh38
rs775591825 Chr.16: 29994679-29994679 on Build GRCh38
rs777990033 Chr.16: 29994573-29994573 on Build GRCh38
rs967763356 Chr.16: 29994439-29994439 on Build GRCh38
rs749619336 Chr.16: 29994323-29994323 on Build GRCh38
rs763351517 Chr.16: 29994623-29994623 on Build GRCh38
rs139422561 Chr.16: 29994389-29994389 on Build GRCh38
rs146230037 Chr.16: 29994300-29994300 on Build GRCh38
rs1019037181 Chr.16: 29994581-29994581 on Build GRCh38
rs766711969 Chr.16: 29994624-29994624 on Build GRCh38
rs554496381 Chr.16: 29994582-29994582 on Build GRCh38
rs763931620 Chr.16: 29994515-29994517 on Build GRCh38
rs746427887 Chr.16: 29994330-29994330 on Build GRCh38
rs202044584 Chr.16: 29994348-29994348 on Build GRCh38
rs780718925 Chr.16: 29994316-29994316 on Build GRCh38
rs747065079 Chr.16: 29994335-29994335 on Build GRCh38
rs1051446072 Chr.16: 29994339-29994339 on Build GRCh38
rs776889758 Chr.16: 29994469-29994469 on Build GRCh38
rs757667702 Chr.16: 29994271-29994271 on Build GRCh38
rs770750812 Chr.16: 29994658-29994658 on Build GRCh38
rs1009020499 Chr.16: 29994579-29994579 on Build GRCh38
rs200362133 Chr.16: 29994657-29994657 on Build GRCh38
rs1006950842 Chr.16: 29994424-29994424 on Build GRCh38
rs367564028 Chr.16: 29994604-29994604 on Build GRCh38
rs375385061 Chr.16: 29994650-29994650 on Build GRCh38
rs763791833 Chr.16: 29994621-29994621 on Build GRCh38
rs1033222920 Chr.16: 29994371-29994371 on Build GRCh38
rs200020004 Chr.16: 29994257-29994257 on Build GRCh38
rs183400729 Chr.16: 29994328-29994328 on Build GRCh38
rs775025103 Chr.16: 29994243-29994243 on Build GRCh38
rs751021230 Chr.16: 29994392-29994392 on Build GRCh38

View Full Product Details