Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30428842-30429387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTPP1
Entrez Gene ID
79077
Gene Name
dCTP pyrophosphatase 1
Gene Aliases
CDA03, RS21C6, XTP3TPA
UniGene
Hs.632191
Gene Chromosome Location
Chr.16: 30423698-30430052 on Build GRCh38
Transcripts
NM_024096.1 NR_134470.1 NR_134471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3112176 c.169C>A p.L57I Chr.16: 30429100-30429100 on Build GRCh38
1203175 c.197A>G p.E66G Chr.16: 30429072-30429072 on Build GRCh38
3794795 c.134G>A p.R45Q Chr.16: 30429135-30429135 on Build GRCh38
3957462 c.212+8G>A p.? Chr.16: 30429049-30429049 on Build GRCh38
1609199 c.117T>C p.A39A Chr.16: 30429152-30429152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772532200 Chr.16: 30428923-30428923 on Build GRCh38
rs570167074 Chr.16: 30428866-30428866 on Build GRCh38
rs755541131 Chr.16: 30429203-30429203 on Build GRCh38
rs562189592 Chr.16: 30429240-30429240 on Build GRCh38
rs1037287993 Chr.16: 30429137-30429137 on Build GRCh38
rs202012351 Chr.16: 30429198-30429198 on Build GRCh38
rs949228798 Chr.16: 30429019-30429019 on Build GRCh38
rs969141537 Chr.16: 30428909-30428909 on Build GRCh38
rs765328444 Chr.16: 30429010-30429010 on Build GRCh38
rs899717557 Chr.16: 30429339-30429339 on Build GRCh38
rs1041375512 Chr.16: 30429333-30429333 on Build GRCh38
rs553640729 Chr.16: 30428934-30428934 on Build GRCh38
rs759280402 Chr.16: 30429252-30429252 on Build GRCh38
rs931382152 Chr.16: 30429239-30429239 on Build GRCh38
rs996922568 Chr.16: 30429355-30429355 on Build GRCh38
rs755762163 Chr.16: 30429204-30429204 on Build GRCh38
rs574073883 Chr.16: 30428918-30428918 on Build GRCh38
rs189783642 Chr.16: 30428903-30428903 on Build GRCh38
rs551316027 Chr.16: 30429073-30429073 on Build GRCh38
rs762778828 Chr.16: 30429017-30429017 on Build GRCh38
rs763613465 Chr.16: 30429028-30429029 on Build GRCh38
rs369795540 Chr.16: 30429086-30429086 on Build GRCh38
rs1031927228 Chr.16: 30429359-30429359 on Build GRCh38
rs750311765 Chr.16: 30429015-30429015 on Build GRCh38
rs760150137 Chr.16: 30429154-30429154 on Build GRCh38
rs755224155 Chr.16: 30429239-30429239 on Build GRCh38
rs753237536 Chr.16: 30429332-30429332 on Build GRCh38
rs992902925 Chr.16: 30428965-30428965 on Build GRCh38
rs375231646 Chr.16: 30429006-30429006 on Build GRCh38
rs780583576 Chr.16: 30429185-30429185 on Build GRCh38
rs754842539 Chr.16: 30429057-30429057 on Build GRCh38
rs756906562 Chr.16: 30429174-30429174 on Build GRCh38
rs773357649 Chr.16: 30429129-30429129 on Build GRCh38
rs376311792 Chr.16: 30429210-30429210 on Build GRCh38
rs148049786 Chr.16: 30429246-30429246 on Build GRCh38
rs956071038 Chr.16: 30428945-30428945 on Build GRCh38
rs567045591 Chr.16: 30429258-30429258 on Build GRCh38
rs967691956 Chr.16: 30428957-30428957 on Build GRCh38
rs777328069 Chr.16: 30429077-30429077 on Build GRCh38
rs751563473 Chr.16: 30429039-30429039 on Build GRCh38
rs200063128 Chr.16: 30429097-30429097 on Build GRCh38
rs1031718310 Chr.16: 30429277-30429277 on Build GRCh38
rs780561744 Chr.16: 30428891-30428891 on Build GRCh38
rs371544904 Chr.16: 30429011-30429011 on Build GRCh38
rs766676952 Chr.16: 30429081-30429082 on Build GRCh38
rs781354874 Chr.16: 30429116-30429116 on Build GRCh38
rs763649498 Chr.16: 30429161-30429161 on Build GRCh38
rs897492409 Chr.16: 30429162-30429162 on Build GRCh38
rs767068046 Chr.16: 30429065-30429065 on Build GRCh38
rs181156575 Chr.16: 30429187-30429187 on Build GRCh38
rs750289625 Chr.16: 30429188-30429191 on Build GRCh38
rs747840395 Chr.16: 30428892-30428892 on Build GRCh38
rs555862703 Chr.16: 30428913-30428913 on Build GRCh38
rs755663953 Chr.16: 30428899-30428899 on Build GRCh38
rs926529675 Chr.16: 30429048-30429048 on Build GRCh38
rs780565157 Chr.16: 30429108-30429108 on Build GRCh38
rs760177847 Chr.16: 30428940-30428941 on Build GRCh38
rs956010620 Chr.16: 30429283-30429283 on Build GRCh38
rs982090496 Chr.16: 30429005-30429005 on Build GRCh38
rs753614679 Chr.16: 30429166-30429166 on Build GRCh38
rs1051510854 Chr.16: 30429177-30429177 on Build GRCh38
rs529287977 Chr.16: 30429241-30429241 on Build GRCh38
rs200511564 Chr.16: 30429075-30429075 on Build GRCh38
rs767408935 Chr.16: 30428928-30428928 on Build GRCh38
rs1045405361 Chr.16: 30429021-30429021 on Build GRCh38
rs747527404 Chr.16: 30429109-30429109 on Build GRCh38
rs763156020 Chr.16: 30429138-30429138 on Build GRCh38
rs753733074 Chr.16: 30429049-30429051 on Build GRCh38
rs368160112 Chr.16: 30429008-30429008 on Build GRCh38
rs943993187 Chr.16: 30429265-30429265 on Build GRCh38
rs759492520 Chr.16: 30429141-30429141 on Build GRCh38
rs752645482 Chr.16: 30429153-30429153 on Build GRCh38
rs778652830 Chr.16: 30429124-30429125 on Build GRCh38
rs748138253 Chr.16: 30429122-30429122 on Build GRCh38
rs770715518 Chr.16: 30429140-30429140 on Build GRCh38
rs769888382 Chr.16: 30429125-30429125 on Build GRCh38
rs954894829 Chr.16: 30428879-30428879 on Build GRCh38
rs368463553 Chr.16: 30429213-30429213 on Build GRCh38
rs535115949 Chr.16: 30428932-30428932 on Build GRCh38

View Full Product Details