Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53451568-53452090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBL2
Entrez Gene ID
5934
Gene Name
RB transcriptional corepressor like 2
Gene Aliases
P130, Rb2
UniGene
Hs.513609
Gene Chromosome Location
Chr.16: 53434420-53491648 on Build GRCh38
Transcripts
NM_001323608.1 NM_001323609.1 NM_001323610.1 NM_001323611.1 NM_005611.3 XM_011523253.2 XM_017023513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4720630 c.738T>C p.R246R Chr.16: 53451803-53451803 on Build GRCh38
5772467 c.682C>A p.L228M Chr.16: 53451747-53451747 on Build GRCh38
6222237 c.766+6T>G p.? Chr.16: 53451837-53451837 on Build GRCh38
3672273 c.729T>C p.C243C Chr.16: 53451794-53451794 on Build GRCh38
6246913 c.690G>T p.L230L Chr.16: 53451755-53451755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952011012 Chr.16: 53452066-53452066 on Build GRCh38
rs751787000 Chr.16: 53451867-53451867 on Build GRCh38
rs540728293 Chr.16: 53451679-53451679 on Build GRCh38
rs771248856 Chr.16: 53451657-53451657 on Build GRCh38
rs972060336 Chr.16: 53451999-53451999 on Build GRCh38
rs901517651 Chr.16: 53451822-53451822 on Build GRCh38
rs771667410 Chr.16: 53451818-53451818 on Build GRCh38
rs202133927 Chr.16: 53451868-53451868 on Build GRCh38
rs776478588 Chr.16: 53451837-53451837 on Build GRCh38
rs201324508 Chr.16: 53451740-53451740 on Build GRCh38
rs754876750 Chr.16: 53451731-53451731 on Build GRCh38
rs981292114 Chr.16: 53451633-53451633 on Build GRCh38
rs753729896 Chr.16: 53451730-53451730 on Build GRCh38
rs777219034 Chr.16: 53451663-53451663 on Build GRCh38
rs930432323 Chr.16: 53451681-53451681 on Build GRCh38
rs766148068 Chr.16: 53451727-53451727 on Build GRCh38
rs772276831 Chr.16: 53452036-53452036 on Build GRCh38
rs368582246 Chr.16: 53451812-53451812 on Build GRCh38
rs199920381 Chr.16: 53451824-53451824 on Build GRCh38
rs1037380313 Chr.16: 53451641-53451641 on Build GRCh38
rs778164259 Chr.16: 53451653-53451653 on Build GRCh38
rs560609018 Chr.16: 53451710-53451710 on Build GRCh38
rs1002290061 Chr.16: 53451943-53451943 on Build GRCh38
rs760032937 Chr.16: 53451665-53451665 on Build GRCh38
rs770323707 Chr.16: 53451802-53451802 on Build GRCh38
rs766090408 Chr.16: 53451835-53451835 on Build GRCh38
rs146824919 Chr.16: 53451996-53451996 on Build GRCh38
rs756049355 Chr.16: 53451975-53451975 on Build GRCh38
rs370735742 Chr.16: 53451789-53451789 on Build GRCh38
rs1002032045 Chr.16: 53451668-53451668 on Build GRCh38
rs910266767 Chr.16: 53451760-53451760 on Build GRCh38
rs927931876 Chr.16: 53451638-53451638 on Build GRCh38
rs761617948 Chr.16: 53451669-53451669 on Build GRCh38
rs1011469971 Chr.16: 53451897-53451897 on Build GRCh38
rs200891640 Chr.16: 53451742-53451742 on Build GRCh38
rs771282059 Chr.16: 53451632-53451632 on Build GRCh38
rs764106214 Chr.16: 53451866-53451866 on Build GRCh38
rs765195808 Chr.16: 53451855-53451855 on Build GRCh38
rs777029960 Chr.16: 53451658-53451658 on Build GRCh38
rs927888074 Chr.16: 53452063-53452063 on Build GRCh38
rs760344148 Chr.16: 53451826-53451826 on Build GRCh38
rs910690102 Chr.16: 53451924-53451924 on Build GRCh38
rs374075321 Chr.16: 53451848-53451848 on Build GRCh38
rs181633989 Chr.16: 53451667-53451667 on Build GRCh38
rs752811169 Chr.16: 53451862-53451862 on Build GRCh38
rs75138965 Chr.16: 53452035-53452035 on Build GRCh38
rs533781352 Chr.16: 53451919-53451919 on Build GRCh38
rs767256003 Chr.16: 53451699-53451699 on Build GRCh38
rs548473274 Chr.16: 53451874-53451874 on Build GRCh38
rs760577001 Chr.16: 53451720-53451720 on Build GRCh38
rs774053725 Chr.16: 53451813-53451813 on Build GRCh38
rs1046061437 Chr.16: 53452039-53452039 on Build GRCh38
rs1033432463 Chr.16: 53451696-53451696 on Build GRCh38
rs778006189 Chr.16: 53451743-53451743 on Build GRCh38
rs747188044 Chr.16: 53451746-53451746 on Build GRCh38
rs973538766 Chr.16: 53452028-53452028 on Build GRCh38
rs1022764214 Chr.16: 53451606-53451606 on Build GRCh38
rs1035137593 Chr.16: 53451978-53451978 on Build GRCh38
rs969512349 Chr.16: 53451630-53451637 on Build GRCh38
rs760773957 Chr.16: 53451852-53451852 on Build GRCh38
rs547077641 Chr.16: 53451942-53451942 on Build GRCh38
rs758498439 Chr.16: 53451863-53451863 on Build GRCh38
rs777484546 Chr.16: 53452031-53452031 on Build GRCh38
rs967807078 Chr.16: 53451807-53451807 on Build GRCh38
rs781556948 Chr.16: 53451782-53451782 on Build GRCh38
rs747631806 Chr.16: 53451817-53451817 on Build GRCh38
rs928967829 Chr.16: 53451647-53451647 on Build GRCh38
rs775497273 Chr.16: 53451655-53451656 on Build GRCh38
rs757578853 Chr.16: 53451781-53451781 on Build GRCh38
rs561287810 Chr.16: 53451646-53451646 on Build GRCh38
rs769479574 Chr.16: 53451961-53451961 on Build GRCh38
rs16952254 Chr.16: 53451888-53451888 on Build GRCh38
rs375056854 Chr.16: 53451957-53451957 on Build GRCh38
rs1011083548 Chr.16: 53451595-53451595 on Build GRCh38

View Full Product Details