Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53637984-53638525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPGRIP1L
Entrez Gene ID
23322
Gene Name
RPGRIP1 like
Gene Aliases
CORS3, FTM, JBTS7, MKS5, NPHP8, PPP1R134
UniGene
Hs.298382
Gene Chromosome Location
Chr.16: 53599239-53703934 on Build GRCh38
Transcripts
XM_005255867.1 XM_005255868.1 XM_011522970.1 XM_011522971.2 XM_011522973.2 XM_017023094.1 XM_017023095.1 XM_017023096.1 XM_017023098.1 XM_017023099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971821 c.2997A>G p.S999S Chr.16: 53638373-53638373 on Build GRCh38
7304922 c.3033T>A p.N1011K Chr.16: 53638337-53638337 on Build GRCh38
6769132 c.3060G>T p.K1020N Chr.16: 53638310-53638310 on Build GRCh38
5384992 c.2959-1G>A p.? Chr.16: 53638412-53638412 on Build GRCh38
3510257 c.2959-1G>T p.? Chr.16: 53638412-53638412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762752860 Chr.16: 53638409-53638409 on Build GRCh38
rs888005204 Chr.16: 53638007-53638007 on Build GRCh38
rs536439986 Chr.16: 53638050-53638050 on Build GRCh38
rs7203525 Chr.16: 53638443-53638443 on Build GRCh38
rs750665065 Chr.16: 53638348-53638348 on Build GRCh38
rs1026056729 Chr.16: 53638233-53638233 on Build GRCh38
rs574430009 Chr.16: 53638360-53638360 on Build GRCh38
rs34076982 Chr.16: 53638312-53638313 on Build GRCh38
rs774428262 Chr.16: 53638422-53638422 on Build GRCh38
rs531008300 Chr.16: 53638492-53638492 on Build GRCh38
rs865856552 Chr.16: 53638402-53638402 on Build GRCh38
rs775539641 Chr.16: 53638313-53638313 on Build GRCh38
rs937851840 Chr.16: 53638171-53638171 on Build GRCh38
rs780310164 Chr.16: 53638357-53638357 on Build GRCh38
rs764920843 Chr.16: 53638309-53638309 on Build GRCh38
rs756538968 Chr.16: 53638354-53638354 on Build GRCh38
rs375794046 Chr.16: 53638269-53638269 on Build GRCh38
rs561339531 Chr.16: 53638499-53638499 on Build GRCh38
rs909371199 Chr.16: 53638465-53638465 on Build GRCh38
rs538524550 Chr.16: 53638115-53638115 on Build GRCh38
rs11863101 Chr.16: 53638426-53638426 on Build GRCh38
rs1032187766 Chr.16: 53638322-53638322 on Build GRCh38
rs752076060 Chr.16: 53638394-53638396 on Build GRCh38
rs745480425 Chr.16: 53638392-53638392 on Build GRCh38
rs776259551 Chr.16: 53638281-53638281 on Build GRCh38
rs538093589 Chr.16: 53638121-53638121 on Build GRCh38
rs775171959 Chr.16: 53638407-53638407 on Build GRCh38
rs779454020 Chr.16: 53638459-53638459 on Build GRCh38
rs191331892 Chr.16: 53638029-53638029 on Build GRCh38
rs757343133 Chr.16: 53638330-53638330 on Build GRCh38
rs190689604 Chr.16: 53638442-53638442 on Build GRCh38
rs558459915 Chr.16: 53638118-53638118 on Build GRCh38
rs554791000 Chr.16: 53638098-53638099 on Build GRCh38
rs570158682 Chr.16: 53638159-53638159 on Build GRCh38
rs769448029 Chr.16: 53638404-53638404 on Build GRCh38
rs767686945 Chr.16: 53638432-53638432 on Build GRCh38
rs537109143 Chr.16: 53638230-53638230 on Build GRCh38
rs757949061 Chr.16: 53638244-53638244 on Build GRCh38
rs750487146 Chr.16: 53638436-53638436 on Build GRCh38
rs767718757 Chr.16: 53638445-53638447 on Build GRCh38
rs749613626 Chr.16: 53638359-53638359 on Build GRCh38
rs767018622 Chr.16: 53638378-53638378 on Build GRCh38
rs540344173 Chr.16: 53638416-53638416 on Build GRCh38
rs762829599 Chr.16: 53638319-53638319 on Build GRCh38
rs1056261797 Chr.16: 53638182-53638182 on Build GRCh38
rs754156397 Chr.16: 53638445-53638445 on Build GRCh38
rs767920573 Chr.16: 53638346-53638346 on Build GRCh38
rs764044965 Chr.16: 53638320-53638320 on Build GRCh38
rs769296095 Chr.16: 53638095-53638095 on Build GRCh38
rs746586531 Chr.16: 53638363-53638363 on Build GRCh38
rs946702867 Chr.16: 53638134-53638134 on Build GRCh38
rs1043665062 Chr.16: 53638142-53638142 on Build GRCh38
rs776313021 Chr.16: 53638378-53638378 on Build GRCh38
rs770414663 Chr.16: 53638374-53638374 on Build GRCh38
rs940865058 Chr.16: 53638466-53638466 on Build GRCh38
rs988118018 Chr.16: 53638124-53638124 on Build GRCh38
rs554467783 Chr.16: 53638211-53638211 on Build GRCh38
rs984400687 Chr.16: 53638028-53638028 on Build GRCh38
rs369829038 Chr.16: 53638283-53638283 on Build GRCh38
rs975599188 Chr.16: 53638062-53638062 on Build GRCh38
rs148279382 Chr.16: 53638120-53638120 on Build GRCh38
rs1036646636 Chr.16: 53638070-53638070 on Build GRCh38
rs779323809 Chr.16: 53638362-53638362 on Build GRCh38
rs965006608 Chr.16: 53638366-53638366 on Build GRCh38
rs183588912 Chr.16: 53638055-53638055 on Build GRCh38
rs368259444 Chr.16: 53638060-53638063 on Build GRCh38
rs546555847 Chr.16: 53638048-53638048 on Build GRCh38
rs186619460 Chr.16: 53638247-53638247 on Build GRCh38
rs536357397 Chr.16: 53638037-53638037 on Build GRCh38
rs896483389 Chr.16: 53638085-53638085 on Build GRCh38
rs951623252 Chr.16: 53638008-53638008 on Build GRCh38
rs183233028 Chr.16: 53638496-53638496 on Build GRCh38
rs199542430 Chr.16: 53638450-53638450 on Build GRCh38
rs913873824 Chr.16: 53638127-53638127 on Build GRCh38
rs770716076 Chr.16: 53638386-53638386 on Build GRCh38
rs759986075 Chr.16: 53638403-53638405 on Build GRCh38

View Full Product Details