Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56192127-56192632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434H168
Entrez Gene ID
26077
Gene Name
uncharacterized LOC26077
Gene Aliases
-
UniGene
Hs.721567
Gene Chromosome Location
Chr.16: 56192617-56194525 on Build GRCh38
Transcripts
NR_026889.1 NM_020988.2 NM_138736.2 XM_011523003.2

Gene Symbol
GNAO1
Entrez Gene ID
2775
Gene Name
G protein subunit alpha o1
Gene Aliases
EIEE17, G-ALPHA-o, GNAO, HLA-DQB1
UniGene
Hs.644524
Gene Chromosome Location
Chr.16: 56191339-56357444 on Build GRCh38
Transcripts
NR_026889.1 NM_020988.2 NM_138736.2 XM_011523003.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4525720 c.134G>A p.G45E Chr.16: 56192589-56192589 on Build GRCh38
6532283 c.9T>C p.C3C Chr.16: 56192244-56192244 on Build GRCh38
3969598 c.84C>G p.I28M Chr.16: 56192319-56192319 on Build GRCh38
6031883 c.12T>G p.T4T Chr.16: 56192247-56192247 on Build GRCh38
1208396 c.35C>T p.A12V Chr.16: 56192270-56192270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049369452 Chr.16: 56192510-56192510 on Build GRCh38
rs757827374 Chr.16: 56192228-56192228 on Build GRCh38
rs370572969 Chr.16: 56192403-56192403 on Build GRCh38
rs563968251 Chr.16: 56192375-56192375 on Build GRCh38
rs751695021 Chr.16: 56192391-56192391 on Build GRCh38
rs774575085 Chr.16: 56192556-56192559 on Build GRCh38
rs372787855 Chr.16: 56192361-56192361 on Build GRCh38
rs752677340 Chr.16: 56192204-56192204 on Build GRCh38
rs900646926 Chr.16: 56192496-56192496 on Build GRCh38
rs775212759 Chr.16: 56192569-56192569 on Build GRCh38
rs376241293 Chr.16: 56192209-56192209 on Build GRCh38
rs912013571 Chr.16: 56192445-56192445 on Build GRCh38
rs776729306 Chr.16: 56192552-56192552 on Build GRCh38
rs944775964 Chr.16: 56192456-56192456 on Build GRCh38
rs781694164 Chr.16: 56192392-56192392 on Build GRCh38
rs760689463 Chr.16: 56192371-56192371 on Build GRCh38
rs886041715 Chr.16: 56192353-56192353 on Build GRCh38
rs764320976 Chr.16: 56192526-56192526 on Build GRCh38
rs182812404 Chr.16: 56192310-56192310 on Build GRCh38
rs776531651 Chr.16: 56192372-56192372 on Build GRCh38
rs375695166 Chr.16: 56192379-56192380 on Build GRCh38
rs1020237471 Chr.16: 56192378-56192378 on Build GRCh38
rs765324424 Chr.16: 56192379-56192379 on Build GRCh38
rs886041766 Chr.16: 56192574-56192574 on Build GRCh38
rs367670802 Chr.16: 56192516-56192516 on Build GRCh38
rs769634112 Chr.16: 56192560-56192560 on Build GRCh38
rs755224987 Chr.16: 56192190-56192190 on Build GRCh38
rs763223472 Chr.16: 56192600-56192600 on Build GRCh38
rs886672142 Chr.16: 56192522-56192522 on Build GRCh38
rs552528169 Chr.16: 56192507-56192507 on Build GRCh38
rs921791681 Chr.16: 56192424-56192424 on Build GRCh38
rs201977449 Chr.16: 56192544-56192544 on Build GRCh38
rs374312361 Chr.16: 56192367-56192367 on Build GRCh38
rs774085068 Chr.16: 56192347-56192347 on Build GRCh38
rs369237309 Chr.16: 56192235-56192235 on Build GRCh38
rs916381009 Chr.16: 56192233-56192233 on Build GRCh38
rs766825059 Chr.16: 56192385-56192385 on Build GRCh38
rs777412763 Chr.16: 56192531-56192531 on Build GRCh38
rs139334934 Chr.16: 56192262-56192262 on Build GRCh38
rs778082773 Chr.16: 56192212-56192212 on Build GRCh38
rs763242415 Chr.16: 56192381-56192418 on Build GRCh38
rs759192540 Chr.16: 56192557-56192557 on Build GRCh38
rs752976425 Chr.16: 56192380-56192380 on Build GRCh38
rs771692814 Chr.16: 56192530-56192530 on Build GRCh38
rs765531417 Chr.16: 56192201-56192201 on Build GRCh38
rs869312939 Chr.16: 56192588-56192588 on Build GRCh38
rs1004538154 Chr.16: 56192160-56192160 on Build GRCh38
rs1034791551 Chr.16: 56192162-56192162 on Build GRCh38
rs1039614887 Chr.16: 56192495-56192495 on Build GRCh38
rs113550994 Chr.16: 56192405-56192405 on Build GRCh38
rs770947758 Chr.16: 56192551-56192551 on Build GRCh38
rs1014492184 Chr.16: 56192167-56192167 on Build GRCh38
rs530274427 Chr.16: 56192149-56192149 on Build GRCh38
rs964496051 Chr.16: 56192402-56192402 on Build GRCh38
rs747516640 Chr.16: 56192214-56192214 on Build GRCh38
rs570729468 Chr.16: 56192521-56192521 on Build GRCh38
rs762872543 Chr.16: 56192385-56192385 on Build GRCh38
rs747565910 Chr.16: 56192352-56192352 on Build GRCh38
rs974587597 Chr.16: 56192408-56192408 on Build GRCh38
rs978738893 Chr.16: 56192386-56192386 on Build GRCh38
rs764230140 Chr.16: 56192388-56192388 on Build GRCh38
rs377201271 Chr.16: 56192529-56192529 on Build GRCh38
rs377122941 Chr.16: 56192398-56192398 on Build GRCh38

View Full Product Details