Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 61652333-61652837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH8
Entrez Gene ID
1006
Gene Name
cadherin 8
Gene Aliases
Nbla04261
UniGene
Hs.368322
Gene Chromosome Location
Chr.16: 61640435-62036835 on Build GRCh38
Transcripts
NM_001796.4 XM_005255760.3 XM_017022829.1

SNP Detail

SNP ID Chromosome Location
rs1041808035 Chr.16: 61652600-61652600 on Build GRCh38
rs902129195 Chr.16: 61652714-61652714 on Build GRCh38
rs953559303 Chr.16: 61652516-61652516 on Build GRCh38
rs143472529 Chr.16: 61652613-61652613 on Build GRCh38
rs1056065480 Chr.16: 61652652-61652652 on Build GRCh38
rs952067859 Chr.16: 61652786-61652786 on Build GRCh38
rs575315613 Chr.16: 61652719-61652719 on Build GRCh38
rs534420775 Chr.16: 61652572-61652572 on Build GRCh38
rs991381449 Chr.16: 61652559-61652559 on Build GRCh38
rs555449473 Chr.16: 61652669-61652669 on Build GRCh38
rs935771641 Chr.16: 61652658-61652658 on Build GRCh38
rs544478903 Chr.16: 61652746-61652746 on Build GRCh38
rs917122470 Chr.16: 61652582-61652582 on Build GRCh38
rs997208972 Chr.16: 61652782-61652782 on Build GRCh38
rs796536234 Chr.16: 61652711-61652711 on Build GRCh38
rs748623566 Chr.16: 61652466-61652466 on Build GRCh38
rs949753355 Chr.16: 61652592-61652592 on Build GRCh38
rs902846079 Chr.16: 61652619-61652619 on Build GRCh38
rs775031217 Chr.16: 61652455-61652455 on Build GRCh38
rs1019934067 Chr.16: 61652686-61652686 on Build GRCh38
rs16963765 Chr.16: 61652504-61652504 on Build GRCh38
rs1054193034 Chr.16: 61652660-61652660 on Build GRCh38
rs574699097 Chr.16: 61652793-61652793 on Build GRCh38
rs889299964 Chr.16: 61652675-61652675 on Build GRCh38
rs760846448 Chr.16: 61652790-61652790 on Build GRCh38
rs553014260 Chr.16: 61652511-61652511 on Build GRCh38
rs905306491 Chr.16: 61652622-61652622 on Build GRCh38
rs919263570 Chr.16: 61652591-61652591 on Build GRCh38
rs937185465 Chr.16: 61652639-61652639 on Build GRCh38
rs1019178694 Chr.16: 61652406-61652406 on Build GRCh38
rs953506252 Chr.16: 61652361-61652361 on Build GRCh38
rs937148881 Chr.16: 61652542-61652542 on Build GRCh38
rs551793613 Chr.16: 61652479-61652479 on Build GRCh38
rs564239163 Chr.16: 61652791-61652791 on Build GRCh38
rs1019968348 Chr.16: 61652441-61652441 on Build GRCh38
rs150462483 Chr.16: 61652792-61652792 on Build GRCh38
rs4131634 Chr.16: 61652495-61652495 on Build GRCh38
rs909748739 Chr.16: 61652525-61652525 on Build GRCh38
rs141235665 Chr.16: 61652580-61652580 on Build GRCh38
rs972005471 Chr.16: 61652583-61652583 on Build GRCh38
rs1019216127 Chr.16: 61652685-61652685 on Build GRCh38
rs752275245 Chr.16: 61652748-61652748 on Build GRCh38
rs5002375 Chr.16: 61652377-61652377 on Build GRCh38
rs1007693637 Chr.16: 61652677-61652677 on Build GRCh38
rs966037547 Chr.16: 61652415-61652415 on Build GRCh38
rs1026759634 Chr.16: 61652768-61652768 on Build GRCh38

View Full Product Details