Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86530009-86530502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTHFSD
Entrez Gene ID
64779
Gene Name
methenyltetrahydrofolate synthetase domain containing
Gene Aliases
-
UniGene
Hs.343627
Gene Chromosome Location
Chr.16: 86530176-86555235 on Build GRCh38
Transcripts
NM_001159377.1 NM_001159378.1 NM_001159379.1 NM_001159380.1 NM_022764.2 NR_027489.1 NR_027490.1 XM_005256101.1 XM_005256105.1 XM_005256106.1 XM_006721247.3 XM_011523280.2 XM_011523285.1 XM_011523286.1 XM_011523287.1 XM_011523288.1 XM_011523289.1 XM_017023571.1 XM_017023572.1 XM_017023573.1 XM_017023574.1

SNP Detail

SNP ID Chromosome Location
rs972658962 Chr.16: 86530408-86530408 on Build GRCh38
rs894893974 Chr.16: 86530261-86530261 on Build GRCh38
rs1037997876 Chr.16: 86530190-86530190 on Build GRCh38
rs778075830 Chr.16: 86530110-86530110 on Build GRCh38
rs569251499 Chr.16: 86530387-86530387 on Build GRCh38
rs538608364 Chr.16: 86530210-86530210 on Build GRCh38
rs1008896636 Chr.16: 86530314-86530314 on Build GRCh38
rs1054853036 Chr.16: 86530252-86530252 on Build GRCh38
rs200175812 Chr.16: 86530482-86530482 on Build GRCh38
rs965214451 Chr.16: 86530339-86530339 on Build GRCh38
rs111992926 Chr.16: 86530398-86530398 on Build GRCh38
rs75554489 Chr.16: 86530058-86530058 on Build GRCh38
rs949148936 Chr.16: 86530133-86530133 on Build GRCh38
rs556326511 Chr.16: 86530137-86530137 on Build GRCh38
rs554069145 Chr.16: 86530376-86530376 on Build GRCh38
rs916389167 Chr.16: 86530433-86530433 on Build GRCh38
rs760581815 Chr.16: 86530460-86530460 on Build GRCh38
rs899105608 Chr.16: 86530211-86530211 on Build GRCh38
rs970060263 Chr.16: 86530436-86530436 on Build GRCh38
rs763440655 Chr.16: 86530234-86530234 on Build GRCh38
rs758893905 Chr.16: 86530351-86530352 on Build GRCh38
rs926240671 Chr.16: 86530074-86530074 on Build GRCh38
rs532887131 Chr.16: 86530237-86530237 on Build GRCh38
rs578152732 Chr.16: 86530269-86530269 on Build GRCh38
rs115577033 Chr.16: 86530421-86530421 on Build GRCh38
rs8776 Chr.16: 86530288-86530288 on Build GRCh38
rs758620321 Chr.16: 86530073-86530073 on Build GRCh38
rs569780535 Chr.16: 86530173-86530173 on Build GRCh38
rs768952461 Chr.16: 86530273-86530273 on Build GRCh38
rs571425511 Chr.16: 86530062-86530062 on Build GRCh38
rs913609819 Chr.16: 86530402-86530402 on Build GRCh38
rs999801718 Chr.16: 86530316-86530316 on Build GRCh38
rs949185082 Chr.16: 86530435-86530435 on Build GRCh38
rs181608895 Chr.16: 86530118-86530118 on Build GRCh38
rs937822511 Chr.16: 86530461-86530461 on Build GRCh38
rs979387669 Chr.16: 86530063-86530063 on Build GRCh38
rs1017694441 Chr.16: 86530336-86530336 on Build GRCh38
rs974861397 Chr.16: 86530122-86530122 on Build GRCh38
rs531630901 Chr.16: 86530052-86530052 on Build GRCh38
rs987831562 Chr.16: 86530114-86530114 on Build GRCh38
rs562609137 Chr.16: 86530471-86530471 on Build GRCh38
rs547256800 Chr.16: 86530069-86530069 on Build GRCh38
rs970385898 Chr.16: 86530406-86530406 on Build GRCh38
rs1046262096 Chr.16: 86530156-86530156 on Build GRCh38
rs1009009704 Chr.16: 86530051-86530051 on Build GRCh38
rs370587118 Chr.16: 86530119-86530119 on Build GRCh38
rs536327837 Chr.16: 86530135-86530135 on Build GRCh38
rs926410879 Chr.16: 86530160-86530160 on Build GRCh38
rs16941417 Chr.16: 86530077-86530077 on Build GRCh38
rs987544247 Chr.16: 86530397-86530397 on Build GRCh38
rs868570882 Chr.16: 86530127-86530127 on Build GRCh38
rs954820830 Chr.16: 86530040-86530040 on Build GRCh38
rs1000388796 Chr.16: 86530345-86530345 on Build GRCh38
rs890536058 Chr.16: 86530294-86530294 on Build GRCh38
rs919852176 Chr.16: 86530414-86530414 on Build GRCh38
rs373187986 Chr.16: 86530302-86530302 on Build GRCh38
rs376493913 Chr.16: 86530139-86530139 on Build GRCh38
rs369576867 Chr.16: 86530033-86530033 on Build GRCh38
rs144409979 Chr.16: 86530236-86530236 on Build GRCh38
rs949142430 Chr.16: 86530283-86530283 on Build GRCh38
rs761886796 Chr.16: 86530318-86530318 on Build GRCh38

View Full Product Details