Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 6996735-6997229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12
Entrez Gene ID
239
Gene Name
arachidonate 12-lipoxygenase, 12S type
Gene Aliases
12-LOX, 12S-LOX, LOG12
UniGene
Hs.654431
Gene Chromosome Location
Chr.17: 6993791-7010736 on Build GRCh38
Transcripts
NM_000697.2 XM_011523780.2 NR_040089.1

Gene Symbol
ALOX12-AS1
Entrez Gene ID
100506713
Gene Name
ALOX12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.422967
Gene Chromosome Location
Chr.17: 6985123-7012334 on Build GRCh38
Transcripts
NM_000697.2 XM_011523780.2 NR_040089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
39826 c.263C>T p.A88V Chr.17: 6996953-6996953 on Build GRCh38
5387161 c.192G>A p.R64R Chr.17: 6996882-6996882 on Build GRCh38
5747803 c.181C>T p.Q61* Chr.17: 6996871-6996871 on Build GRCh38
437255 c.309G>C p.E103D Chr.17: 6996999-6996999 on Build GRCh38
2975936 c.170T>C p.L57S Chr.17: 6996860-6996860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41388649 Chr.17: 6996783-6996783 on Build GRCh38
rs1036200284 Chr.17: 6997053-6997053 on Build GRCh38
rs763628706 Chr.17: 6996798-6996798 on Build GRCh38
rs1022375923 Chr.17: 6997145-6997145 on Build GRCh38
rs752933794 Chr.17: 6996993-6996993 on Build GRCh38
rs867235749 Chr.17: 6997084-6997084 on Build GRCh38
rs898001340 Chr.17: 6996769-6996769 on Build GRCh38
rs770436423 Chr.17: 6997048-6997048 on Build GRCh38
rs539504455 Chr.17: 6996764-6996764 on Build GRCh38
rs751995761 Chr.17: 6996972-6996972 on Build GRCh38
rs991905210 Chr.17: 6997065-6997065 on Build GRCh38
rs766775354 Chr.17: 6996962-6996962 on Build GRCh38
rs995628050 Chr.17: 6997025-6997025 on Build GRCh38
rs777774835 Chr.17: 6996858-6996858 on Build GRCh38
rs966014348 Chr.17: 6996824-6996826 on Build GRCh38
rs1018806893 Chr.17: 6996805-6996805 on Build GRCh38
rs771903250 Chr.17: 6996877-6996877 on Build GRCh38
rs764789807 Chr.17: 6996811-6996811 on Build GRCh38
rs934428232 Chr.17: 6996914-6996914 on Build GRCh38
rs312468 Chr.17: 6997064-6997064 on Build GRCh38
rs1035681852 Chr.17: 6996794-6996794 on Build GRCh38
rs969368935 Chr.17: 6996990-6996990 on Build GRCh38
rs775400651 Chr.17: 6996879-6996879 on Build GRCh38
rs56342631 Chr.17: 6996801-6996801 on Build GRCh38
rs369870849 Chr.17: 6996978-6996978 on Build GRCh38
rs771917931 Chr.17: 6996785-6996785 on Build GRCh38
rs533384960 Chr.17: 6997090-6997090 on Build GRCh38
rs1027363840 Chr.17: 6996786-6996786 on Build GRCh38
rs11571329 Chr.17: 6997040-6997040 on Build GRCh38
rs978580072 Chr.17: 6997070-6997070 on Build GRCh38
rs147402591 Chr.17: 6996929-6996929 on Build GRCh38
rs756736044 Chr.17: 6997015-6997015 on Build GRCh38
rs566663426 Chr.17: 6997002-6997002 on Build GRCh38
rs781363418 Chr.17: 6996838-6996838 on Build GRCh38
rs746746196 Chr.17: 6996882-6996882 on Build GRCh38
rs1022872611 Chr.17: 6997066-6997066 on Build GRCh38
rs774744541 Chr.17: 6997054-6997054 on Build GRCh38
rs372860181 Chr.17: 6996995-6996995 on Build GRCh38
rs375708757 Chr.17: 6996834-6996834 on Build GRCh38
rs751857816 Chr.17: 6996832-6996832 on Build GRCh38
rs768721604 Chr.17: 6997099-6997099 on Build GRCh38
rs998747369 Chr.17: 6997082-6997082 on Build GRCh38
rs139609698 Chr.17: 6997010-6997010 on Build GRCh38
rs750978849 Chr.17: 6996814-6996814 on Build GRCh38
rs2070590 Chr.17: 6997125-6997125 on Build GRCh38
rs768809125 Chr.17: 6997046-6997046 on Build GRCh38
rs2070589 Chr.17: 6997043-6997043 on Build GRCh38
rs915815128 Chr.17: 6996804-6996804 on Build GRCh38
rs764265918 Chr.17: 6997073-6997073 on Build GRCh38
rs777900096 Chr.17: 6996997-6996997 on Build GRCh38
rs758764573 Chr.17: 6996815-6996815 on Build GRCh38
rs943032943 Chr.17: 6996964-6996964 on Build GRCh38
rs374987218 Chr.17: 6997038-6997038 on Build GRCh38
rs746121469 Chr.17: 6996924-6996924 on Build GRCh38
rs760950012 Chr.17: 6997072-6997072 on Build GRCh38
rs761436579 Chr.17: 6996802-6996802 on Build GRCh38
rs537773303 Chr.17: 6997127-6997127 on Build GRCh38
rs554860166 Chr.17: 6997205-6997205 on Build GRCh38
rs745825725 Chr.17: 6996870-6996870 on Build GRCh38
rs768584209 Chr.17: 6997037-6997037 on Build GRCh38
rs867516551 Chr.17: 6997051-6997051 on Build GRCh38
rs746887910 Chr.17: 6997067-6997067 on Build GRCh38
rs149287091 Chr.17: 6996894-6996894 on Build GRCh38
rs531141172 Chr.17: 6996780-6996780 on Build GRCh38
rs2073438 Chr.17: 6996757-6996757 on Build GRCh38
rs779883218 Chr.17: 6997023-6997023 on Build GRCh38
rs1036508479 Chr.17: 6996765-6996765 on Build GRCh38
rs761497528 Chr.17: 6996904-6996904 on Build GRCh38
rs761657767 Chr.17: 6997153-6997153 on Build GRCh38
rs138589208 Chr.17: 6996847-6996847 on Build GRCh38
rs375590131 Chr.17: 6996977-6996977 on Build GRCh38
rs775085688 Chr.17: 6996787-6996787 on Build GRCh38
rs763572123 Chr.17: 6996950-6996950 on Build GRCh38
rs183070992 Chr.17: 6996797-6996797 on Build GRCh38
rs908440369 Chr.17: 6996905-6996905 on Build GRCh38
rs957188858 Chr.17: 6997123-6997123 on Build GRCh38
rs753058889 Chr.17: 6997069-6997069 on Build GRCh38
rs776082345 Chr.17: 6996853-6996853 on Build GRCh38
rs546698755 Chr.17: 6996856-6996856 on Build GRCh38
rs199823896 Chr.17: 6996897-6996897 on Build GRCh38
rs939591074 Chr.17: 6997017-6997017 on Build GRCh38
rs927952762 Chr.17: 6997188-6997188 on Build GRCh38
rs373195321 Chr.17: 6997044-6997044 on Build GRCh38
rs111935092 Chr.17: 6997104-6997104 on Build GRCh38
rs980866518 Chr.17: 6997179-6997179 on Build GRCh38
rs911639766 Chr.17: 6997191-6997191 on Build GRCh38
rs766833130 Chr.17: 6996829-6996829 on Build GRCh38
rs755332420 Chr.17: 6996974-6996974 on Build GRCh38
rs746913082 Chr.17: 6997030-6997030 on Build GRCh38

View Full Product Details