Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7220357-7220853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADVL
Entrez Gene ID
37
Gene Name
acyl-CoA dehydrogenase, very long chain
Gene Aliases
ACAD6, LCACD, VLCAD
UniGene
Hs.437178
Gene Chromosome Location
Chr.17: 7217125-7225267 on Build GRCh38
Transcripts
NM_000018.3 NM_001033859.2 NM_001270447.1 NM_001270448.1 XM_006721516.2 XM_011523829.1 XM_011523830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588601 c.139-7T>G p.? Chr.17: 7220457-7220457 on Build GRCh38
140115 c.166C>T p.P56S Chr.17: 7220491-7220491 on Build GRCh38
6591704 c.139-9_139-8delTT p.? Chr.17: 7220455-7220456 on Build GRCh38
437270 c.163C>T p.H55Y Chr.17: 7220488-7220488 on Build GRCh38
7139920 c.234C>A p.F78L Chr.17: 7220633-7220633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057516979 Chr.17: 7220796-7220797 on Build GRCh38
rs996950993 Chr.17: 7220400-7220400 on Build GRCh38
rs786204713 Chr.17: 7220786-7220787 on Build GRCh38
rs776422793 Chr.17: 7220682-7220682 on Build GRCh38
rs773003854 Chr.17: 7220711-7220711 on Build GRCh38
rs113809829 Chr.17: 7220601-7220601 on Build GRCh38
rs201085520 Chr.17: 7220654-7220654 on Build GRCh38
rs758471890 Chr.17: 7220759-7220759 on Build GRCh38
rs753490272 Chr.17: 7220440-7220440 on Build GRCh38
rs758986446 Chr.17: 7220448-7220448 on Build GRCh38
rs199945418 Chr.17: 7220700-7220700 on Build GRCh38
rs751162358 Chr.17: 7220718-7220718 on Build GRCh38
rs372863720 Chr.17: 7220466-7220466 on Build GRCh38
rs749482177 Chr.17: 7220669-7220669 on Build GRCh38
rs958166043 Chr.17: 7220820-7220820 on Build GRCh38
rs147357106 Chr.17: 7220663-7220663 on Build GRCh38
rs1022457539 Chr.17: 7220720-7220720 on Build GRCh38
rs762599719 Chr.17: 7220634-7220634 on Build GRCh38
rs958328801 Chr.17: 7220534-7220534 on Build GRCh38
rs369835906 Chr.17: 7220421-7220421 on Build GRCh38
rs774141572 Chr.17: 7220787-7220787 on Build GRCh38
rs775298132 Chr.17: 7220701-7220701 on Build GRCh38
rs762537169 Chr.17: 7220714-7220714 on Build GRCh38
rs769865745 Chr.17: 7220467-7220467 on Build GRCh38
rs954670873 Chr.17: 7220402-7220402 on Build GRCh38
rs773109868 Chr.17: 7220414-7220414 on Build GRCh38
rs756969179 Chr.17: 7220529-7220529 on Build GRCh38
rs557260142 Chr.17: 7220828-7220828 on Build GRCh38
rs377243702 Chr.17: 7220553-7220553 on Build GRCh38
rs530278910 Chr.17: 7220675-7220675 on Build GRCh38
rs754188342 Chr.17: 7220741-7220741 on Build GRCh38
rs776453042 Chr.17: 7220611-7220611 on Build GRCh38
rs778993830 Chr.17: 7220672-7220672 on Build GRCh38
rs201498083 Chr.17: 7220778-7220778 on Build GRCh38
rs780688207 Chr.17: 7220452-7220452 on Build GRCh38
rs780118841 Chr.17: 7220761-7220761 on Build GRCh38
rs751066230 Chr.17: 7220648-7220648 on Build GRCh38
rs753577095 Chr.17: 7220526-7220526 on Build GRCh38
rs750653177 Chr.17: 7220827-7220827 on Build GRCh38
rs766001030 Chr.17: 7220637-7220637 on Build GRCh38
rs780773899 Chr.17: 7220545-7220545 on Build GRCh38
rs778504571 Chr.17: 7220443-7220443 on Build GRCh38
rs374535729 Chr.17: 7220444-7220444 on Build GRCh38
rs765432568 Chr.17: 7220524-7220524 on Build GRCh38
rs886053372 Chr.17: 7220505-7220505 on Build GRCh38
rs113500833 Chr.17: 7220764-7220764 on Build GRCh38
rs755486571 Chr.17: 7220548-7220548 on Build GRCh38
rs886053373 Chr.17: 7220766-7220766 on Build GRCh38
rs752537465 Chr.17: 7220427-7220427 on Build GRCh38
rs750043368 Chr.17: 7220626-7220626 on Build GRCh38
rs769138546 Chr.17: 7220459-7220459 on Build GRCh38
rs564913508 Chr.17: 7220377-7220377 on Build GRCh38
rs756582043 Chr.17: 7220736-7220736 on Build GRCh38
rs370156298 Chr.17: 7220625-7220625 on Build GRCh38
rs768632138 Chr.17: 7220676-7220676 on Build GRCh38
rs142745084 Chr.17: 7220521-7220521 on Build GRCh38
rs755889997 Chr.17: 7220433-7220433 on Build GRCh38
rs979893163 Chr.17: 7220516-7220516 on Build GRCh38
rs759197049 Chr.17: 7220789-7220789 on Build GRCh38
rs1057517012 Chr.17: 7220677-7220677 on Build GRCh38
rs761739412 Chr.17: 7220699-7220699 on Build GRCh38
rs771055189 Chr.17: 7220512-7220512 on Build GRCh38
rs139427392 Chr.17: 7220774-7220774 on Build GRCh38
rs794727695 Chr.17: 7220782-7220782 on Build GRCh38
rs767548421 Chr.17: 7220499-7220499 on Build GRCh38
rs756800545 Chr.17: 7220441-7220441 on Build GRCh38
rs886043235 Chr.17: 7220810-7220810 on Build GRCh38
rs769481185 Chr.17: 7220707-7220707 on Build GRCh38
rs1057518506 Chr.17: 7220804-7220813 on Build GRCh38
rs764767909 Chr.17: 7220533-7220533 on Build GRCh38
rs28934585 Chr.17: 7220519-7220519 on Build GRCh38
rs750675692 Chr.17: 7220786-7220786 on Build GRCh38
rs755575335 Chr.17: 7220649-7220649 on Build GRCh38
rs747439748 Chr.17: 7220454-7220454 on Build GRCh38
rs940857936 Chr.17: 7220621-7220621 on Build GRCh38
rs760625298 Chr.17: 7220597-7220597 on Build GRCh38
rs727503790 Chr.17: 7220514-7220514 on Build GRCh38
rs768204728 Chr.17: 7220734-7220734 on Build GRCh38
rs990837045 Chr.17: 7220542-7220542 on Build GRCh38
rs901293987 Chr.17: 7220382-7220382 on Build GRCh38
rs757508022 Chr.17: 7220743-7220743 on Build GRCh38
rs2049851 Chr.17: 7220376-7220377 on Build GRCh38
rs398123087 Chr.17: 7220670-7220670 on Build GRCh38
rs781758731 Chr.17: 7220652-7220652 on Build GRCh38
rs757608507 Chr.17: 7220817-7220817 on Build GRCh38
rs140566084 Chr.17: 7220796-7220796 on Build GRCh38
rs753149462 Chr.17: 7220735-7220735 on Build GRCh38
rs530515395 Chr.17: 7220411-7220411 on Build GRCh38
rs774353448 Chr.17: 7220596-7220596 on Build GRCh38
rs763799450 Chr.17: 7220437-7220437 on Build GRCh38
rs759548707 Chr.17: 7220415-7220415 on Build GRCh38
rs771808680 Chr.17: 7220663-7220663 on Build GRCh38
rs761492981 Chr.17: 7220615-7220615 on Build GRCh38
rs750003292 Chr.17: 7220541-7220541 on Build GRCh38
rs775398187 Chr.17: 7220506-7220506 on Build GRCh38
rs769723013 Chr.17: 7220773-7220773 on Build GRCh38
rs774676423 Chr.17: 7220582-7220582 on Build GRCh38
rs754207297 Chr.17: 7220813-7220813 on Build GRCh38
rs768537914 Chr.17: 7220599-7220599 on Build GRCh38
rs764028320 Chr.17: 7220821-7220821 on Build GRCh38
rs544122648 Chr.17: 7220493-7220493 on Build GRCh38
rs748048602 Chr.17: 7220767-7220767 on Build GRCh38
rs376629743 Chr.17: 7220572-7220572 on Build GRCh38
rs370929055 Chr.17: 7220819-7220819 on Build GRCh38
rs748428641 Chr.17: 7220568-7220568 on Build GRCh38
rs773378857 Chr.17: 7220470-7220470 on Build GRCh38
rs754620326 Chr.17: 7220803-7220803 on Build GRCh38
rs770101303 Chr.17: 7220570-7220570 on Build GRCh38
rs764384199 Chr.17: 7220738-7220738 on Build GRCh38
rs746009227 Chr.17: 7220755-7220755 on Build GRCh38
rs760543441 Chr.17: 7220778-7220783 on Build GRCh38
rs1014076291 Chr.17: 7220485-7220485 on Build GRCh38
rs747072789 Chr.17: 7220832-7220832 on Build GRCh38
rs202244937 Chr.17: 7220727-7220727 on Build GRCh38
rs749451023 Chr.17: 7220577-7220577 on Build GRCh38
rs796051907 Chr.17: 7220659-7220659 on Build GRCh38
rs745642661 Chr.17: 7220590-7220591 on Build GRCh38
rs1005937282 Chr.17: 7220445-7220445 on Build GRCh38
rs1008843538 Chr.17: 7220760-7220760 on Build GRCh38
rs796051912 Chr.17: 7220609-7220610 on Build GRCh38
rs780020193 Chr.17: 7220831-7220831 on Build GRCh38
rs779087862 Chr.17: 7220748-7220748 on Build GRCh38
rs770999464 Chr.17: 7220582-7220582 on Build GRCh38
rs537031882 Chr.17: 7220824-7220824 on Build GRCh38
rs760535113 Chr.17: 7220508-7220508 on Build GRCh38
rs1029039003 Chr.17: 7220398-7220398 on Build GRCh38
rs967186367 Chr.17: 7220511-7220512 on Build GRCh38
rs764628802 Chr.17: 7220812-7220812 on Build GRCh38
rs766091747 Chr.17: 7220715-7220715 on Build GRCh38
rs1015495051 Chr.17: 7220450-7220450 on Build GRCh38
rs1041233613 Chr.17: 7220381-7220381 on Build GRCh38

View Full Product Details