Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16441832-16442336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC75A
Entrez Gene ID
388341
Gene Name
leucine rich repeat containing 75A
Gene Aliases
C17orf76, FAM211A
UniGene
Hs.25425
Gene Chromosome Location
Chr.17: 16442005-16492191 on Build GRCh38
Transcripts
NM_001113567.2 NM_207387.3 XM_011523845.2 XM_017024619.1 XM_017024620.1 NR_027158.1 NR_027159.1 NR_027160.1 NR_027161.1 NR_027162.1 NR_027163.1 NR_027164.1 NR_027165.1 NR_027166.1 NR_027167.1 NR_027168.2 NR_027169.1 NR_027170.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_027667.1 NR_045021.1 NR_045022.1 NR_045023.1 NR_045025.1 NR_045027.1 NR_045028.1 NR_045029.1

Gene Symbol
LRRC75A-AS1
Entrez Gene ID
125144
Gene Name
LRRC75A antisense RNA 1
Gene Aliases
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
UniGene
Hs.368934
Gene Chromosome Location
Chr.17: 16438987-16470648 on Build GRCh38
Transcripts
NM_001113567.2 NM_207387.3 XM_011523845.2 XM_017024619.1 XM_017024620.1 NR_027158.1 NR_027159.1 NR_027160.1 NR_027161.1 NR_027162.1 NR_027163.1 NR_027164.1 NR_027165.1 NR_027166.1 NR_027167.1 NR_027168.2 NR_027169.1 NR_027170.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_027667.1 NR_045021.1 NR_045022.1 NR_045023.1 NR_045025.1 NR_045027.1 NR_045028.1 NR_045029.1

SNP Detail

SNP ID Chromosome Location
rs115049073 Chr.17: 16442246-16442246 on Build GRCh38
rs549698106 Chr.17: 16442224-16442224 on Build GRCh38
rs973700387 Chr.17: 16442282-16442284 on Build GRCh38
rs77792135 Chr.17: 16442267-16442267 on Build GRCh38
rs954865141 Chr.17: 16442033-16442033 on Build GRCh38
rs1133342 Chr.17: 16441865-16441865 on Build GRCh38
rs976446113 Chr.17: 16442300-16442300 on Build GRCh38
rs528562232 Chr.17: 16442046-16442047 on Build GRCh38
rs746129656 Chr.17: 16442005-16442006 on Build GRCh38
rs999561567 Chr.17: 16442226-16442226 on Build GRCh38
rs184545038 Chr.17: 16442303-16442303 on Build GRCh38
rs948027874 Chr.17: 16442104-16442104 on Build GRCh38
rs374508794 Chr.17: 16441920-16441921 on Build GRCh38
rs562605254 Chr.17: 16442107-16442107 on Build GRCh38
rs1005086247 Chr.17: 16442266-16442266 on Build GRCh38
rs569577855 Chr.17: 16442299-16442299 on Build GRCh38
rs966703931 Chr.17: 16441896-16441896 on Build GRCh38
rs540857518 Chr.17: 16441906-16441906 on Build GRCh38
rs985333502 Chr.17: 16441976-16441976 on Build GRCh38
rs1025081810 Chr.17: 16442198-16442198 on Build GRCh38
rs910967457 Chr.17: 16441977-16441977 on Build GRCh38
rs935169102 Chr.17: 16442137-16442137 on Build GRCh38
rs546844594 Chr.17: 16442239-16442239 on Build GRCh38
rs963987794 Chr.17: 16442279-16442279 on Build GRCh38
rs1032272788 Chr.17: 16441921-16441921 on Build GRCh38
rs973838582 Chr.17: 16442074-16442074 on Build GRCh38
rs757638892 Chr.17: 16442254-16442254 on Build GRCh38
rs771770519 Chr.17: 16441982-16441982 on Build GRCh38
rs763312437 Chr.17: 16441935-16441935 on Build GRCh38
rs985688733 Chr.17: 16442259-16442259 on Build GRCh38
rs1048437878 Chr.17: 16442089-16442089 on Build GRCh38
rs370529510 Chr.17: 16442112-16442112 on Build GRCh38
rs397768741 Chr.17: 16441922-16441922 on Build GRCh38
rs916568785 Chr.17: 16442092-16442092 on Build GRCh38
rs114046767 Chr.17: 16442126-16442126 on Build GRCh38
rs942192514 Chr.17: 16442140-16442140 on Build GRCh38
rs368477611 Chr.17: 16442305-16442305 on Build GRCh38
rs963652759 Chr.17: 16442062-16442062 on Build GRCh38
rs1133356 Chr.17: 16441871-16441871 on Build GRCh38
rs895325415 Chr.17: 16442142-16442142 on Build GRCh38
rs191253610 Chr.17: 16442055-16442055 on Build GRCh38
rs1040003822 Chr.17: 16442199-16442199 on Build GRCh38
rs560500713 Chr.17: 16441987-16441987 on Build GRCh38
rs149439336 Chr.17: 16442060-16442060 on Build GRCh38
rs5819572 Chr.17: 16441920-16441920 on Build GRCh38
rs533309714 Chr.17: 16442110-16442110 on Build GRCh38
rs918211185 Chr.17: 16442056-16442056 on Build GRCh38
rs762960477 Chr.17: 16442194-16442194 on Build GRCh38
rs564436423 Chr.17: 16442190-16442190 on Build GRCh38
rs752779427 Chr.17: 16442187-16442187 on Build GRCh38
rs376445816 Chr.17: 16441920-16441920 on Build GRCh38
rs533053984 Chr.17: 16442207-16442207 on Build GRCh38
rs982232500 Chr.17: 16442119-16442119 on Build GRCh38
rs978390005 Chr.17: 16441915-16441915 on Build GRCh38
rs899509850 Chr.17: 16442213-16442214 on Build GRCh38
rs976477333 Chr.17: 16442005-16442005 on Build GRCh38
rs147539693 Chr.17: 16442141-16442141 on Build GRCh38
rs893758399 Chr.17: 16442185-16442185 on Build GRCh38
rs965184104 Chr.17: 16442287-16442287 on Build GRCh38

View Full Product Details