Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16447749-16448241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC75A
Entrez Gene ID
388341
Gene Name
leucine rich repeat containing 75A
Gene Aliases
C17orf76, FAM211A
UniGene
Hs.25425
Gene Chromosome Location
Chr.17: 16442005-16492191 on Build GRCh38
Transcripts
NM_001113567.2 NM_207387.3 XM_011523845.2 XM_017024619.1 XM_017024620.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_045023.1 NR_045025.1

Gene Symbol
LRRC75A-AS1
Entrez Gene ID
125144
Gene Name
LRRC75A antisense RNA 1
Gene Aliases
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
UniGene
Hs.368934
Gene Chromosome Location
Chr.17: 16438987-16470648 on Build GRCh38
Transcripts
NM_001113567.2 NM_207387.3 XM_011523845.2 XM_017024619.1 XM_017024620.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_045023.1 NR_045025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6500915 c.413G>A p.R138Q Chr.17: 16447923-16447923 on Build GRCh38
2739333 c.454C>T p.R152W Chr.17: 16447882-16447882 on Build GRCh38
6363121 c.488C>T p.A163V Chr.17: 16447848-16447848 on Build GRCh38
1588689 c.486G>T p.Q162H Chr.17: 16447850-16447850 on Build GRCh38
6244749 c.439C>A p.H147N Chr.17: 16447897-16447897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746601653 Chr.17: 16447821-16447821 on Build GRCh38
rs983847134 Chr.17: 16448004-16448004 on Build GRCh38
rs542773464 Chr.17: 16448204-16448204 on Build GRCh38
rs750336093 Chr.17: 16447989-16447989 on Build GRCh38
rs1052518593 Chr.17: 16448082-16448082 on Build GRCh38
rs141608934 Chr.17: 16448039-16448039 on Build GRCh38
rs534158974 Chr.17: 16448038-16448038 on Build GRCh38
rs752428551 Chr.17: 16448054-16448054 on Build GRCh38
rs751269172 Chr.17: 16447824-16447825 on Build GRCh38
rs945029436 Chr.17: 16448020-16448020 on Build GRCh38
rs761099123 Chr.17: 16447881-16447881 on Build GRCh38
rs549028561 Chr.17: 16447871-16447871 on Build GRCh38
rs867114797 Chr.17: 16447811-16447811 on Build GRCh38
rs375595106 Chr.17: 16447795-16447795 on Build GRCh38
rs996341873 Chr.17: 16448180-16448180 on Build GRCh38
rs369833733 Chr.17: 16447828-16447828 on Build GRCh38
rs568769683 Chr.17: 16447873-16447873 on Build GRCh38
rs35661885 Chr.17: 16447798-16447799 on Build GRCh38
rs535531962 Chr.17: 16447882-16447882 on Build GRCh38
rs772568108 Chr.17: 16448097-16448097 on Build GRCh38
rs906206166 Chr.17: 16447803-16447803 on Build GRCh38
rs755860256 Chr.17: 16447876-16447876 on Build GRCh38
rs779960352 Chr.17: 16447918-16447918 on Build GRCh38
rs746310646 Chr.17: 16448090-16448090 on Build GRCh38
rs529018556 Chr.17: 16447867-16447867 on Build GRCh38
rs755533397 Chr.17: 16448061-16448061 on Build GRCh38
rs750383502 Chr.17: 16447847-16447847 on Build GRCh38
rs1042022326 Chr.17: 16448022-16448022 on Build GRCh38
rs758576943 Chr.17: 16448058-16448058 on Build GRCh38
rs571600380 Chr.17: 16448028-16448028 on Build GRCh38
rs762997128 Chr.17: 16448037-16448037 on Build GRCh38
rs573871131 Chr.17: 16448192-16448192 on Build GRCh38
rs754870279 Chr.17: 16448040-16448040 on Build GRCh38
rs777359337 Chr.17: 16448075-16448075 on Build GRCh38
rs780016788 Chr.17: 16447819-16447819 on Build GRCh38
rs769102378 Chr.17: 16448216-16448216 on Build GRCh38
rs900681161 Chr.17: 16448178-16448178 on Build GRCh38
rs966421208 Chr.17: 16447872-16447872 on Build GRCh38
rs780678444 Chr.17: 16448053-16448053 on Build GRCh38
rs889271265 Chr.17: 16448101-16448101 on Build GRCh38
rs147047205 Chr.17: 16448215-16448215 on Build GRCh38
rs1019442508 Chr.17: 16448181-16448181 on Build GRCh38
rs926419401 Chr.17: 16447942-16447942 on Build GRCh38
rs747590368 Chr.17: 16447859-16447859 on Build GRCh38
rs992303812 Chr.17: 16447955-16447955 on Build GRCh38
rs754603305 Chr.17: 16447828-16447828 on Build GRCh38
rs868134004 Chr.17: 16447911-16447911 on Build GRCh38
rs1007752125 Chr.17: 16448176-16448176 on Build GRCh38
rs767873362 Chr.17: 16447875-16447875 on Build GRCh38
rs765388071 Chr.17: 16447958-16447958 on Build GRCh38
rs963411122 Chr.17: 16448032-16448032 on Build GRCh38
rs752926305 Chr.17: 16448061-16448065 on Build GRCh38
rs138809782 Chr.17: 16447827-16447827 on Build GRCh38
rs761766426 Chr.17: 16447952-16447952 on Build GRCh38
rs1010681819 Chr.17: 16448135-16448135 on Build GRCh38
rs1054649956 Chr.17: 16448078-16448078 on Build GRCh38
rs901958792 Chr.17: 16448199-16448199 on Build GRCh38
rs557066360 Chr.17: 16447943-16447943 on Build GRCh38
rs917715438 Chr.17: 16448002-16448002 on Build GRCh38
rs775839843 Chr.17: 16447877-16447877 on Build GRCh38

View Full Product Details