Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28769594-28770091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YTHDF2
Entrez Gene ID
51441
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Gene Aliases
CAHL, HGRG8, NY-REN-2
UniGene
Hs.532286
Gene Chromosome Location
Chr.1: 28736621-28769775 on Build GRCh38
Transcripts
NM_001172828.1 NM_001173128.1 NM_016258.2

SNP Detail

SNP ID Chromosome Location
rs764638256 Chr.1: 28770000-28770000 on Build GRCh38
rs532657427 Chr.1: 28770067-28770067 on Build GRCh38
rs895777362 Chr.1: 28769738-28769738 on Build GRCh38
rs765667566 Chr.1: 28769711-28769711 on Build GRCh38
rs951168885 Chr.1: 28769757-28769757 on Build GRCh38
rs746057489 Chr.1: 28769754-28769754 on Build GRCh38
rs560565520 Chr.1: 28770046-28770046 on Build GRCh38
rs574371397 Chr.1: 28769650-28769650 on Build GRCh38
rs934268709 Chr.1: 28770016-28770016 on Build GRCh38
rs933570794 Chr.1: 28769656-28769656 on Build GRCh38
rs891932324 Chr.1: 28769667-28769667 on Build GRCh38
rs904200471 Chr.1: 28769678-28769678 on Build GRCh38
rs1014174143 Chr.1: 28769745-28769745 on Build GRCh38
rs945790154 Chr.1: 28769671-28769671 on Build GRCh38
rs973205142 Chr.1: 28769795-28769798 on Build GRCh38
rs570483462 Chr.1: 28769959-28769959 on Build GRCh38
rs946207631 Chr.1: 28769972-28769972 on Build GRCh38
rs140261662 Chr.1: 28770041-28770041 on Build GRCh38
rs990555146 Chr.1: 28769868-28769868 on Build GRCh38
rs562046759 Chr.1: 28770038-28770038 on Build GRCh38
rs116002631 Chr.1: 28770031-28770031 on Build GRCh38
rs1007008308 Chr.1: 28769714-28769714 on Build GRCh38
rs895451279 Chr.1: 28770053-28770053 on Build GRCh38
rs1051340306 Chr.1: 28770023-28770023 on Build GRCh38
rs530029515 Chr.1: 28770040-28770040 on Build GRCh38
rs950654001 Chr.1: 28769808-28769808 on Build GRCh38
rs937681379 Chr.1: 28769853-28769853 on Build GRCh38
rs186550777 Chr.1: 28770030-28770030 on Build GRCh38
rs374032666 Chr.1: 28769703-28769703 on Build GRCh38
rs1025483017 Chr.1: 28769746-28769746 on Build GRCh38
rs182370004 Chr.1: 28769795-28769795 on Build GRCh38
rs1042788960 Chr.1: 28769672-28769672 on Build GRCh38
rs1006872556 Chr.1: 28770058-28770058 on Build GRCh38
rs371452625 Chr.1: 28769651-28769651 on Build GRCh38
rs914961969 Chr.1: 28769869-28769870 on Build GRCh38
rs937087178 Chr.1: 28770042-28770042 on Build GRCh38
rs974810233 Chr.1: 28769895-28769895 on Build GRCh38
rs149703134 Chr.1: 28769910-28769910 on Build GRCh38
rs761104066 Chr.1: 28769954-28769954 on Build GRCh38
rs942010805 Chr.1: 28769625-28769625 on Build GRCh38
rs913422283 Chr.1: 28769932-28769932 on Build GRCh38
rs963773012 Chr.1: 28769840-28769840 on Build GRCh38
rs762476185 Chr.1: 28770043-28770043 on Build GRCh38
rs533811724 Chr.1: 28769659-28769659 on Build GRCh38
rs370958900 Chr.1: 28769958-28769958 on Build GRCh38
rs527591643 Chr.1: 28769782-28769782 on Build GRCh38
rs979245356 Chr.1: 28769826-28769827 on Build GRCh38
rs562345026 Chr.1: 28769947-28769947 on Build GRCh38
rs145509594 Chr.1: 28769942-28769942 on Build GRCh38
rs774455849 Chr.1: 28769666-28769666 on Build GRCh38
rs921927801 Chr.1: 28769902-28769902 on Build GRCh38
rs35926776 Chr.1: 28769759-28769760 on Build GRCh38
rs998197720 Chr.1: 28769632-28769632 on Build GRCh38
rs975134788 Chr.1: 28769636-28769636 on Build GRCh38
rs1001239354 Chr.1: 28769698-28769698 on Build GRCh38
rs1016376789 Chr.1: 28769828-28769828 on Build GRCh38
rs1038476189 Chr.1: 28770066-28770066 on Build GRCh38
rs775677324 Chr.1: 28769824-28769824 on Build GRCh38
rs922211111 Chr.1: 28769640-28769640 on Build GRCh38
rs987644733 Chr.1: 28769913-28769913 on Build GRCh38

View Full Product Details