Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29574134-29574637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIT1
Entrez Gene ID
28964
Gene Name
GIT ArfGAP 1
Gene Aliases
-
UniGene
Hs.514051
Gene Chromosome Location
Chr.17: 29573469-29589592 on Build GRCh38
Transcripts
NM_001085454.1 NM_014030.3 XM_011524684.1 XM_011524685.1

SNP Detail

SNP ID Chromosome Location
rs773459082 Chr.17: 29574551-29574558 on Build GRCh38
rs1004725302 Chr.17: 29574217-29574217 on Build GRCh38
rs758822363 Chr.17: 29574272-29574272 on Build GRCh38
rs554739611 Chr.17: 29574402-29574402 on Build GRCh38
rs757604901 Chr.17: 29574585-29574585 on Build GRCh38
rs540096788 Chr.17: 29574334-29574334 on Build GRCh38
rs553504640 Chr.17: 29574539-29574539 on Build GRCh38
rs1027077204 Chr.17: 29574587-29574587 on Build GRCh38
rs755750925 Chr.17: 29574411-29574411 on Build GRCh38
rs763135559 Chr.17: 29574600-29574600 on Build GRCh38
rs189416822 Chr.17: 29574342-29574342 on Build GRCh38
rs1025129248 Chr.17: 29574383-29574383 on Build GRCh38
rs931766242 Chr.17: 29574458-29574458 on Build GRCh38
rs372467014 Chr.17: 29574545-29574546 on Build GRCh38
rs772363287 Chr.17: 29574512-29574513 on Build GRCh38
rs398030605 Chr.17: 29574520-29574521 on Build GRCh38
rs545359898 Chr.17: 29574542-29574542 on Build GRCh38
rs552908027 Chr.17: 29574455-29574455 on Build GRCh38
rs992346779 Chr.17: 29574367-29574367 on Build GRCh38
rs568042218 Chr.17: 29574430-29574430 on Build GRCh38
rs370229539 Chr.17: 29574564-29574564 on Build GRCh38
rs781462538 Chr.17: 29574487-29574487 on Build GRCh38
rs980897929 Chr.17: 29574545-29574545 on Build GRCh38
rs969493051 Chr.17: 29574391-29574391 on Build GRCh38
rs756150649 Chr.17: 29574595-29574595 on Build GRCh38
rs186422605 Chr.17: 29574399-29574399 on Build GRCh38
rs191283001 Chr.17: 29574183-29574183 on Build GRCh38
rs765980878 Chr.17: 29574537-29574537 on Build GRCh38
rs983153554 Chr.17: 29574431-29574431 on Build GRCh38
rs184533236 Chr.17: 29574311-29574311 on Build GRCh38
rs764047945 Chr.17: 29574565-29574565 on Build GRCh38
rs35948647 Chr.17: 29574519-29574520 on Build GRCh38
rs766710389 Chr.17: 29574273-29574273 on Build GRCh38
rs760383808 Chr.17: 29574526-29574526 on Build GRCh38
rs770227191 Chr.17: 29574511-29574511 on Build GRCh38
rs907796196 Chr.17: 29574433-29574433 on Build GRCh38
rs756793889 Chr.17: 29574547-29574547 on Build GRCh38
rs1040205335 Chr.17: 29574452-29574452 on Build GRCh38
rs940586687 Chr.17: 29574451-29574451 on Build GRCh38
rs995863241 Chr.17: 29574263-29574263 on Build GRCh38
rs557945815 Chr.17: 29574336-29574336 on Build GRCh38
rs1040152395 Chr.17: 29574228-29574228 on Build GRCh38
rs1031260995 Chr.17: 29574349-29574349 on Build GRCh38
rs775905598 Chr.17: 29574513-29574515 on Build GRCh38
rs776699500 Chr.17: 29574534-29574535 on Build GRCh38
rs753392472 Chr.17: 29574538-29574538 on Build GRCh38
rs776024034 Chr.17: 29574513-29574513 on Build GRCh38
rs41274276 Chr.17: 29574310-29574310 on Build GRCh38
rs771274837 Chr.17: 29574484-29574484 on Build GRCh38
rs769006617 Chr.17: 29574519-29574519 on Build GRCh38
rs886314182 Chr.17: 29574204-29574204 on Build GRCh38
rs745922170 Chr.17: 29574506-29574506 on Build GRCh38
rs549757366 Chr.17: 29574240-29574240 on Build GRCh38
rs563552803 Chr.17: 29574584-29574584 on Build GRCh38
rs901616196 Chr.17: 29574262-29574262 on Build GRCh38
rs371571846 Chr.17: 29574541-29574541 on Build GRCh38
rs780439070 Chr.17: 29574608-29574608 on Build GRCh38
rs952877001 Chr.17: 29574599-29574599 on Build GRCh38
rs765471563 Chr.17: 29574545-29574545 on Build GRCh38
rs181408672 Chr.17: 29574393-29574393 on Build GRCh38
rs538028952 Chr.17: 29574350-29574350 on Build GRCh38
rs763230033 Chr.17: 29574515-29574515 on Build GRCh38
rs749864276 Chr.17: 29574612-29574612 on Build GRCh38
rs777170597 Chr.17: 29574415-29574415 on Build GRCh38
rs1050130055 Chr.17: 29574464-29574464 on Build GRCh38
rs9913116 Chr.17: 29574549-29574549 on Build GRCh38
rs748856148 Chr.17: 29574461-29574461 on Build GRCh38
rs1000551274 Chr.17: 29574498-29574498 on Build GRCh38
rs745964877 Chr.17: 29574594-29574594 on Build GRCh38
rs903566710 Chr.17: 29574478-29574478 on Build GRCh38
rs573668174 Chr.17: 29574418-29574418 on Build GRCh38
rs540675350 Chr.17: 29574444-29574444 on Build GRCh38
rs547730491 Chr.17: 29574298-29574298 on Build GRCh38
rs1031061919 Chr.17: 29574517-29574517 on Build GRCh38

View Full Product Details