Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18120633-18121124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO15A
Entrez Gene ID
51168
Gene Name
myosin XVA
Gene Aliases
DFNB3, MYO15
UniGene
Hs.462390
Gene Chromosome Location
Chr.17: 18108706-18180227 on Build GRCh38
Transcripts
NM_016239.3 XM_011523918.2 XM_017024714.1 XM_017024715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5747823 c.1951C>T p.P651S Chr.17: 18120751-18120751 on Build GRCh38
5493537 c.1936G>A p.A646T Chr.17: 18120736-18120736 on Build GRCh38
6397026 c.2323T>C p.S775P Chr.17: 18121123-18121123 on Build GRCh38
6556214 c.2277G>T p.G759G Chr.17: 18121077-18121077 on Build GRCh38
6145973 c.1855C>A p.P619T Chr.17: 18120655-18120655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757367598 Chr.17: 18120703-18120703 on Build GRCh38
rs778896303 Chr.17: 18120706-18120706 on Build GRCh38
rs886038623 Chr.17: 18120654-18120654 on Build GRCh38
rs770493586 Chr.17: 18121054-18121054 on Build GRCh38
rs557225435 Chr.17: 18120820-18120820 on Build GRCh38
rs764581419 Chr.17: 18120903-18120903 on Build GRCh38
rs776218805 Chr.17: 18120737-18120737 on Build GRCh38
rs534687248 Chr.17: 18120666-18120666 on Build GRCh38
rs993096170 Chr.17: 18120845-18120845 on Build GRCh38
rs373561242 Chr.17: 18121102-18121102 on Build GRCh38
rs749541790 Chr.17: 18120702-18120702 on Build GRCh38
rs932191004 Chr.17: 18120916-18120916 on Build GRCh38
rs750535004 Chr.17: 18120951-18120951 on Build GRCh38
rs767978786 Chr.17: 18120824-18120824 on Build GRCh38
rs746972322 Chr.17: 18120959-18120959 on Build GRCh38
rs559233698 Chr.17: 18120656-18120656 on Build GRCh38
rs954909427 Chr.17: 18120767-18120767 on Build GRCh38
rs973362601 Chr.17: 18120814-18120814 on Build GRCh38
rs962124239 Chr.17: 18120812-18120812 on Build GRCh38
rs886052670 Chr.17: 18121059-18121059 on Build GRCh38
rs370722452 Chr.17: 18120672-18120672 on Build GRCh38
rs994845796 Chr.17: 18121075-18121075 on Build GRCh38
rs767906576 Chr.17: 18120694-18120694 on Build GRCh38
rs768453845 Chr.17: 18120736-18120736 on Build GRCh38
rs938528364 Chr.17: 18120842-18120842 on Build GRCh38
rs772650400 Chr.17: 18120796-18120796 on Build GRCh38
rs771865217 Chr.17: 18120657-18120657 on Build GRCh38
rs761069955 Chr.17: 18120870-18120870 on Build GRCh38
rs769317162 Chr.17: 18120758-18120758 on Build GRCh38
rs1036464353 Chr.17: 18121072-18121072 on Build GRCh38
rs913090043 Chr.17: 18120997-18120997 on Build GRCh38
rs754311954 Chr.17: 18121087-18121087 on Build GRCh38
rs969421672 Chr.17: 18120978-18120978 on Build GRCh38
rs761299340 Chr.17: 18121074-18121074 on Build GRCh38
rs886052669 Chr.17: 18120819-18120819 on Build GRCh38
rs2955366 Chr.17: 18120699-18120699 on Build GRCh38
rs757598891 Chr.17: 18120942-18120942 on Build GRCh38
rs945794067 Chr.17: 18120866-18120866 on Build GRCh38
rs537016494 Chr.17: 18120741-18120741 on Build GRCh38
rs779892754 Chr.17: 18120725-18120725 on Build GRCh38
rs747777586 Chr.17: 18120986-18120986 on Build GRCh38
rs761256749 Chr.17: 18120678-18120678 on Build GRCh38
rs760308017 Chr.17: 18120663-18120663 on Build GRCh38
rs1009513538 Chr.17: 18120781-18120781 on Build GRCh38
rs960260126 Chr.17: 18120991-18120991 on Build GRCh38
rs978516095 Chr.17: 18121019-18121019 on Build GRCh38
rs554641348 Chr.17: 18120698-18120698 on Build GRCh38
rs876657897 Chr.17: 18120815-18120815 on Build GRCh38
rs905301484 Chr.17: 18121101-18121101 on Build GRCh38
rs981212271 Chr.17: 18120979-18120979 on Build GRCh38
rs572234398 Chr.17: 18120963-18120963 on Build GRCh38
rs890742592 Chr.17: 18120945-18120945 on Build GRCh38
rs79760961 Chr.17: 18121038-18121038 on Build GRCh38
rs781006180 Chr.17: 18120974-18120974 on Build GRCh38
rs2955367 Chr.17: 18120952-18120952 on Build GRCh38
rs1015006421 Chr.17: 18120788-18120788 on Build GRCh38
rs925726957 Chr.17: 18121022-18121022 on Build GRCh38
rs1050691951 Chr.17: 18120927-18120927 on Build GRCh38
rs746737801 Chr.17: 18120731-18120731 on Build GRCh38
rs1042721945 Chr.17: 18120899-18120899 on Build GRCh38
rs1056888186 Chr.17: 18120697-18120697 on Build GRCh38
rs772776336 Chr.17: 18121025-18121025 on Build GRCh38
rs1050537476 Chr.17: 18121026-18121026 on Build GRCh38
rs764527969 Chr.17: 18121082-18121082 on Build GRCh38
rs376451611 Chr.17: 18120994-18120994 on Build GRCh38
rs771960641 Chr.17: 18120722-18120722 on Build GRCh38
rs754077336 Chr.17: 18120919-18120919 on Build GRCh38
rs764751762 Chr.17: 18120688-18120688 on Build GRCh38
rs904748871 Chr.17: 18120734-18120734 on Build GRCh38
rs546018470 Chr.17: 18120950-18120950 on Build GRCh38
rs952606016 Chr.17: 18120826-18120826 on Build GRCh38
rs912974134 Chr.17: 18120855-18120855 on Build GRCh38
rs776096881 Chr.17: 18121057-18121057 on Build GRCh38
rs945740176 Chr.17: 18120998-18120998 on Build GRCh38
rs755479632 Chr.17: 18120691-18120691 on Build GRCh38
rs577023485 Chr.17: 18120828-18120828 on Build GRCh38

View Full Product Details