Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45168933-45169429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXIM2
Entrez Gene ID
124790
Gene Name
hexamethylene bisacetamide inducible 2
Gene Aliases
L3
UniGene
Hs.56382
Gene Chromosome Location
Chr.17: 45160174-45170040 on Build GRCh38
Transcripts
NM_001303436.1 NM_001303437.1 NM_001303438.1 NM_001303439.1 NM_001303440.1 NM_001303441.1 NM_001303442.1 NM_001303443.1 NM_001303444.1 NM_144608.2 XM_006721687.3 XM_011524306.2 XM_011524307.2 XM_011524308.2 XM_017024167.1 XM_017024168.1 XM_017024169.1 XM_017024170.1 NR_135646.1

Gene Symbol
LOC105371795
Entrez Gene ID
105371795
Gene Name
uncharacterized LOC105371795
Gene Aliases
-
UniGene
Hs.569733
Gene Chromosome Location
Chr.17: 45168805-45171485 on Build GRCh38
Transcripts
NM_001303436.1 NM_001303437.1 NM_001303438.1 NM_001303439.1 NM_001303440.1 NM_001303441.1 NM_001303442.1 NM_001303443.1 NM_001303444.1 NM_144608.2 XM_006721687.3 XM_011524306.2 XM_011524307.2 XM_011524308.2 XM_017024167.1 XM_017024168.1 XM_017024169.1 XM_017024170.1 NR_135646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1383782 c.170A>T p.D57V Chr.17: 45169118-45169118 on Build GRCh38
5040754 c.325A>C p.S109R Chr.17: 45169273-45169273 on Build GRCh38
1563805 c.370G>A p.A124T Chr.17: 45169318-45169318 on Build GRCh38
1662132 c.179G>A p.G60E Chr.17: 45169127-45169127 on Build GRCh38
166937 c.107G>A p.R36H Chr.17: 45169055-45169055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376981377 Chr.17: 45169373-45169373 on Build GRCh38
rs567137721 Chr.17: 45168957-45168957 on Build GRCh38
rs1025031474 Chr.17: 45169017-45169017 on Build GRCh38
rs779522379 Chr.17: 45169111-45169111 on Build GRCh38
rs774574870 Chr.17: 45169041-45169041 on Build GRCh38
rs769935397 Chr.17: 45169240-45169240 on Build GRCh38
rs779248569 Chr.17: 45169194-45169194 on Build GRCh38
rs774272919 Chr.17: 45169269-45169269 on Build GRCh38
rs780558286 Chr.17: 45169037-45169037 on Build GRCh38
rs768778728 Chr.17: 45169265-45169265 on Build GRCh38
rs749621054 Chr.17: 45168981-45168981 on Build GRCh38
rs774469146 Chr.17: 45169357-45169357 on Build GRCh38
rs762877238 Chr.17: 45169084-45169084 on Build GRCh38
rs543432373 Chr.17: 45169125-45169125 on Build GRCh38
rs899367390 Chr.17: 45169236-45169236 on Build GRCh38
rs370295565 Chr.17: 45169171-45169171 on Build GRCh38
rs201451805 Chr.17: 45169210-45169210 on Build GRCh38
rs780570108 Chr.17: 45169103-45169103 on Build GRCh38
rs772683379 Chr.17: 45169364-45169364 on Build GRCh38
rs758705838 Chr.17: 45169216-45169216 on Build GRCh38
rs751903343 Chr.17: 45169324-45169324 on Build GRCh38
rs372890339 Chr.17: 45169166-45169166 on Build GRCh38
rs756078837 Chr.17: 45169331-45169331 on Build GRCh38
rs371493294 Chr.17: 45169359-45169359 on Build GRCh38
rs940976513 Chr.17: 45169209-45169209 on Build GRCh38
rs756765702 Chr.17: 45169090-45169090 on Build GRCh38
rs747444125 Chr.17: 45169225-45169225 on Build GRCh38
rs748603320 Chr.17: 45169048-45169048 on Build GRCh38
rs777629298 Chr.17: 45169029-45169029 on Build GRCh38
rs777924513 Chr.17: 45169126-45169126 on Build GRCh38
rs767792709 Chr.17: 45168993-45168993 on Build GRCh38
rs764646055 Chr.17: 45169320-45169320 on Build GRCh38
rs766275543 Chr.17: 45169009-45169009 on Build GRCh38
rs772260628 Chr.17: 45169049-45169049 on Build GRCh38
rs911858855 Chr.17: 45169191-45169191 on Build GRCh38
rs754754344 Chr.17: 45169019-45169019 on Build GRCh38
rs764959645 Chr.17: 45169026-45169026 on Build GRCh38
rs756855212 Chr.17: 45169031-45169031 on Build GRCh38
rs775507935 Chr.17: 45169241-45169241 on Build GRCh38
rs73319050 Chr.17: 45169151-45169151 on Build GRCh38
rs746111578 Chr.17: 45169329-45169329 on Build GRCh38
rs906175857 Chr.17: 45169283-45169283 on Build GRCh38
rs773266071 Chr.17: 45168998-45168998 on Build GRCh38
rs753750323 Chr.17: 45169016-45169016 on Build GRCh38
rs368642121 Chr.17: 45169222-45169222 on Build GRCh38
rs755426705 Chr.17: 45169110-45169110 on Build GRCh38
rs143018255 Chr.17: 45169082-45169082 on Build GRCh38
rs757502659 Chr.17: 45169407-45169407 on Build GRCh38
rs771910802 Chr.17: 45168975-45168975 on Build GRCh38
rs760300422 Chr.17: 45169028-45169029 on Build GRCh38
rs758905931 Chr.17: 45169313-45169313 on Build GRCh38
rs768996427 Chr.17: 45169040-45169040 on Build GRCh38
rs770399823 Chr.17: 45168966-45168966 on Build GRCh38
rs760708659 Chr.17: 45169055-45169055 on Build GRCh38
rs763238860 Chr.17: 45169286-45169288 on Build GRCh38
rs764558003 Chr.17: 45169387-45169387 on Build GRCh38
rs750013841 Chr.17: 45169104-45169104 on Build GRCh38
rs760054542 Chr.17: 45169281-45169281 on Build GRCh38
rs765650594 Chr.17: 45169064-45169065 on Build GRCh38
rs564827087 Chr.17: 45169138-45169138 on Build GRCh38
rs141737517 Chr.17: 45169274-45169274 on Build GRCh38
rs759100217 Chr.17: 45169378-45169378 on Build GRCh38
rs556056680 Chr.17: 45169117-45169117 on Build GRCh38
rs768921212 Chr.17: 45168986-45168986 on Build GRCh38
rs567654324 Chr.17: 45169330-45169330 on Build GRCh38
rs1055935242 Chr.17: 45169250-45169250 on Build GRCh38
rs755691733 Chr.17: 45169188-45169188 on Build GRCh38
rs149023557 Chr.17: 45169077-45169077 on Build GRCh38
rs1012832720 Chr.17: 45169163-45169163 on Build GRCh38
rs776577740 Chr.17: 45169073-45169073 on Build GRCh38
rs773017611 Chr.17: 45169028-45169028 on Build GRCh38
rs775525339 Chr.17: 45169165-45169165 on Build GRCh38
rs776571452 Chr.17: 45169140-45169140 on Build GRCh38
rs1032289939 Chr.17: 45168956-45168956 on Build GRCh38
rs75548629 Chr.17: 45169234-45169234 on Build GRCh38
rs771022140 Chr.17: 45169068-45169068 on Build GRCh38
rs773722760 Chr.17: 45169093-45169094 on Build GRCh38
rs760794704 Chr.17: 45169001-45169001 on Build GRCh38
rs752687454 Chr.17: 45169172-45169172 on Build GRCh38
rs773463146 Chr.17: 45169050-45169050 on Build GRCh38
rs753138219 Chr.17: 45169293-45169293 on Build GRCh38
rs867315326 Chr.17: 45169177-45169177 on Build GRCh38
rs957909206 Chr.17: 45169345-45169345 on Build GRCh38
rs542824190 Chr.17: 45168988-45168988 on Build GRCh38
rs768692619 Chr.17: 45169351-45169351 on Build GRCh38
rs749435782 Chr.17: 45169342-45169342 on Build GRCh38
rs200362016 Chr.17: 45169243-45169243 on Build GRCh38
rs552072619 Chr.17: 45169152-45169152 on Build GRCh38
rs574339066 Chr.17: 45169370-45169370 on Build GRCh38
rs763795808 Chr.17: 45169087-45169087 on Build GRCh38
rs34207107 Chr.17: 45169089-45169089 on Build GRCh38
rs1012384506 Chr.17: 45169278-45169278 on Build GRCh38
rs375887446 Chr.17: 45169175-45169175 on Build GRCh38
rs139634584 Chr.17: 45169389-45169389 on Build GRCh38
rs761550683 Chr.17: 45169173-45169173 on Build GRCh38
rs766758832 Chr.17: 45169314-45169315 on Build GRCh38
rs772950713 Chr.17: 45169279-45169279 on Build GRCh38
rs199878861 Chr.17: 45169080-45169080 on Build GRCh38
rs946340345 Chr.17: 45169046-45169046 on Build GRCh38
rs776178356 Chr.17: 45168971-45168971 on Build GRCh38
rs765928774 Chr.17: 45169287-45169287 on Build GRCh38
rs202210860 Chr.17: 45169170-45169170 on Build GRCh38
rs757683854 Chr.17: 45169325-45169325 on Build GRCh38
rs372907073 Chr.17: 45169344-45169344 on Build GRCh38
rs765289985 Chr.17: 45169079-45169079 on Build GRCh38
rs769271180 Chr.17: 45168975-45168976 on Build GRCh38
rs767046158 Chr.17: 45169275-45169275 on Build GRCh38
rs766089364 Chr.17: 45169371-45169371 on Build GRCh38
rs749772272 Chr.17: 45169038-45169038 on Build GRCh38
rs764953150 Chr.17: 45169182-45169182 on Build GRCh38
rs142750178 Chr.17: 45169129-45169129 on Build GRCh38
rs150581722 Chr.17: 45169332-45169332 on Build GRCh38
rs112826052 Chr.17: 45169130-45169130 on Build GRCh38
rs760380482 Chr.17: 45169365-45169365 on Build GRCh38
rs771049148 Chr.17: 45169229-45169229 on Build GRCh38
rs781530427 Chr.17: 45169233-45169233 on Build GRCh38
rs774836342 Chr.17: 45168991-45168993 on Build GRCh38
rs781218556 Chr.17: 45169327-45169327 on Build GRCh38

View Full Product Details