Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45270823-45271320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K14
Entrez Gene ID
9020
Gene Name
mitogen-activated protein kinase kinase kinase 14
Gene Aliases
FTDCR1B, HS, HSNIK, NIK
UniGene
Hs.404183
Gene Chromosome Location
Chr.17: 45263119-45317064 on Build GRCh38
Transcripts
NM_003954.4 XM_011525441.2

SNP Detail

SNP ID Chromosome Location
rs565220770 Chr.17: 45270979-45270980 on Build GRCh38
rs750836356 Chr.17: 45271106-45271106 on Build GRCh38
rs775109572 Chr.17: 45271249-45271249 on Build GRCh38
rs755279962 Chr.17: 45271047-45271047 on Build GRCh38
rs938147862 Chr.17: 45271002-45271002 on Build GRCh38
rs546776472 Chr.17: 45270972-45270972 on Build GRCh38
rs770924893 Chr.17: 45271219-45271219 on Build GRCh38
rs770568550 Chr.17: 45271061-45271061 on Build GRCh38
rs1009624584 Chr.17: 45271018-45271018 on Build GRCh38
rs779083529 Chr.17: 45271048-45271048 on Build GRCh38
rs764688712 Chr.17: 45271024-45271024 on Build GRCh38
rs11574830 Chr.17: 45271045-45271045 on Build GRCh38
rs957174587 Chr.17: 45270898-45270898 on Build GRCh38
rs757953953 Chr.17: 45271032-45271032 on Build GRCh38
rs986988109 Chr.17: 45271252-45271252 on Build GRCh38
rs762446609 Chr.17: 45271089-45271089 on Build GRCh38
rs766258829 Chr.17: 45270982-45270982 on Build GRCh38
rs368589648 Chr.17: 45271085-45271085 on Build GRCh38
rs762679399 Chr.17: 45271263-45271263 on Build GRCh38
rs1015841356 Chr.17: 45271273-45271273 on Build GRCh38
rs772228998 Chr.17: 45271049-45271049 on Build GRCh38
rs777910932 Chr.17: 45271130-45271131 on Build GRCh38
rs975981134 Chr.17: 45270905-45270905 on Build GRCh38
rs755349338 Chr.17: 45270983-45270984 on Build GRCh38
rs759194750 Chr.17: 45271023-45271023 on Build GRCh38
rs939983303 Chr.17: 45270976-45270976 on Build GRCh38
rs748561730 Chr.17: 45271125-45271126 on Build GRCh38
rs779242638 Chr.17: 45271166-45271166 on Build GRCh38
rs928848235 Chr.17: 45271289-45271289 on Build GRCh38
rs950764554 Chr.17: 45271169-45271169 on Build GRCh38
rs769644556 Chr.17: 45271242-45271242 on Build GRCh38
rs368083566 Chr.17: 45271034-45271034 on Build GRCh38
rs370019844 Chr.17: 45271279-45271279 on Build GRCh38
rs745632319 Chr.17: 45271233-45271233 on Build GRCh38
rs34171366 Chr.17: 45271041-45271041 on Build GRCh38
rs761246981 Chr.17: 45271116-45271116 on Build GRCh38
rs766610044 Chr.17: 45271121-45271121 on Build GRCh38
rs552468703 Chr.17: 45271055-45271055 on Build GRCh38
rs371145129 Chr.17: 45271076-45271076 on Build GRCh38
rs553580666 Chr.17: 45271156-45271156 on Build GRCh38
rs753030532 Chr.17: 45271183-45271183 on Build GRCh38
rs534894306 Chr.17: 45271090-45271090 on Build GRCh38
rs757382324 Chr.17: 45270875-45270875 on Build GRCh38
rs773929665 Chr.17: 45271185-45271185 on Build GRCh38
rs970149542 Chr.17: 45271260-45271260 on Build GRCh38
rs1044508517 Chr.17: 45270952-45270952 on Build GRCh38
rs751005102 Chr.17: 45271036-45271036 on Build GRCh38
rs754918334 Chr.17: 45271122-45271122 on Build GRCh38
rs781674912 Chr.17: 45271028-45271029 on Build GRCh38
rs753572716 Chr.17: 45271081-45271081 on Build GRCh38
rs577831952 Chr.17: 45270950-45270950 on Build GRCh38
rs546145436 Chr.17: 45270871-45270871 on Build GRCh38
rs767983116 Chr.17: 45271271-45271271 on Build GRCh38
rs139849447 Chr.17: 45270975-45270975 on Build GRCh38
rs768796911 Chr.17: 45270943-45270943 on Build GRCh38
rs770134033 Chr.17: 45271126-45271127 on Build GRCh38
rs909308443 Chr.17: 45271253-45271253 on Build GRCh38
rs950762595 Chr.17: 45270857-45270857 on Build GRCh38
rs371866775 Chr.17: 45271184-45271184 on Build GRCh38
rs773013851 Chr.17: 45271054-45271054 on Build GRCh38
rs746883259 Chr.17: 45271217-45271217 on Build GRCh38
rs550389689 Chr.17: 45271039-45271039 on Build GRCh38
rs755439399 Chr.17: 45271157-45271157 on Build GRCh38
rs746432224 Chr.17: 45271151-45271151 on Build GRCh38
rs564343681 Chr.17: 45270923-45270923 on Build GRCh38
rs996613711 Chr.17: 45271159-45271159 on Build GRCh38
rs749578780 Chr.17: 45270937-45270937 on Build GRCh38
rs957347930 Chr.17: 45271077-45271077 on Build GRCh38
rs770434730 Chr.17: 45271162-45271162 on Build GRCh38
rs776444527 Chr.17: 45271070-45271070 on Build GRCh38
rs777915925 Chr.17: 45271211-45271211 on Build GRCh38
rs781157704 Chr.17: 45271221-45271221 on Build GRCh38
rs773247049 Chr.17: 45270974-45270974 on Build GRCh38
rs776093851 Chr.17: 45271017-45271017 on Build GRCh38
rs529025769 Chr.17: 45271028-45271028 on Build GRCh38

View Full Product Details