Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76001939-76002440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK3
Entrez Gene ID
1018
Gene Name
cyclin dependent kinase 3
Gene Aliases
-
UniGene
Hs.706766
Gene Chromosome Location
Chr.17: 76000906-76005999 on Build GRCh38
Transcripts
NM_001258.2 NR_037709.1

Gene Symbol
TEN1-CDK3
Entrez Gene ID
100529145
Gene Name
TEN1-CDK3 readthrough (NMD candidate)
Gene Aliases
C17orf106-CDK3
UniGene
Hs.706766
Gene Chromosome Location
Chr.17: 75979217-76005999 on Build GRCh38
Transcripts
NM_001258.2 NR_037709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4935092 c.482A>G p.H161R Chr.17: 76002414-76002414 on Build GRCh38
3522379 c.192C>T p.V64V Chr.17: 76001949-76001949 on Build GRCh38
3522381 c.278C>T p.S93F Chr.17: 76002105-76002105 on Build GRCh38
5055787 c.297delC p.L101fs*15 Chr.17: 76002124-76002124 on Build GRCh38
3522380 c.247C>T p.L83F Chr.17: 76002074-76002074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372288050 Chr.17: 76002121-76002121 on Build GRCh38
rs773664890 Chr.17: 76002382-76002382 on Build GRCh38
rs201685548 Chr.17: 76002296-76002296 on Build GRCh38
rs201794034 Chr.17: 76002313-76002313 on Build GRCh38
rs757877562 Chr.17: 76002231-76002231 on Build GRCh38
rs777412070 Chr.17: 76001976-76001976 on Build GRCh38
rs761223956 Chr.17: 76002213-76002213 on Build GRCh38
rs780129446 Chr.17: 76002349-76002349 on Build GRCh38
rs764812585 Chr.17: 76002223-76002223 on Build GRCh38
rs1041369601 Chr.17: 76002314-76002314 on Build GRCh38
rs745652971 Chr.17: 76002147-76002150 on Build GRCh38
rs371481878 Chr.17: 76002212-76002212 on Build GRCh38
rs745355663 Chr.17: 76002004-76002004 on Build GRCh38
rs377464628 Chr.17: 76002376-76002376 on Build GRCh38
rs148982467 Chr.17: 76002395-76002395 on Build GRCh38
rs142329959 Chr.17: 76002344-76002344 on Build GRCh38
rs746058055 Chr.17: 76002254-76002254 on Build GRCh38
rs774617198 Chr.17: 76002197-76002197 on Build GRCh38
rs746925159 Chr.17: 76002139-76002139 on Build GRCh38
rs755912088 Chr.17: 76002238-76002238 on Build GRCh38
rs201655552 Chr.17: 76002405-76002405 on Build GRCh38
rs753726718 Chr.17: 76002411-76002411 on Build GRCh38
rs751010139 Chr.17: 76002233-76002233 on Build GRCh38
rs2069531 Chr.17: 76002196-76002196 on Build GRCh38
rs375401067 Chr.17: 76002219-76002219 on Build GRCh38
rs866091789 Chr.17: 76002308-76002308 on Build GRCh38
rs763937816 Chr.17: 76002406-76002406 on Build GRCh38
rs771254581 Chr.17: 76002058-76002058 on Build GRCh38
rs779004517 Chr.17: 76002252-76002252 on Build GRCh38
rs758337188 Chr.17: 76002005-76002005 on Build GRCh38
rs144506173 Chr.17: 76002046-76002046 on Build GRCh38
rs567520898 Chr.17: 76002037-76002037 on Build GRCh38
rs374525420 Chr.17: 76002403-76002403 on Build GRCh38
rs762715989 Chr.17: 76002297-76002297 on Build GRCh38
rs776003784 Chr.17: 76002103-76002103 on Build GRCh38
rs374666997 Chr.17: 76002187-76002187 on Build GRCh38
rs750026024 Chr.17: 76002229-76002229 on Build GRCh38
rs954274711 Chr.17: 76002042-76002042 on Build GRCh38
rs748605731 Chr.17: 76002371-76002371 on Build GRCh38
rs267605055 Chr.17: 76002126-76002126 on Build GRCh38
rs747107316 Chr.17: 76002352-76002352 on Build GRCh38
rs1028524830 Chr.17: 76002032-76002032 on Build GRCh38
rs761027879 Chr.17: 76002106-76002106 on Build GRCh38
rs781181073 Chr.17: 76002168-76002168 on Build GRCh38
rs757867913 Chr.17: 76002127-76002127 on Build GRCh38
rs780226863 Chr.17: 76002273-76002273 on Build GRCh38
rs756946506 Chr.17: 76002115-76002115 on Build GRCh38
rs201007083 Chr.17: 76002222-76002222 on Build GRCh38
rs779860792 Chr.17: 76002138-76002138 on Build GRCh38
rs1013779100 Chr.17: 76002008-76002008 on Build GRCh38
rs747919533 Chr.17: 76002178-76002178 on Build GRCh38
rs764867113 Chr.17: 76002119-76002119 on Build GRCh38
rs778770257 Chr.17: 76002064-76002064 on Build GRCh38
rs896420828 Chr.17: 76002245-76002245 on Build GRCh38
rs765915980 Chr.17: 76002298-76002298 on Build GRCh38
rs141186503 Chr.17: 76002040-76002040 on Build GRCh38
rs754841709 Chr.17: 76002143-76002143 on Build GRCh38
rs556171163 Chr.17: 76002043-76002043 on Build GRCh38
rs754183853 Chr.17: 76002109-76002109 on Build GRCh38
rs1039702806 Chr.17: 76002393-76002393 on Build GRCh38
rs558068161 Chr.17: 76002329-76002329 on Build GRCh38
rs368718895 Chr.17: 76001998-76001998 on Build GRCh38
rs777571429 Chr.17: 76002334-76002334 on Build GRCh38
rs536521071 Chr.17: 76002081-76002081 on Build GRCh38
rs777067823 Chr.17: 76002282-76002282 on Build GRCh38
rs951322395 Chr.17: 76002409-76002409 on Build GRCh38
rs201961284 Chr.17: 76002201-76002201 on Build GRCh38
rs373239489 Chr.17: 76002323-76002323 on Build GRCh38
rs145098458 Chr.17: 76001959-76001959 on Build GRCh38
rs774332704 Chr.17: 76002086-76002086 on Build GRCh38
rs1009961179 Chr.17: 76002256-76002256 on Build GRCh38
rs140567861 Chr.17: 76002044-76002044 on Build GRCh38
rs759655147 Chr.17: 76002088-76002088 on Build GRCh38
rs144754110 Chr.17: 76002370-76002370 on Build GRCh38
rs772166842 Chr.17: 76002270-76002270 on Build GRCh38
rs17880945 Chr.17: 76001987-76001987 on Build GRCh38
rs760154229 Chr.17: 76002271-76002271 on Build GRCh38
rs17881521 Chr.17: 76001988-76001988 on Build GRCh38
rs752611765 Chr.17: 76002320-76002320 on Build GRCh38
rs369829842 Chr.17: 76002380-76002380 on Build GRCh38
rs752595166 Chr.17: 76002235-76002235 on Build GRCh38
rs762171036 Chr.17: 76002286-76002286 on Build GRCh38
rs764397013 Chr.17: 76002108-76002108 on Build GRCh38
rs768656670 Chr.17: 76002362-76002362 on Build GRCh38
rs534113059 Chr.17: 76002034-76002034 on Build GRCh38
rs763221782 Chr.17: 76002388-76002388 on Build GRCh38
rs1007422043 Chr.17: 76002180-76002180 on Build GRCh38
rs368861061 Chr.17: 76002389-76002389 on Build GRCh38
rs937087926 Chr.17: 76002291-76002291 on Build GRCh38
rs764558017 Chr.17: 76002214-76002214 on Build GRCh38
rs767964507 Chr.17: 76002097-76002097 on Build GRCh38
rs759118252 Chr.17: 76002394-76002394 on Build GRCh38
rs770156329 Chr.17: 76002288-76002288 on Build GRCh38
rs771688809 Chr.17: 76002201-76002202 on Build GRCh38
rs145748425 Chr.17: 76002392-76002392 on Build GRCh38
rs368962392 Chr.17: 76002202-76002202 on Build GRCh38
rs769212297 Chr.17: 76002275-76002275 on Build GRCh38
rs972125613 Chr.17: 76002381-76002381 on Build GRCh38
rs1001427164 Chr.17: 76002031-76002031 on Build GRCh38
rs775177528 Chr.17: 76002248-76002260 on Build GRCh38
rs547553625 Chr.17: 76002036-76002036 on Build GRCh38
rs747891051 Chr.17: 76002092-76002092 on Build GRCh38
rs774077788 Chr.17: 76002039-76002041 on Build GRCh38
rs747090635 Chr.17: 76002274-76002274 on Build GRCh38
rs34918446 Chr.17: 76002303-76002303 on Build GRCh38
rs369742590 Chr.17: 76002309-76002309 on Build GRCh38
rs755004179 Chr.17: 76002234-76002234 on Build GRCh38
rs765863589 Chr.17: 76002232-76002232 on Build GRCh38

View Full Product Details