Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76353377-76353876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPSAP1
Entrez Gene ID
5635
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 1
Gene Aliases
PAP39
UniGene
Hs.77498
Gene Chromosome Location
Chr.17: 76310787-76354149 on Build GRCh38
Transcripts
NM_002766.2 XM_011525034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391246 c.7G>T p.A3S Chr.17: 76353610-76353610 on Build GRCh38
6391245 c.94G>T p.A32S Chr.17: 76353610-76353610 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs990168663 Chr.17: 76353846-76353846 on Build GRCh38
rs771880666 Chr.17: 76353496-76353496 on Build GRCh38
rs905042281 Chr.17: 76353743-76353743 on Build GRCh38
rs965558275 Chr.17: 76353624-76353624 on Build GRCh38
rs767321204 Chr.17: 76353632-76353632 on Build GRCh38
rs138247677 Chr.17: 76353637-76353637 on Build GRCh38
rs752382509 Chr.17: 76353634-76353634 on Build GRCh38
rs770541411 Chr.17: 76353795-76353795 on Build GRCh38
rs533391952 Chr.17: 76353659-76353659 on Build GRCh38
rs1040600228 Chr.17: 76353698-76353698 on Build GRCh38
rs150684296 Chr.17: 76353825-76353825 on Build GRCh38
rs769992731 Chr.17: 76353677-76353677 on Build GRCh38
rs779420316 Chr.17: 76353731-76353731 on Build GRCh38
rs369969047 Chr.17: 76353666-76353666 on Build GRCh38
rs549423653 Chr.17: 76353784-76353784 on Build GRCh38
rs748332694 Chr.17: 76353672-76353672 on Build GRCh38
rs768914187 Chr.17: 76353589-76353589 on Build GRCh38
rs866285631 Chr.17: 76353544-76353544 on Build GRCh38
rs141639978 Chr.17: 76353505-76353505 on Build GRCh38
rs931654392 Chr.17: 76353476-76353476 on Build GRCh38
rs754985420 Chr.17: 76353722-76353722 on Build GRCh38
rs1027012901 Chr.17: 76353412-76353412 on Build GRCh38
rs917057671 Chr.17: 76353450-76353450 on Build GRCh38
rs983787549 Chr.17: 76353619-76353619 on Build GRCh38
rs747423840 Chr.17: 76353766-76353766 on Build GRCh38
rs949886665 Chr.17: 76353645-76353645 on Build GRCh38
rs924960712 Chr.17: 76353486-76353486 on Build GRCh38
rs902120996 Chr.17: 76353719-76353719 on Build GRCh38
rs114276513 Chr.17: 76353434-76353434 on Build GRCh38
rs918348296 Chr.17: 76353433-76353433 on Build GRCh38
rs561753788 Chr.17: 76353746-76353746 on Build GRCh38
rs889075439 Chr.17: 76353675-76353675 on Build GRCh38
rs887100400 Chr.17: 76353754-76353754 on Build GRCh38
rs909780288 Chr.17: 76353440-76353440 on Build GRCh38
rs564402310 Chr.17: 76353641-76353641 on Build GRCh38
rs1019144912 Chr.17: 76353400-76353400 on Build GRCh38
rs1054409482 Chr.17: 76353496-76353496 on Build GRCh38
rs530389118 Chr.17: 76353566-76353566 on Build GRCh38
rs755375884 Chr.17: 76353628-76353628 on Build GRCh38
rs748270957 Chr.17: 76353770-76353770 on Build GRCh38
rs566984691 Chr.17: 76353692-76353692 on Build GRCh38
rs1056028738 Chr.17: 76353443-76353443 on Build GRCh38
rs780947358 Chr.17: 76353539-76353539 on Build GRCh38
rs376769808 Chr.17: 76353779-76353779 on Build GRCh38
rs773197500 Chr.17: 76353678-76353678 on Build GRCh38
rs557302132 Chr.17: 76353849-76353849 on Build GRCh38
rs935018351 Chr.17: 76353667-76353667 on Build GRCh38
rs764260026 Chr.17: 76353502-76353502 on Build GRCh38
rs776898411 Chr.17: 76353668-76353668 on Build GRCh38
rs751998561 Chr.17: 76353621-76353621 on Build GRCh38
rs770831023 Chr.17: 76353567-76353567 on Build GRCh38
rs546877754 Chr.17: 76353685-76353685 on Build GRCh38
rs759278025 Chr.17: 76353587-76353587 on Build GRCh38
rs901226332 Chr.17: 76353817-76353817 on Build GRCh38
rs976487687 Chr.17: 76353408-76353408 on Build GRCh38
rs769190777 Chr.17: 76353834-76353834 on Build GRCh38
rs1000140669 Chr.17: 76353488-76353488 on Build GRCh38
rs917173551 Chr.17: 76353638-76353638 on Build GRCh38
rs187023074 Chr.17: 76353837-76353837 on Build GRCh38
rs774133987 Chr.17: 76353500-76353500 on Build GRCh38
rs763374509 Chr.17: 76353600-76353600 on Build GRCh38
rs866165981 Chr.17: 76353591-76353591 on Build GRCh38
rs993660985 Chr.17: 76353736-76353736 on Build GRCh38
rs754713021 Chr.17: 76353536-76353536 on Build GRCh38
rs774252579 Chr.17: 76353855-76353855 on Build GRCh38
rs760886959 Chr.17: 76353501-76353501 on Build GRCh38
rs559438702 Chr.17: 76353504-76353504 on Build GRCh38
rs561510536 Chr.17: 76353528-76353528 on Build GRCh38
rs758832617 Chr.17: 76353472-76353472 on Build GRCh38
rs372518573 Chr.17: 76353753-76353753 on Build GRCh38
rs994083580 Chr.17: 76353531-76353531 on Build GRCh38
rs776801816 Chr.17: 76353592-76353592 on Build GRCh38
rs776333473 Chr.17: 76353802-76353802 on Build GRCh38
rs779937859 Chr.17: 76353607-76353607 on Build GRCh38
rs979479580 Chr.17: 76353479-76353479 on Build GRCh38
rs975690734 Chr.17: 76353468-76353468 on Build GRCh38
rs115365276 Chr.17: 76353829-76353829 on Build GRCh38
rs765811737 Chr.17: 76353516-76353516 on Build GRCh38
rs968922889 Chr.17: 76353832-76353832 on Build GRCh38
rs772445671 Chr.17: 76353750-76353750 on Build GRCh38
rs1040799778 Chr.17: 76353856-76353856 on Build GRCh38
rs1004195673 Chr.17: 76353768-76353768 on Build GRCh38
rs545979106 Chr.17: 76353438-76353438 on Build GRCh38
rs977279762 Chr.17: 76353831-76353831 on Build GRCh38
rs762017642 Chr.17: 76353598-76353598 on Build GRCh38
rs747713077 Chr.17: 76353545-76353545 on Build GRCh38
rs868747402 Chr.17: 76353580-76353580 on Build GRCh38
rs769158329 Chr.17: 76353555-76353555 on Build GRCh38
rs777822649 Chr.17: 76353561-76353561 on Build GRCh38
rs543273617 Chr.17: 76353727-76353727 on Build GRCh38
rs937283242 Chr.17: 76353442-76353442 on Build GRCh38
rs753960055 Chr.17: 76353483-76353483 on Build GRCh38
rs764142082 Chr.17: 76353847-76353847 on Build GRCh38
rs745701771 Chr.17: 76353653-76353653 on Build GRCh38
rs1045271521 Chr.17: 76353724-76353724 on Build GRCh38
rs763825576 Chr.17: 76353407-76353407 on Build GRCh38
rs771847777 Chr.17: 76353665-76353665 on Build GRCh38
rs765220128 Chr.17: 76353599-76353599 on Build GRCh38
rs866624969 Chr.17: 76353597-76353597 on Build GRCh38
rs774159301 Chr.17: 76353584-76353584 on Build GRCh38
rs910648196 Chr.17: 76353848-76353848 on Build GRCh38
rs868428787 Chr.17: 76353778-76353778 on Build GRCh38
rs949233864 Chr.17: 76353711-76353711 on Build GRCh38
rs750624348 Chr.17: 76353418-76353418 on Build GRCh38
rs147075140 Chr.17: 76353526-76353526 on Build GRCh38
rs551696186 Chr.17: 76353697-76353697 on Build GRCh38
rs755730825 Chr.17: 76353636-76353636 on Build GRCh38
rs949927951 Chr.17: 76353452-76353452 on Build GRCh38
rs1045719418 Chr.17: 76353629-76353629 on Build GRCh38
rs766869956 Chr.17: 76353608-76353608 on Build GRCh38
rs903280637 Chr.17: 76353475-76353475 on Build GRCh38

View Full Product Details