Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76736474-76736962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD11
Entrez Gene ID
79157
Gene Name
major facilitator superfamily domain containing 11
Gene Aliases
ET
UniGene
Hs.732019
Gene Chromosome Location
Chr.17: 76736565-76803805 on Build GRCh38
Transcripts
NM_001242534.1 NR_030366.1 NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

Gene Symbol
MIR636
Entrez Gene ID
693221
Gene Name
microRNA 636
Gene Aliases
MIRN636, hsa-mir-636
UniGene
Gene Chromosome Location
Chr.17: 76736450-76736548 on Build GRCh38
Transcripts
NM_001242534.1 NR_030366.1 NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

Gene Symbol
SRSF2
Entrez Gene ID
6427
Gene Name
serine and arginine rich splicing factor 2
Gene Aliases
PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b
UniGene
Hs.584801
Gene Chromosome Location
Chr.17: 76734115-76737431 on Build GRCh38
Transcripts
NM_001242534.1 NR_030366.1 NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408527 c.314C>T p.P105L Chr.17: 76736847-76736847 on Build GRCh38
6213192 c.302+7G>A p.? Chr.17: 76736792-76736792 on Build GRCh38
6223050 c.318A>G p.P106P Chr.17: 76736843-76736843 on Build GRCh38
4385020 c.230G>A p.G77E Chr.17: 76736931-76736931 on Build GRCh38
6965897 c.274T>C p.Y92H Chr.17: 76736887-76736887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949427164 Chr.17: 76736674-76736674 on Build GRCh38
rs769278515 Chr.17: 76736576-76736576 on Build GRCh38
rs901491098 Chr.17: 76736738-76736738 on Build GRCh38
rs1024568288 Chr.17: 76736622-76736622 on Build GRCh38
rs753844933 Chr.17: 76736912-76736912 on Build GRCh38
rs775685820 Chr.17: 76736493-76736493 on Build GRCh38
rs780851745 Chr.17: 76736791-76736791 on Build GRCh38
rs1050014392 Chr.17: 76736873-76736873 on Build GRCh38
rs548868861 Chr.17: 76736663-76736663 on Build GRCh38
rs764750794 Chr.17: 76736502-76736502 on Build GRCh38
rs770680008 Chr.17: 76736580-76736580 on Build GRCh38
rs762284664 Chr.17: 76736851-76736851 on Build GRCh38
rs762261079 Chr.17: 76736591-76736591 on Build GRCh38
rs756714282 Chr.17: 76736501-76736501 on Build GRCh38
rs757305177 Chr.17: 76736544-76736544 on Build GRCh38
rs908674875 Chr.17: 76736820-76736820 on Build GRCh38
rs751713049 Chr.17: 76736877-76736877 on Build GRCh38
rs750310954 Chr.17: 76736519-76736532 on Build GRCh38
rs931513044 Chr.17: 76736808-76736808 on Build GRCh38
rs761166094 Chr.17: 76736538-76736538 on Build GRCh38
rs760680088 Chr.17: 76736900-76736900 on Build GRCh38
rs979272036 Chr.17: 76736666-76736666 on Build GRCh38
rs754741757 Chr.17: 76736780-76736780 on Build GRCh38
rs763006490 Chr.17: 76736869-76736869 on Build GRCh38
rs976254083 Chr.17: 76736815-76736815 on Build GRCh38
rs750478335 Chr.17: 76736596-76736596 on Build GRCh38
rs1026735111 Chr.17: 76736507-76736507 on Build GRCh38
rs773771490 Chr.17: 76736867-76736867 on Build GRCh38
rs377064670 Chr.17: 76736542-76736542 on Build GRCh38
rs750602613 Chr.17: 76736545-76736545 on Build GRCh38
rs977762039 Chr.17: 76736630-76736630 on Build GRCh38
rs536602994 Chr.17: 76736706-76736706 on Build GRCh38
rs887566542 Chr.17: 76736634-76736634 on Build GRCh38
rs1025085922 Chr.17: 76736764-76736764 on Build GRCh38
rs998771977 Chr.17: 76736750-76736750 on Build GRCh38
rs910340182 Chr.17: 76736667-76736667 on Build GRCh38
rs766200080 Chr.17: 76736854-76736877 on Build GRCh38
rs971312072 Chr.17: 76736628-76736628 on Build GRCh38
rs779853165 Chr.17: 76736512-76736512 on Build GRCh38
rs923446001 Chr.17: 76736696-76736696 on Build GRCh38
rs754830314 Chr.17: 76736559-76736559 on Build GRCh38
rs761880835 Chr.17: 76736510-76736511 on Build GRCh38
rs1017230226 Chr.17: 76736781-76736781 on Build GRCh38
rs777172872 Chr.17: 76736578-76736578 on Build GRCh38
rs1052900167 Chr.17: 76736617-76736617 on Build GRCh38
rs910710355 Chr.17: 76736579-76736579 on Build GRCh38
rs981007517 Chr.17: 76736676-76736676 on Build GRCh38
rs768995336 Chr.17: 76736629-76736629 on Build GRCh38
rs754601892 Chr.17: 76736873-76736874 on Build GRCh38
rs535244752 Chr.17: 76736650-76736650 on Build GRCh38
rs958526203 Chr.17: 76736618-76736618 on Build GRCh38
rs770165979 Chr.17: 76736861-76736861 on Build GRCh38
rs931467362 Chr.17: 76736680-76736680 on Build GRCh38
rs774219066 Chr.17: 76736581-76736581 on Build GRCh38
rs529999340 Chr.17: 76736553-76736553 on Build GRCh38
rs932978794 Chr.17: 76736849-76736849 on Build GRCh38
rs563622861 Chr.17: 76736598-76736598 on Build GRCh38
rs151217137 Chr.17: 76736703-76736703 on Build GRCh38
rs1013691314 Chr.17: 76736739-76736739 on Build GRCh38
rs755985389 Chr.17: 76736807-76736807 on Build GRCh38
rs935742869 Chr.17: 76736560-76736560 on Build GRCh38
rs775978250 Chr.17: 76736536-76736536 on Build GRCh38
rs748814345 Chr.17: 76736548-76736548 on Build GRCh38
rs931498432 Chr.17: 76736632-76736632 on Build GRCh38
rs765186728 Chr.17: 76736517-76736535 on Build GRCh38
rs993549885 Chr.17: 76736506-76736506 on Build GRCh38
rs556956650 Chr.17: 76736707-76736707 on Build GRCh38
rs746774617 Chr.17: 76736557-76736557 on Build GRCh38
rs763059673 Chr.17: 76736754-76736754 on Build GRCh38
rs561929258 Chr.17: 76736687-76736687 on Build GRCh38
rs751363805 Chr.17: 76736861-76736878 on Build GRCh38
rs1048824664 Chr.17: 76736633-76736633 on Build GRCh38
rs997214274 Chr.17: 76736742-76736742 on Build GRCh38
rs762727557 Chr.17: 76736525-76736525 on Build GRCh38
rs957030560 Chr.17: 76736770-76736770 on Build GRCh38
rs769242167 Chr.17: 76736521-76736521 on Build GRCh38
rs532688400 Chr.17: 76736615-76736615 on Build GRCh38
rs971724555 Chr.17: 76736541-76736541 on Build GRCh38
rs199683955 Chr.17: 76736505-76736505 on Build GRCh38
rs1043146559 Chr.17: 76736686-76736686 on Build GRCh38
rs768558412 Chr.17: 76736515-76736515 on Build GRCh38
rs752405402 Chr.17: 76736878-76736878 on Build GRCh38
rs777901001 Chr.17: 76736816-76736816 on Build GRCh38
rs765310328 Chr.17: 76736594-76736594 on Build GRCh38
rs1055133139 Chr.17: 76736732-76736732 on Build GRCh38
rs769026966 Chr.17: 76736843-76736843 on Build GRCh38
rs895199480 Chr.17: 76736734-76736734 on Build GRCh38
rs550160657 Chr.17: 76736583-76736583 on Build GRCh38
rs772860153 Chr.17: 76736524-76736524 on Build GRCh38
rs1007596585 Chr.17: 76736517-76736517 on Build GRCh38
rs976507913 Chr.17: 76736624-76736647 on Build GRCh38
rs940411297 Chr.17: 76736714-76736714 on Build GRCh38
rs764657547 Chr.17: 76736539-76736539 on Build GRCh38
rs949630594 Chr.17: 76736624-76736624 on Build GRCh38
rs776469568 Chr.17: 76736519-76736519 on Build GRCh38
rs1048472740 Chr.17: 76736704-76736704 on Build GRCh38
rs770622960 Chr.17: 76736825-76736825 on Build GRCh38
rs917889791 Chr.17: 76736673-76736673 on Build GRCh38
rs989848999 Chr.17: 76736655-76736655 on Build GRCh38
rs976027453 Chr.17: 76736592-76736592 on Build GRCh38
rs1045340694 Chr.17: 76736626-76736626 on Build GRCh38
rs1004642046 Chr.17: 76736637-76736637 on Build GRCh38
rs552667347 Chr.17: 76736796-76736796 on Build GRCh38
rs879534548 Chr.17: 76736586-76736586 on Build GRCh38
rs576861200 Chr.17: 76736743-76736743 on Build GRCh38
rs1029048657 Chr.17: 76736679-76736701 on Build GRCh38
rs1014834570 Chr.17: 76736588-76736588 on Build GRCh38
rs953194620 Chr.17: 76736658-76736658 on Build GRCh38
rs777130901 Chr.17: 76736585-76736585 on Build GRCh38
rs747685241 Chr.17: 76736520-76736520 on Build GRCh38
rs1012716864 Chr.17: 76736619-76736619 on Build GRCh38
rs976186117 Chr.17: 76736682-76736682 on Build GRCh38
rs537883100 Chr.17: 76736819-76736819 on Build GRCh38
rs781172050 Chr.17: 76736563-76736563 on Build GRCh38
rs758640296 Chr.17: 76736547-76736547 on Build GRCh38
rs552780754 Chr.17: 76736638-76736638 on Build GRCh38
rs761108823 Chr.17: 76736582-76736582 on Build GRCh38
rs957294548 Chr.17: 76736644-76736644 on Build GRCh38
rs1000715601 Chr.17: 76736918-76736918 on Build GRCh38
rs541687424 Chr.17: 76736535-76736535 on Build GRCh38
rs779792125 Chr.17: 76736566-76736566 on Build GRCh38
rs1004980521 Chr.17: 76736587-76736587 on Build GRCh38
rs1006291460 Chr.17: 76736593-76736593 on Build GRCh38
rs943123753 Chr.17: 76736669-76736669 on Build GRCh38
rs935342207 Chr.17: 76736616-76736616 on Build GRCh38
rs776880365 Chr.17: 76736844-76736844 on Build GRCh38
rs369604599 Chr.17: 76736510-76736510 on Build GRCh38
rs566309566 Chr.17: 76736647-76736647 on Build GRCh38
rs1036126519 Chr.17: 76736726-76736726 on Build GRCh38
rs766578856 Chr.17: 76736776-76736776 on Build GRCh38
rs971118278 Chr.17: 76736788-76736788 on Build GRCh38
rs766090246 Chr.17: 76736933-76736933 on Build GRCh38
rs778651965 Chr.17: 76736831-76736831 on Build GRCh38
rs976353866 Chr.17: 76736494-76736494 on Build GRCh38
rs953224132 Chr.17: 76736605-76736605 on Build GRCh38
rs559151830 Chr.17: 76736513-76736513 on Build GRCh38
rs539555945 Chr.17: 76736778-76736778 on Build GRCh38
rs762854633 Chr.17: 76736542-76736544 on Build GRCh38
rs907056127 Chr.17: 76736746-76736746 on Build GRCh38
rs762750742 Chr.17: 76736730-76736730 on Build GRCh38
rs988605293 Chr.17: 76736758-76736762 on Build GRCh38
rs766459684 Chr.17: 76736870-76736870 on Build GRCh38
rs1047694766 Chr.17: 76736589-76736589 on Build GRCh38
rs988248974 Chr.17: 76736671-76736671 on Build GRCh38
rs756005065 Chr.17: 76736885-76736885 on Build GRCh38
rs953594529 Chr.17: 76736748-76736768 on Build GRCh38
rs745521484 Chr.17: 76736834-76736834 on Build GRCh38
rs757347241 Chr.17: 76736509-76736509 on Build GRCh38
rs372808564 Chr.17: 76736516-76736516 on Build GRCh38

View Full Product Details