Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80223167-80223659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC26A11
Entrez Gene ID
284129
Gene Name
solute carrier family 26 member 11
Gene Aliases
-
UniGene
Hs.4866
Gene Chromosome Location
Chr.17: 80220287-80253509 on Build GRCh38
Transcripts
NM_001166347.1 NM_001166348.1 NM_001166349.1 NM_173626.3 XM_006721833.1 XM_011524652.1 XM_011524653.1 XM_011524654.1 XM_017024505.1 XM_017024506.1 XM_017024507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5504389 c.428-9G>A p.? Chr.17: 80223243-80223243 on Build GRCh38
7424134 c.435G>T p.L145L Chr.17: 80223259-80223259 on Build GRCh38
6856312 c.457G>A p.V153I Chr.17: 80223281-80223281 on Build GRCh38
5981319 c.507G>T p.Q169H Chr.17: 80223331-80223331 on Build GRCh38
4726899 c.483T>C p.A161A Chr.17: 80223307-80223307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746273040 Chr.17: 80223233-80223233 on Build GRCh38
rs575090026 Chr.17: 80223484-80223484 on Build GRCh38
rs757796994 Chr.17: 80223305-80223305 on Build GRCh38
rs536655493 Chr.17: 80223331-80223331 on Build GRCh38
rs577677706 Chr.17: 80223624-80223624 on Build GRCh38
rs760268559 Chr.17: 80223359-80223359 on Build GRCh38
rs118084276 Chr.17: 80223613-80223613 on Build GRCh38
rs553626825 Chr.17: 80223552-80223552 on Build GRCh38
rs893224051 Chr.17: 80223354-80223354 on Build GRCh38
rs200793605 Chr.17: 80223278-80223278 on Build GRCh38
rs144577972 Chr.17: 80223317-80223317 on Build GRCh38
rs140824671 Chr.17: 80223298-80223298 on Build GRCh38
rs773204443 Chr.17: 80223345-80223345 on Build GRCh38
rs372688197 Chr.17: 80223218-80223218 on Build GRCh38
rs138520983 Chr.17: 80223319-80223319 on Build GRCh38
rs751000853 Chr.17: 80223382-80223382 on Build GRCh38
rs750061094 Chr.17: 80223206-80223206 on Build GRCh38
rs967317412 Chr.17: 80223486-80223486 on Build GRCh38
rs752202894 Chr.17: 80223353-80223353 on Build GRCh38
rs890783083 Chr.17: 80223400-80223400 on Build GRCh38
rs762818859 Chr.17: 80223280-80223280 on Build GRCh38
rs117339976 Chr.17: 80223371-80223371 on Build GRCh38
rs761780106 Chr.17: 80223268-80223268 on Build GRCh38
rs999810309 Chr.17: 80223383-80223383 on Build GRCh38
rs867080518 Chr.17: 80223473-80223473 on Build GRCh38
rs772130174 Chr.17: 80223234-80223234 on Build GRCh38
rs990493290 Chr.17: 80223237-80223237 on Build GRCh38
rs903148192 Chr.17: 80223601-80223601 on Build GRCh38
rs1031041327 Chr.17: 80223499-80223499 on Build GRCh38
rs765408204 Chr.17: 80223383-80223385 on Build GRCh38
rs893237336 Chr.17: 80223571-80223571 on Build GRCh38
rs960997868 Chr.17: 80223513-80223513 on Build GRCh38
rs558709397 Chr.17: 80223391-80223391 on Build GRCh38
rs573410262 Chr.17: 80223579-80223579 on Build GRCh38
rs371733210 Chr.17: 80223540-80223540 on Build GRCh38
rs148133286 Chr.17: 80223528-80223528 on Build GRCh38
rs532485121 Chr.17: 80223303-80223303 on Build GRCh38
rs367684406 Chr.17: 80223311-80223311 on Build GRCh38
rs898762402 Chr.17: 80223375-80223375 on Build GRCh38
rs538351608 Chr.17: 80223428-80223428 on Build GRCh38
rs754521004 Chr.17: 80223214-80223214 on Build GRCh38
rs757670522 Chr.17: 80223223-80223223 on Build GRCh38
rs73429464 Chr.17: 80223349-80223349 on Build GRCh38
rs374761070 Chr.17: 80223443-80223443 on Build GRCh38
rs143955079 Chr.17: 80223487-80223487 on Build GRCh38
rs930213069 Chr.17: 80223376-80223376 on Build GRCh38
rs1041721441 Chr.17: 80223582-80223582 on Build GRCh38
rs757949125 Chr.17: 80223209-80223209 on Build GRCh38
rs577405821 Chr.17: 80223420-80223420 on Build GRCh38
rs766076262 Chr.17: 80223210-80223210 on Build GRCh38
rs371524422 Chr.17: 80223348-80223348 on Build GRCh38
rs1020267192 Chr.17: 80223378-80223378 on Build GRCh38
rs368317235 Chr.17: 80223217-80223217 on Build GRCh38
rs1035386246 Chr.17: 80223605-80223605 on Build GRCh38
rs1051291745 Chr.17: 80223333-80223333 on Build GRCh38
rs767644878 Chr.17: 80223538-80223538 on Build GRCh38
rs769659229 Chr.17: 80223334-80223334 on Build GRCh38
rs765879820 Chr.17: 80223283-80223283 on Build GRCh38
rs564299523 Chr.17: 80223606-80223606 on Build GRCh38
rs575009277 Chr.17: 80223314-80223314 on Build GRCh38
rs143824845 Chr.17: 80223253-80223253 on Build GRCh38
rs774660252 Chr.17: 80223281-80223281 on Build GRCh38
rs755648076 Chr.17: 80223521-80223521 on Build GRCh38
rs374680442 Chr.17: 80223273-80223273 on Build GRCh38
rs759099809 Chr.17: 80223300-80223300 on Build GRCh38
rs912806369 Chr.17: 80223527-80223527 on Build GRCh38
rs974237840 Chr.17: 80223541-80223541 on Build GRCh38
rs1051751228 Chr.17: 80223505-80223506 on Build GRCh38
rs762651109 Chr.17: 80223346-80223346 on Build GRCh38
rs1018450295 Chr.17: 80223439-80223439 on Build GRCh38
rs148979132 Chr.17: 80223252-80223252 on Build GRCh38
rs955178083 Chr.17: 80223189-80223189 on Build GRCh38
rs1002866213 Chr.17: 80223603-80223603 on Build GRCh38
rs150676297 Chr.17: 80223565-80223565 on Build GRCh38
rs1047753567 Chr.17: 80223577-80223577 on Build GRCh38
rs781211086 Chr.17: 80223320-80223320 on Build GRCh38
rs779456643 Chr.17: 80223228-80223228 on Build GRCh38
rs1011643614 Chr.17: 80223578-80223578 on Build GRCh38
rs557020714 Chr.17: 80223444-80223444 on Build GRCh38
rs143511610 Chr.17: 80223262-80223262 on Build GRCh38
rs141954243 Chr.17: 80223558-80223558 on Build GRCh38
rs779579035 Chr.17: 80223310-80223310 on Build GRCh38
rs909463987 Chr.17: 80223562-80223562 on Build GRCh38
rs750974075 Chr.17: 80223212-80223212 on Build GRCh38

View Full Product Details