Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80246062-80246559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC26A11
Entrez Gene ID
284129
Gene Name
solute carrier family 26 member 11
Gene Aliases
-
UniGene
Hs.4866
Gene Chromosome Location
Chr.17: 80220287-80253509 on Build GRCh38
Transcripts
NM_001166347.1 NM_001166348.1 NM_001166349.1 NM_173626.3 XM_006721833.1 XM_011524652.1 XM_011524653.1 XM_011524654.1 XM_017024505.1 XM_017024506.1 XM_017024507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5910951 c.1098-7C>T p.? Chr.17: 80246147-80246147 on Build GRCh38
4726902 c.1136_1137insG p.L382fs*50 Chr.17: 80246192-80246193 on Build GRCh38
6780927 c.1102G>T p.A368S Chr.17: 80246158-80246158 on Build GRCh38
5571332 c.1105G>A p.V369M Chr.17: 80246161-80246161 on Build GRCh38
3672615 c.1136C>A p.A379E Chr.17: 80246192-80246192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190164309 Chr.17: 80246410-80246410 on Build GRCh38
rs758972363 Chr.17: 80246497-80246497 on Build GRCh38
rs770727130 Chr.17: 80246486-80246486 on Build GRCh38
rs756905343 Chr.17: 80246466-80246467 on Build GRCh38
rs148525144 Chr.17: 80246160-80246160 on Build GRCh38
rs745943087 Chr.17: 80246189-80246189 on Build GRCh38
rs754174767 Chr.17: 80246175-80246175 on Build GRCh38
rs565984961 Chr.17: 80246451-80246451 on Build GRCh38
rs755075019 Chr.17: 80246529-80246529 on Build GRCh38
rs555866920 Chr.17: 80246216-80246216 on Build GRCh38
rs755767772 Chr.17: 80246460-80246461 on Build GRCh38
rs187677879 Chr.17: 80246308-80246308 on Build GRCh38
rs927937230 Chr.17: 80246228-80246228 on Build GRCh38
rs200625063 Chr.17: 80246133-80246133 on Build GRCh38
rs562132181 Chr.17: 80246377-80246377 on Build GRCh38
rs779006768 Chr.17: 80246183-80246183 on Build GRCh38
rs374707575 Chr.17: 80246174-80246174 on Build GRCh38
rs750810741 Chr.17: 80246510-80246510 on Build GRCh38
rs941516313 Chr.17: 80246272-80246272 on Build GRCh38
rs910076833 Chr.17: 80246268-80246268 on Build GRCh38
rs1037298945 Chr.17: 80246365-80246365 on Build GRCh38
rs1020806822 Chr.17: 80246304-80246304 on Build GRCh38
rs1006667966 Chr.17: 80246299-80246299 on Build GRCh38
rs758652476 Chr.17: 80246255-80246255 on Build GRCh38
rs531435822 Chr.17: 80246313-80246313 on Build GRCh38
rs942337142 Chr.17: 80246456-80246456 on Build GRCh38
rs777551332 Chr.17: 80246495-80246495 on Build GRCh38
rs763016810 Chr.17: 80246204-80246206 on Build GRCh38
rs889430447 Chr.17: 80246292-80246292 on Build GRCh38
rs751654101 Chr.17: 80246462-80246462 on Build GRCh38
rs77106895 Chr.17: 80246155-80246155 on Build GRCh38
rs34818744 Chr.17: 80246109-80246109 on Build GRCh38
rs746784872 Chr.17: 80246120-80246120 on Build GRCh38
rs750560049 Chr.17: 80246109-80246109 on Build GRCh38
rs7217586 Chr.17: 80246086-80246086 on Build GRCh38
rs918732342 Chr.17: 80246434-80246434 on Build GRCh38
rs1054910852 Chr.17: 80246415-80246415 on Build GRCh38
rs959184813 Chr.17: 80246315-80246315 on Build GRCh38
rs58412367 Chr.17: 80246505-80246505 on Build GRCh38
rs141146194 Chr.17: 80246166-80246166 on Build GRCh38
rs541390179 Chr.17: 80246286-80246286 on Build GRCh38
rs371729311 Chr.17: 80246224-80246224 on Build GRCh38
rs913747784 Chr.17: 80246095-80246095 on Build GRCh38
rs746911139 Chr.17: 80246198-80246198 on Build GRCh38
rs766310412 Chr.17: 80246211-80246213 on Build GRCh38
rs934248550 Chr.17: 80246436-80246436 on Build GRCh38
rs567774859 Chr.17: 80246516-80246516 on Build GRCh38
rs747965657 Chr.17: 80246471-80246471 on Build GRCh38
rs773060485 Chr.17: 80246211-80246211 on Build GRCh38
rs374231395 Chr.17: 80246312-80246312 on Build GRCh38
rs1051329937 Chr.17: 80246450-80246450 on Build GRCh38
rs199912887 Chr.17: 80246179-80246179 on Build GRCh38
rs757441989 Chr.17: 80246181-80246181 on Build GRCh38
rs781062288 Chr.17: 80246468-80246468 on Build GRCh38
rs72853684 Chr.17: 80246334-80246334 on Build GRCh38
rs537218999 Chr.17: 80246492-80246492 on Build GRCh38
rs368506262 Chr.17: 80246116-80246116 on Build GRCh38
rs35712944 Chr.17: 80246205-80246206 on Build GRCh38
rs913925454 Chr.17: 80246092-80246092 on Build GRCh38
rs75009752 Chr.17: 80246421-80246421 on Build GRCh38
rs111963541 Chr.17: 80246480-80246480 on Build GRCh38
rs768538219 Chr.17: 80246132-80246132 on Build GRCh38
rs771657842 Chr.17: 80246404-80246404 on Build GRCh38
rs553355999 Chr.17: 80246300-80246300 on Build GRCh38
rs764520809 Chr.17: 80246236-80246236 on Build GRCh38
rs1041957339 Chr.17: 80246484-80246484 on Build GRCh38
rs76617433 Chr.17: 80246284-80246284 on Build GRCh38
rs112831546 Chr.17: 80246507-80246507 on Build GRCh38
rs922877321 Chr.17: 80246373-80246373 on Build GRCh38
rs759207972 Chr.17: 80246238-80246238 on Build GRCh38
rs752510279 Chr.17: 80246251-80246252 on Build GRCh38
rs984241311 Chr.17: 80246330-80246331 on Build GRCh38
rs375180567 Chr.17: 80246114-80246114 on Build GRCh38
rs751442949 Chr.17: 80246211-80246212 on Build GRCh38
rs1032863312 Chr.17: 80246190-80246190 on Build GRCh38
rs537929586 Chr.17: 80246207-80246207 on Build GRCh38
rs746573931 Chr.17: 80246498-80246498 on Build GRCh38
rs768502056 Chr.17: 80246201-80246201 on Build GRCh38
rs754827712 Chr.17: 80246216-80246226 on Build GRCh38
rs934194469 Chr.17: 80246285-80246285 on Build GRCh38
rs367624389 Chr.17: 80246208-80246208 on Build GRCh38
rs1035004795 Chr.17: 80246376-80246376 on Build GRCh38
rs539743945 Chr.17: 80246167-80246167 on Build GRCh38
rs761193637 Chr.17: 80246227-80246227 on Build GRCh38
rs147759020 Chr.17: 80246161-80246161 on Build GRCh38
rs750508292 Chr.17: 80246135-80246135 on Build GRCh38
rs758522694 Chr.17: 80246191-80246191 on Build GRCh38
rs570207728 Chr.17: 80246491-80246491 on Build GRCh38
rs370173673 Chr.17: 80246303-80246303 on Build GRCh38
rs780196773 Chr.17: 80246253-80246253 on Build GRCh38
rs780061511 Chr.17: 80246458-80246458 on Build GRCh38
rs928051520 Chr.17: 80246115-80246115 on Build GRCh38
rs1012203624 Chr.17: 80246517-80246517 on Build GRCh38
rs747927879 Chr.17: 80246395-80246395 on Build GRCh38
rs1010625733 Chr.17: 80246418-80246418 on Build GRCh38
rs535219796 Chr.17: 80246520-80246520 on Build GRCh38
rs981298090 Chr.17: 80246336-80246336 on Build GRCh38
rs142863782 Chr.17: 80246535-80246535 on Build GRCh38
rs780813210 Chr.17: 80246247-80246248 on Build GRCh38
rs1037324179 Chr.17: 80246281-80246281 on Build GRCh38
rs925741830 Chr.17: 80246082-80246082 on Build GRCh38
rs936108580 Chr.17: 80246411-80246411 on Build GRCh38
rs776431841 Chr.17: 80246205-80246205 on Build GRCh38
rs146947468 Chr.17: 80246307-80246307 on Build GRCh38
rs1032285977 Chr.17: 80246146-80246146 on Build GRCh38
rs957322393 Chr.17: 80246083-80246083 on Build GRCh38
rs945357450 Chr.17: 80246108-80246108 on Build GRCh38
rs762525575 Chr.17: 80246212-80246212 on Build GRCh38
rs777062452 Chr.17: 80246501-80246501 on Build GRCh38
rs547764311 Chr.17: 80246426-80246426 on Build GRCh38
rs940290673 Chr.17: 80246188-80246188 on Build GRCh38
rs756400489 Chr.17: 80246171-80246171 on Build GRCh38
rs373430553 Chr.17: 80246504-80246504 on Build GRCh38
rs891815929 Chr.17: 80246417-80246417 on Build GRCh38
rs990895285 Chr.17: 80246398-80246398 on Build GRCh38
rs779936680 Chr.17: 80246196-80246196 on Build GRCh38
rs1049962581 Chr.17: 80246403-80246404 on Build GRCh38
rs748905494 Chr.17: 80246485-80246485 on Build GRCh38
rs777532950 Chr.17: 80246483-80246483 on Build GRCh38
rs377404252 Chr.17: 80246290-80246290 on Build GRCh38
rs934214000 Chr.17: 80246397-80246397 on Build GRCh38
rs962785568 Chr.17: 80246420-80246420 on Build GRCh38
rs770602275 Chr.17: 80246222-80246222 on Build GRCh38
rs533665848 Chr.17: 80246466-80246466 on Build GRCh38
rs570193518 Chr.17: 80246192-80246192 on Build GRCh38
rs751658402 Chr.17: 80246527-80246527 on Build GRCh38

View Full Product Details