Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81239154-81239642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf89
Entrez Gene ID
284184
Gene Name
chromosome 17 open reading frame 89
Gene Aliases
-
UniGene
Hs.356545
Gene Chromosome Location
Chr.17: 81239311-81241298 on Build GRCh38
Transcripts
NM_001086521.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318167 c.150C>T p.F50F Chr.17: 81239633-81239633 on Build GRCh38
6408910 c.8C>T p.A3V Chr.17: 81239371-81239371 on Build GRCh38
6410768 c.97G>A p.G33S Chr.17: 81239580-81239580 on Build GRCh38
6391440 c.110A>T p.Q37L Chr.17: 81239593-81239593 on Build GRCh38
5038053 c.148T>C p.F50L Chr.17: 81239631-81239631 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs976701620 Chr.17: 81239403-81239403 on Build GRCh38
rs765426612 Chr.17: 81239360-81239360 on Build GRCh38
rs901084116 Chr.17: 81239596-81239596 on Build GRCh38
rs757211482 Chr.17: 81239573-81239573 on Build GRCh38
rs371087327 Chr.17: 81239361-81239361 on Build GRCh38
rs745674132 Chr.17: 81239325-81239325 on Build GRCh38
rs888243307 Chr.17: 81239555-81239555 on Build GRCh38
rs367930497 Chr.17: 81239485-81239485 on Build GRCh38
rs568014963 Chr.17: 81239394-81239394 on Build GRCh38
rs932776839 Chr.17: 81239301-81239301 on Build GRCh38
rs952593458 Chr.17: 81239375-81239375 on Build GRCh38
rs756358496 Chr.17: 81239495-81239495 on Build GRCh38
rs370099258 Chr.17: 81239528-81239528 on Build GRCh38
rs1043605607 Chr.17: 81239324-81239324 on Build GRCh38
rs988425462 Chr.17: 81239417-81239417 on Build GRCh38
rs780328302 Chr.17: 81239302-81239302 on Build GRCh38
rs1002442907 Chr.17: 81239216-81239216 on Build GRCh38
rs1048146456 Chr.17: 81239546-81239546 on Build GRCh38
rs543919003 Chr.17: 81239221-81239221 on Build GRCh38
rs978885560 Chr.17: 81239432-81239432 on Build GRCh38
rs965351413 Chr.17: 81239235-81239235 on Build GRCh38
rs776680789 Chr.17: 81239319-81239319 on Build GRCh38
rs958426509 Chr.17: 81239393-81239393 on Build GRCh38
rs976755469 Chr.17: 81239255-81239255 on Build GRCh38
rs557714636 Chr.17: 81239175-81239175 on Build GRCh38
rs377540343 Chr.17: 81239391-81239391 on Build GRCh38
rs1001987458 Chr.17: 81239366-81239366 on Build GRCh38
rs917504587 Chr.17: 81239494-81239494 on Build GRCh38
rs921345537 Chr.17: 81239264-81239264 on Build GRCh38
rs552240072 Chr.17: 81239357-81239357 on Build GRCh38
rs921292824 Chr.17: 81239415-81239415 on Build GRCh38
rs779334116 Chr.17: 81239287-81239287 on Build GRCh38
rs529910138 Chr.17: 81239238-81239238 on Build GRCh38
rs74778772 Chr.17: 81239341-81239341 on Build GRCh38
rs374420845 Chr.17: 81239603-81239603 on Build GRCh38
rs376937982 Chr.17: 81239260-81239260 on Build GRCh38
rs949386857 Chr.17: 81239309-81239309 on Build GRCh38
rs531171371 Chr.17: 81239380-81239380 on Build GRCh38
rs934887146 Chr.17: 81239340-81239340 on Build GRCh38
rs2659009 Chr.17: 81239505-81239505 on Build GRCh38
rs573584898 Chr.17: 81239221-81239221 on Build GRCh38
rs558138829 Chr.17: 81239610-81239610 on Build GRCh38
rs1046084819 Chr.17: 81239511-81239511 on Build GRCh38
rs1015404982 Chr.17: 81239563-81239563 on Build GRCh38
rs776068290 Chr.17: 81239377-81239377 on Build GRCh38
rs367877316 Chr.17: 81239620-81239620 on Build GRCh38
rs982732088 Chr.17: 81239227-81239227 on Build GRCh38
rs576488503 Chr.17: 81239186-81239186 on Build GRCh38
rs759930766 Chr.17: 81239420-81239420 on Build GRCh38
rs541999457 Chr.17: 81239261-81239261 on Build GRCh38
rs1032590832 Chr.17: 81239262-81239262 on Build GRCh38
rs969159004 Chr.17: 81239208-81239208 on Build GRCh38
rs768305880 Chr.17: 81239251-81239251 on Build GRCh38
rs1004318257 Chr.17: 81239220-81239221 on Build GRCh38
rs1007712101 Chr.17: 81239382-81239382 on Build GRCh38
rs937865541 Chr.17: 81239342-81239343 on Build GRCh38
rs1004497880 Chr.17: 81239376-81239376 on Build GRCh38
rs767893800 Chr.17: 81239435-81239435 on Build GRCh38
rs1024735619 Chr.17: 81239204-81239204 on Build GRCh38
rs934850676 Chr.17: 81239445-81239445 on Build GRCh38
rs908527098 Chr.17: 81239229-81239229 on Build GRCh38
rs901083886 Chr.17: 81239565-81239565 on Build GRCh38
rs560477763 Chr.17: 81239315-81239315 on Build GRCh38
rs913923787 Chr.17: 81239303-81239303 on Build GRCh38
rs778581463 Chr.17: 81239322-81239322 on Build GRCh38
rs778900987 Chr.17: 81239586-81239586 on Build GRCh38
rs937735565 Chr.17: 81239536-81239536 on Build GRCh38
rs201062379 Chr.17: 81239477-81239477 on Build GRCh38
rs535696128 Chr.17: 81239466-81239466 on Build GRCh38
rs562007746 Chr.17: 81239222-81239222 on Build GRCh38
rs1028254775 Chr.17: 81239609-81239609 on Build GRCh38
rs1026841683 Chr.17: 81239220-81239220 on Build GRCh38
rs1004046531 Chr.17: 81239558-81239558 on Build GRCh38
rs938062074 Chr.17: 81239476-81239476 on Build GRCh38
rs753906325 Chr.17: 81239562-81239562 on Build GRCh38
rs879803337 Chr.17: 81239349-81239349 on Build GRCh38
rs117893942 Chr.17: 81239369-81239369 on Build GRCh38
rs879574216 Chr.17: 81239413-81239413 on Build GRCh38
rs1049371460 Chr.17: 81239613-81239613 on Build GRCh38

View Full Product Details