Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82051347-82051850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPS1
Entrez Gene ID
2873
Gene Name
G protein pathway suppressor 1
Gene Aliases
COPS1, CSN1, SGN1
UniGene
Hs.268530
Gene Chromosome Location
Chr.17: 82051477-82057470 on Build GRCh38
Transcripts
XM_017024528.1 XM_017024529.1 XM_017024530.1 NM_002917.1

Gene Symbol
RFNG
Entrez Gene ID
5986
Gene Name
RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
-
UniGene
Hs.569700
Gene Chromosome Location
Chr.17: 82047902-82051774 on Build GRCh38
Transcripts
XM_017024528.1 XM_017024529.1 XM_017024530.1 NM_002917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7415118 c.202C>T p.R68W Chr.17: 82051565-82051565 on Build GRCh38
6556753 c.216G>A p.G72G Chr.17: 82051551-82051551 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs913615428 Chr.17: 82051472-82051472 on Build GRCh38
rs747023686 Chr.17: 82051547-82051547 on Build GRCh38
rs762216232 Chr.17: 82051375-82051375 on Build GRCh38
rs995631609 Chr.17: 82051608-82051608 on Build GRCh38
rs560366083 Chr.17: 82051756-82051756 on Build GRCh38
rs781369314 Chr.17: 82051554-82051554 on Build GRCh38
rs749521395 Chr.17: 82051591-82051591 on Build GRCh38
rs764874861 Chr.17: 82051818-82051818 on Build GRCh38
rs945168430 Chr.17: 82051486-82051486 on Build GRCh38
rs913438528 Chr.17: 82051705-82051705 on Build GRCh38
rs1043777857 Chr.17: 82051509-82051509 on Build GRCh38
rs867394022 Chr.17: 82051400-82051400 on Build GRCh38
rs1028595259 Chr.17: 82051372-82051372 on Build GRCh38
rs899632144 Chr.17: 82051460-82051460 on Build GRCh38
rs762424036 Chr.17: 82051605-82051605 on Build GRCh38
rs529961429 Chr.17: 82051596-82051596 on Build GRCh38
rs868817429 Chr.17: 82051671-82051671 on Build GRCh38
rs767872701 Chr.17: 82051607-82051607 on Build GRCh38
rs189385594 Chr.17: 82051566-82051566 on Build GRCh38
rs979230785 Chr.17: 82051761-82051761 on Build GRCh38
rs925044106 Chr.17: 82051767-82051767 on Build GRCh38
rs117999554 Chr.17: 82051792-82051792 on Build GRCh38
rs778488967 Chr.17: 82051452-82051452 on Build GRCh38
rs949313617 Chr.17: 82051830-82051830 on Build GRCh38
rs753257942 Chr.17: 82051381-82051381 on Build GRCh38
rs1036852301 Chr.17: 82051831-82051831 on Build GRCh38
rs903548286 Chr.17: 82051527-82051527 on Build GRCh38
rs745892405 Chr.17: 82051565-82051565 on Build GRCh38
rs954047714 Chr.17: 82051646-82051646 on Build GRCh38
rs1055199847 Chr.17: 82051783-82051783 on Build GRCh38
rs965605143 Chr.17: 82051688-82051688 on Build GRCh38
rs374008053 Chr.17: 82051371-82051371 on Build GRCh38
rs996545970 Chr.17: 82051655-82051655 on Build GRCh38
rs12943408 Chr.17: 82051520-82051520 on Build GRCh38
rs554985698 Chr.17: 82051444-82051444 on Build GRCh38
rs765965375 Chr.17: 82051376-82051376 on Build GRCh38
rs754690953 Chr.17: 82051383-82051383 on Build GRCh38
rs985878626 Chr.17: 82051664-82051664 on Build GRCh38
rs527415468 Chr.17: 82051760-82051760 on Build GRCh38
rs1019791172 Chr.17: 82051686-82051686 on Build GRCh38
rs541675429 Chr.17: 82051604-82051604 on Build GRCh38
rs954426577 Chr.17: 82051397-82051397 on Build GRCh38
rs912833330 Chr.17: 82051431-82051431 on Build GRCh38
rs538866826 Chr.17: 82051398-82051398 on Build GRCh38
rs757117555 Chr.17: 82051552-82051552 on Build GRCh38
rs777537861 Chr.17: 82051541-82051541 on Build GRCh38
rs774584429 Chr.17: 82051640-82051657 on Build GRCh38
rs758121697 Chr.17: 82051511-82051511 on Build GRCh38
rs985753896 Chr.17: 82051426-82051426 on Build GRCh38
rs896399896 Chr.17: 82051785-82051785 on Build GRCh38
rs937656480 Chr.17: 82051776-82051776 on Build GRCh38
rs988937753 Chr.17: 82051702-82051702 on Build GRCh38
rs752502537 Chr.17: 82051492-82051492 on Build GRCh38
rs1015473605 Chr.17: 82051593-82051593 on Build GRCh38
rs775792172 Chr.17: 82051581-82051581 on Build GRCh38

View Full Product Details