Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82061087-82061587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS1L
Entrez Gene ID
64118
Gene Name
dihydrouridine synthase 1 like
Gene Aliases
DUS1, PP3111
UniGene
Hs.514599
Gene Chromosome Location
Chr.17: 82057872-82065821 on Build GRCh38
Transcripts
NM_022156.4 XM_005256393.2 XM_005256394.2 XM_006722288.2 XM_006722289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4651328 c.745G>A p.V249M Chr.17: 82061306-82061306 on Build GRCh38
4070818 c.819C>T p.H273H Chr.17: 82061232-82061232 on Build GRCh38
6998896 c.770T>C p.L257P Chr.17: 82061281-82061281 on Build GRCh38
5780623 c.842+1G>A p.? Chr.17: 82061208-82061208 on Build GRCh38
986021 c.781C>T p.R261W Chr.17: 82061270-82061270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527980841 Chr.17: 82061188-82061188 on Build GRCh38
rs370244563 Chr.17: 82061404-82061404 on Build GRCh38
rs764752398 Chr.17: 82061277-82061277 on Build GRCh38
rs369444572 Chr.17: 82061262-82061262 on Build GRCh38
rs894796557 Chr.17: 82061516-82061516 on Build GRCh38
rs750909173 Chr.17: 82061274-82061274 on Build GRCh38
rs752303314 Chr.17: 82061291-82061291 on Build GRCh38
rs373424071 Chr.17: 82061312-82061312 on Build GRCh38
rs768936675 Chr.17: 82061318-82061318 on Build GRCh38
rs781397100 Chr.17: 82061251-82061252 on Build GRCh38
rs770645954 Chr.17: 82061567-82061568 on Build GRCh38
rs902137148 Chr.17: 82061298-82061298 on Build GRCh38
rs751458618 Chr.17: 82061305-82061305 on Build GRCh38
rs376342797 Chr.17: 82061292-82061292 on Build GRCh38
rs200310914 Chr.17: 82061190-82061190 on Build GRCh38
rs893631956 Chr.17: 82061199-82061199 on Build GRCh38
rs776784588 Chr.17: 82061162-82061162 on Build GRCh38
rs369359990 Chr.17: 82061306-82061306 on Build GRCh38
rs546450915 Chr.17: 82061482-82061482 on Build GRCh38
rs111961766 Chr.17: 82061304-82061304 on Build GRCh38
rs377127960 Chr.17: 82061388-82061388 on Build GRCh38
rs759683343 Chr.17: 82061163-82061163 on Build GRCh38
rs769022278 Chr.17: 82061256-82061256 on Build GRCh38
rs945152532 Chr.17: 82061464-82061464 on Build GRCh38
rs748332138 Chr.17: 82061523-82061523 on Build GRCh38
rs775756471 Chr.17: 82061172-82061172 on Build GRCh38
rs777649647 Chr.17: 82061213-82061213 on Build GRCh38
rs1003589158 Chr.17: 82061283-82061283 on Build GRCh38
rs777229714 Chr.17: 82061553-82061553 on Build GRCh38
rs529008524 Chr.17: 82061252-82061252 on Build GRCh38
rs377727284 Chr.17: 82061357-82061357 on Build GRCh38
rs1037230683 Chr.17: 82061137-82061137 on Build GRCh38
rs575592889 Chr.17: 82061527-82061527 on Build GRCh38
rs774547341 Chr.17: 82061328-82061328 on Build GRCh38
rs182101098 Chr.17: 82061497-82061497 on Build GRCh38
rs370137276 Chr.17: 82061369-82061369 on Build GRCh38
rs750343134 Chr.17: 82061384-82061384 on Build GRCh38
rs140560459 Chr.17: 82061220-82061220 on Build GRCh38
rs940299533 Chr.17: 82061135-82061135 on Build GRCh38
rs574987254 Chr.17: 82061370-82061370 on Build GRCh38
rs768911603 Chr.17: 82061206-82061207 on Build GRCh38
rs752083808 Chr.17: 82061180-82061180 on Build GRCh38
rs374455590 Chr.17: 82061337-82061337 on Build GRCh38
rs769804615 Chr.17: 82061355-82061356 on Build GRCh38
rs368812276 Chr.17: 82061170-82061170 on Build GRCh38
rs1049839424 Chr.17: 82061154-82061154 on Build GRCh38
rs535891025 Chr.17: 82061405-82061405 on Build GRCh38
rs114225731 Chr.17: 82061429-82061429 on Build GRCh38
rs543017786 Chr.17: 82061562-82061562 on Build GRCh38
rs772398801 Chr.17: 82061334-82061334 on Build GRCh38
rs922616837 Chr.17: 82061142-82061142 on Build GRCh38
rs768075209 Chr.17: 82061270-82061270 on Build GRCh38
rs771525730 Chr.17: 82061532-82061532 on Build GRCh38
rs1029821901 Chr.17: 82061561-82061561 on Build GRCh38
rs779852456 Chr.17: 82061397-82061397 on Build GRCh38
rs778456630 Chr.17: 82061325-82061325 on Build GRCh38
rs773556454 Chr.17: 82061525-82061525 on Build GRCh38
rs901631135 Chr.17: 82061120-82061120 on Build GRCh38
rs201410540 Chr.17: 82061273-82061273 on Build GRCh38
rs1033788892 Chr.17: 82061478-82061478 on Build GRCh38
rs769884511 Chr.17: 82061251-82061251 on Build GRCh38
rs200452679 Chr.17: 82061181-82061181 on Build GRCh38
rs772436014 Chr.17: 82061524-82061524 on Build GRCh38
rs758147265 Chr.17: 82061211-82061211 on Build GRCh38
rs749125637 Chr.17: 82061513-82061514 on Build GRCh38
rs369716083 Chr.17: 82061317-82061317 on Build GRCh38
rs763269004 Chr.17: 82061179-82061179 on Build GRCh38
rs376595282 Chr.17: 82061362-82061362 on Build GRCh38
rs542386854 Chr.17: 82061373-82061373 on Build GRCh38
rs111383728 Chr.17: 82061343-82061343 on Build GRCh38
rs36067039 Chr.17: 82061236-82061237 on Build GRCh38
rs369502194 Chr.17: 82061307-82061307 on Build GRCh38
rs906480049 Chr.17: 82061472-82061472 on Build GRCh38
rs778481501 Chr.17: 82061209-82061209 on Build GRCh38
rs762590315 Chr.17: 82061554-82061554 on Build GRCh38
rs530548330 Chr.17: 82061563-82061563 on Build GRCh38
rs748078918 Chr.17: 82061251-82061251 on Build GRCh38
rs779149636 Chr.17: 82061509-82061509 on Build GRCh38
rs775607556 Chr.17: 82061253-82061253 on Build GRCh38
rs753416829 Chr.17: 82061198-82061198 on Build GRCh38
rs200828252 Chr.17: 82061316-82061316 on Build GRCh38
rs762164701 Chr.17: 82061269-82061269 on Build GRCh38
rs771714568 Chr.17: 82061469-82061469 on Build GRCh38
rs375227321 Chr.17: 82061244-82061244 on Build GRCh38
rs961527233 Chr.17: 82061548-82061548 on Build GRCh38
rs922742020 Chr.17: 82061496-82061496 on Build GRCh38
rs1054517228 Chr.17: 82061412-82061412 on Build GRCh38
rs764653880 Chr.17: 82061345-82061345 on Build GRCh38
rs1003461114 Chr.17: 82061473-82061473 on Build GRCh38
rs1053643088 Chr.17: 82061197-82061197 on Build GRCh38
rs754501147 Chr.17: 82061208-82061208 on Build GRCh38
rs997164997 Chr.17: 82061140-82061140 on Build GRCh38
rs199812921 Chr.17: 82061239-82061239 on Build GRCh38
rs952732821 Chr.17: 82061401-82061401 on Build GRCh38
rs772977184 Chr.17: 82061372-82061384 on Build GRCh38
rs776980846 Chr.17: 82061541-82061541 on Build GRCh38
rs765626186 Chr.17: 82061152-82061152 on Build GRCh38
rs972975745 Chr.17: 82061333-82061333 on Build GRCh38
rs11542331 Chr.17: 82061331-82061331 on Build GRCh38
rs762537831 Chr.17: 82061363-82061363 on Build GRCh38
rs199598057 Chr.17: 82061385-82061385 on Build GRCh38
rs372820618 Chr.17: 82061240-82061240 on Build GRCh38
rs763888194 Chr.17: 82061365-82061365 on Build GRCh38

View Full Product Details