Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 47035187-47035686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNAL2
Entrez Gene ID
83473
Gene Name
katanin catalytic subunit A1 like 2
Gene Aliases
-
UniGene
Hs.404137
Gene Chromosome Location
Chr.18: 46917602-47104795 on Build GRCh38
Transcripts
NM_031303.2 XM_005258357.4 XM_005258358.4 XM_006722554.3 XM_011526219.2 XM_011526220.1 XM_011526221.2 XM_011526223.2 XM_017026028.1 XM_017026029.1 XM_017026030.1 XM_017026032.1 NM_016427.2

Gene Symbol
TCEB3B
Entrez Gene ID
51224
Gene Name
transcription elongation factor B subunit 3B
Gene Aliases
ELOA2, HsT832, TCEB3L
UniGene
Hs.375035
Gene Chromosome Location
Chr.18: 47032572-47035617 on Build GRCh38
Transcripts
NM_031303.2 XM_005258357.4 XM_005258358.4 XM_006722554.3 XM_011526219.2 XM_011526220.1 XM_011526221.2 XM_011526223.2 XM_017026028.1 XM_017026029.1 XM_017026030.1 XM_017026032.1 NM_016427.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
263994 c.72G>A p.P24P Chr.18: 47035193-47035193 on Build GRCh38
5637072 c.52C>A p.L18M Chr.18: 47035213-47035213 on Build GRCh38
1522931 c.26A>G p.H9R Chr.18: 47035239-47035239 on Build GRCh38
6798172 c.49C>T p.R17C Chr.18: 47035216-47035216 on Build GRCh38
6082724 c.11G>A p.G4E Chr.18: 47035254-47035254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752887648 Chr.18: 47035470-47035470 on Build GRCh38
rs749400801 Chr.18: 47035298-47035298 on Build GRCh38
rs1010603730 Chr.18: 47035517-47035517 on Build GRCh38
rs779660004 Chr.18: 47035232-47035232 on Build GRCh38
rs754608268 Chr.18: 47035646-47035646 on Build GRCh38
rs61746775 Chr.18: 47035259-47035259 on Build GRCh38
rs527456029 Chr.18: 47035630-47035630 on Build GRCh38
rs531466151 Chr.18: 47035299-47035299 on Build GRCh38
rs776373401 Chr.18: 47035297-47035297 on Build GRCh38
rs906457196 Chr.18: 47035499-47035499 on Build GRCh38
rs989212632 Chr.18: 47035376-47035376 on Build GRCh38
rs1020487861 Chr.18: 47035473-47035473 on Build GRCh38
rs1010471296 Chr.18: 47035458-47035458 on Build GRCh38
rs1010695441 Chr.18: 47035664-47035664 on Build GRCh38
rs17853508 Chr.18: 47035217-47035217 on Build GRCh38
rs905908146 Chr.18: 47035415-47035415 on Build GRCh38
rs575047068 Chr.18: 47035481-47035481 on Build GRCh38
rs576106599 Chr.18: 47035375-47035375 on Build GRCh38
rs939261979 Chr.18: 47035655-47035655 on Build GRCh38
rs766789590 Chr.18: 47035274-47035274 on Build GRCh38
rs1048475238 Chr.18: 47035409-47035409 on Build GRCh38
rs774788358 Chr.18: 47035220-47035220 on Build GRCh38
rs572647337 Chr.18: 47035245-47035245 on Build GRCh38
rs948052200 Chr.18: 47035491-47035491 on Build GRCh38
rs140604456 Chr.18: 47035407-47035407 on Build GRCh38
rs750727973 Chr.18: 47035269-47035269 on Build GRCh38
rs369463598 Chr.18: 47035244-47035244 on Build GRCh38
rs2576049 Chr.18: 47035659-47035659 on Build GRCh38
rs56098328 Chr.18: 47035588-47035588 on Build GRCh38
rs538795164 Chr.18: 47035380-47035380 on Build GRCh38
rs61738602 Chr.18: 47035248-47035248 on Build GRCh38
rs547290908 Chr.18: 47035658-47035658 on Build GRCh38
rs775699100 Chr.18: 47035258-47035258 on Build GRCh38
rs796201885 Chr.18: 47035448-47035450 on Build GRCh38
rs746768827 Chr.18: 47035296-47035296 on Build GRCh38
rs779716444 Chr.18: 47035566-47035566 on Build GRCh38
rs937444486 Chr.18: 47035644-47035644 on Build GRCh38
rs985497360 Chr.18: 47035352-47035352 on Build GRCh38
rs925022796 Chr.18: 47035327-47035327 on Build GRCh38
rs748203673 Chr.18: 47035450-47035450 on Build GRCh38
rs925004150 Chr.18: 47035216-47035216 on Build GRCh38
rs955112167 Chr.18: 47035555-47035555 on Build GRCh38
rs147815940 Chr.18: 47035598-47035598 on Build GRCh38
rs541268302 Chr.18: 47035428-47035428 on Build GRCh38
rs1030722820 Chr.18: 47035371-47035371 on Build GRCh38
rs533832703 Chr.18: 47035313-47035313 on Build GRCh38
rs192207549 Chr.18: 47035288-47035288 on Build GRCh38
rs200913463 Chr.18: 47035280-47035280 on Build GRCh38
rs1052336516 Chr.18: 47035657-47035657 on Build GRCh38
rs970547642 Chr.18: 47035345-47035345 on Build GRCh38
rs540106663 Chr.18: 47035530-47035530 on Build GRCh38
rs751782164 Chr.18: 47035261-47035261 on Build GRCh38
rs756699078 Chr.18: 47035272-47035272 on Build GRCh38
rs1003490519 Chr.18: 47035503-47035503 on Build GRCh38
rs753353388 Chr.18: 47035278-47035278 on Build GRCh38
rs571463925 Chr.18: 47035303-47035303 on Build GRCh38
rs948051971 Chr.18: 47035637-47035639 on Build GRCh38
rs757957100 Chr.18: 47035294-47035294 on Build GRCh38
rs975500805 Chr.18: 47035540-47035540 on Build GRCh38
rs762298809 Chr.18: 47035262-47035262 on Build GRCh38
rs369159872 Chr.18: 47035268-47035268 on Build GRCh38
rs542628818 Chr.18: 47035483-47035483 on Build GRCh38
rs979300793 Chr.18: 47035486-47035486 on Build GRCh38
rs908067480 Chr.18: 47035448-47035448 on Build GRCh38
rs2571026 Chr.18: 47035424-47035424 on Build GRCh38
rs1037688427 Chr.18: 47035326-47035326 on Build GRCh38
rs748779768 Chr.18: 47035239-47035239 on Build GRCh38
rs766925346 Chr.18: 47035316-47035316 on Build GRCh38
rs985162763 Chr.18: 47035507-47035507 on Build GRCh38
rs754429422 Chr.18: 47035282-47035282 on Build GRCh38
rs72481245 Chr.18: 47035498-47035498 on Build GRCh38
rs906556880 Chr.18: 47035648-47035648 on Build GRCh38
rs545181502 Chr.18: 47035221-47035221 on Build GRCh38
rs777673621 Chr.18: 47035295-47035295 on Build GRCh38
rs933479162 Chr.18: 47035431-47035431 on Build GRCh38
rs953900057 Chr.18: 47035506-47035506 on Build GRCh38
rs1037826202 Chr.18: 47035462-47035462 on Build GRCh38
rs995614452 Chr.18: 47035253-47035253 on Build GRCh38
rs9951823 Chr.18: 47035574-47035574 on Build GRCh38
rs139193632 Chr.18: 47035250-47035250 on Build GRCh38
rs1029216885 Chr.18: 47035550-47035550 on Build GRCh38
rs773691103 Chr.18: 47035305-47035305 on Build GRCh38
rs371830698 Chr.18: 47035489-47035489 on Build GRCh38
rs368096089 Chr.18: 47035547-47035547 on Build GRCh38
rs1010193121 Chr.18: 47035660-47035660 on Build GRCh38
rs912255275 Chr.18: 47035528-47035528 on Build GRCh38
rs963639339 Chr.18: 47035535-47035535 on Build GRCh38
rs567473315 Chr.18: 47035331-47035331 on Build GRCh38
rs577852592 Chr.18: 47035251-47035251 on Build GRCh38
rs1045460839 Chr.18: 47035639-47035639 on Build GRCh38
rs1001901417 Chr.18: 47035279-47035279 on Build GRCh38
rs568483295 Chr.18: 47035354-47035354 on Build GRCh38
rs555215537 Chr.18: 47035447-47035447 on Build GRCh38
rs778562651 Chr.18: 47035284-47035284 on Build GRCh38
rs545163285 Chr.18: 47035584-47035584 on Build GRCh38
rs113947320 Chr.18: 47035373-47035373 on Build GRCh38
rs897024217 Chr.18: 47035403-47035403 on Build GRCh38
rs536214204 Chr.18: 47035333-47035333 on Build GRCh38
rs556364885 Chr.18: 47035572-47035572 on Build GRCh38
rs1005001060 Chr.18: 47035324-47035324 on Build GRCh38
rs200615274 Chr.18: 47035302-47035302 on Build GRCh38
rs571106429 Chr.18: 47035475-47035475 on Build GRCh38
rs914906365 Chr.18: 47035379-47035379 on Build GRCh38
rs565166831 Chr.18: 47035230-47035230 on Build GRCh38
rs987781382 Chr.18: 47035569-47035569 on Build GRCh38
rs927416728 Chr.18: 47035635-47035635 on Build GRCh38
rs547625855 Chr.18: 47035325-47035325 on Build GRCh38

View Full Product Details