Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43273260-43273751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM125
Entrez Gene ID
128218
Gene Name
transmembrane protein 125
Gene Aliases
-
UniGene
Hs.104476
Gene Chromosome Location
Chr.1: 43269921-43274002 on Build GRCh38
Transcripts
NM_001320244.1 NM_144626.2 XM_005270467.3 XM_011540705.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3930785 c.556A>C p.T186P Chr.1: 43273278-43273278 on Build GRCh38
5420354 c.623G>A p.R208H Chr.1: 43273345-43273345 on Build GRCh38
5816091 c.637T>A p.S213T Chr.1: 43273359-43273359 on Build GRCh38
3386174 c.573C>T p.G191G Chr.1: 43273295-43273295 on Build GRCh38
4414460 c.643A>G p.I215V Chr.1: 43273365-43273365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777373513 Chr.1: 43273434-43273434 on Build GRCh38
rs764519336 Chr.1: 43273304-43273304 on Build GRCh38
rs766647593 Chr.1: 43273421-43273421 on Build GRCh38
rs754813085 Chr.1: 43273423-43273423 on Build GRCh38
rs773120304 Chr.1: 43273418-43273418 on Build GRCh38
rs747725163 Chr.1: 43273355-43273355 on Build GRCh38
rs1030797343 Chr.1: 43273645-43273645 on Build GRCh38
rs917387393 Chr.1: 43273478-43273478 on Build GRCh38
rs761203348 Chr.1: 43273296-43273296 on Build GRCh38
rs919292497 Chr.1: 43273564-43273564 on Build GRCh38
rs899558264 Chr.1: 43273689-43273689 on Build GRCh38
rs573916019 Chr.1: 43273316-43273316 on Build GRCh38
rs560295215 Chr.1: 43273579-43273579 on Build GRCh38
rs1035509357 Chr.1: 43273486-43273486 on Build GRCh38
rs992995876 Chr.1: 43273472-43273472 on Build GRCh38
rs563037803 Chr.1: 43273639-43273639 on Build GRCh38
rs374201465 Chr.1: 43273341-43273341 on Build GRCh38
rs779925429 Chr.1: 43273345-43273345 on Build GRCh38
rs907961404 Chr.1: 43273578-43273578 on Build GRCh38
rs899574668 Chr.1: 43273374-43273374 on Build GRCh38
rs377507863 Chr.1: 43273422-43273422 on Build GRCh38
rs181074833 Chr.1: 43273652-43273652 on Build GRCh38
rs72684070 Chr.1: 43273524-43273524 on Build GRCh38
rs766869148 Chr.1: 43273411-43273412 on Build GRCh38
rs758180120 Chr.1: 43273343-43273343 on Build GRCh38
rs991331441 Chr.1: 43273497-43273497 on Build GRCh38
rs977779790 Chr.1: 43273462-43273462 on Build GRCh38
rs1052475394 Chr.1: 43273593-43273593 on Build GRCh38
rs533543975 Chr.1: 43273716-43273716 on Build GRCh38
rs755618185 Chr.1: 43273363-43273363 on Build GRCh38
rs1040190056 Chr.1: 43273589-43273589 on Build GRCh38
rs147126836 Chr.1: 43273383-43273383 on Build GRCh38
rs540418685 Chr.1: 43273401-43273401 on Build GRCh38
rs554054755 Chr.1: 43273651-43273651 on Build GRCh38
rs549721403 Chr.1: 43273625-43273625 on Build GRCh38
rs752658491 Chr.1: 43273432-43273432 on Build GRCh38
rs1022390311 Chr.1: 43273359-43273359 on Build GRCh38
rs942103702 Chr.1: 43273523-43273523 on Build GRCh38
rs777762458 Chr.1: 43273621-43273621 on Build GRCh38
rs777119291 Chr.1: 43273412-43273412 on Build GRCh38
rs1334051 Chr.1: 43273396-43273396 on Build GRCh38
rs900303540 Chr.1: 43273594-43273594 on Build GRCh38
rs775782469 Chr.1: 43273286-43273286 on Build GRCh38
rs1007920131 Chr.1: 43273354-43273354 on Build GRCh38
rs960171275 Chr.1: 43273489-43273489 on Build GRCh38
rs769210590 Chr.1: 43273409-43273409 on Build GRCh38
rs775837602 Chr.1: 43273395-43273395 on Build GRCh38
rs867716644 Chr.1: 43273585-43273585 on Build GRCh38
rs751329830 Chr.1: 43273349-43273349 on Build GRCh38
rs770449916 Chr.1: 43273384-43273384 on Build GRCh38
rs544286792 Chr.1: 43273460-43273460 on Build GRCh38
rs763270627 Chr.1: 43273420-43273420 on Build GRCh38
rs777338185 Chr.1: 43273376-43273376 on Build GRCh38
rs79558033 Chr.1: 43273518-43273518 on Build GRCh38
rs373542770 Chr.1: 43273350-43273350 on Build GRCh38
rs903548422 Chr.1: 43273643-43273643 on Build GRCh38
rs373611887 Chr.1: 43273413-43273413 on Build GRCh38
rs531975028 Chr.1: 43273644-43273644 on Build GRCh38
rs564270746 Chr.1: 43273484-43273484 on Build GRCh38
rs536163439 Chr.1: 43273333-43273333 on Build GRCh38
rs529545215 Chr.1: 43273602-43273602 on Build GRCh38
rs758727630 Chr.1: 43273399-43273401 on Build GRCh38
rs996512344 Chr.1: 43273604-43273604 on Build GRCh38
rs749381358 Chr.1: 43273730-43273730 on Build GRCh38
rs750774636 Chr.1: 43273287-43273295 on Build GRCh38
rs370543654 Chr.1: 43273433-43273433 on Build GRCh38
rs377708145 Chr.1: 43273358-43273358 on Build GRCh38
rs374960970 Chr.1: 43273628-43273628 on Build GRCh38
rs781260562 Chr.1: 43273351-43273351 on Build GRCh38
rs750629942 Chr.1: 43273342-43273342 on Build GRCh38
rs767599393 Chr.1: 43273428-43273428 on Build GRCh38

View Full Product Details