Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5994233-5994773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100128568
Entrez Gene ID
100128568
Gene Name
uncharacterized LOC100128568
Gene Aliases
-
UniGene
Hs.624525
Gene Chromosome Location
Chr.19: 5978403-6020363 on Build GRCh38
Transcripts
NR_046376.1 NM_000635.3 NM_134433.2 XM_011528171.2 XM_011528172.1 XM_017027107.1 XM_017027108.1

Gene Symbol
RFX2
Entrez Gene ID
5990
Gene Name
regulatory factor X2
Gene Aliases
-
UniGene
Hs.465709
Gene Chromosome Location
Chr.19: 5993164-6110653 on Build GRCh38
Transcripts
NR_046376.1 NM_000635.3 NM_134433.2 XM_011528171.2 XM_011528172.1 XM_017027107.1 XM_017027108.1

SNP Detail

SNP ID Chromosome Location
rs1053761146 Chr.19: 5994615-5994615 on Build GRCh38
rs368690545 Chr.19: 5994608-5994608 on Build GRCh38
rs768432905 Chr.19: 5994728-5994728 on Build GRCh38
rs919123547 Chr.19: 5994491-5994491 on Build GRCh38
rs539274638 Chr.19: 5994414-5994414 on Build GRCh38
rs536375503 Chr.19: 5994347-5994347 on Build GRCh38
rs1031807445 Chr.19: 5994366-5994366 on Build GRCh38
rs559292417 Chr.19: 5994428-5994428 on Build GRCh38
rs999040641 Chr.19: 5994353-5994353 on Build GRCh38
rs1036276529 Chr.19: 5994303-5994303 on Build GRCh38
rs943017076 Chr.19: 5994577-5994577 on Build GRCh38
rs1012150573 Chr.19: 5994689-5994689 on Build GRCh38
rs954031018 Chr.19: 5994547-5994547 on Build GRCh38
rs921385662 Chr.19: 5994276-5994276 on Build GRCh38
rs970093866 Chr.19: 5994694-5994694 on Build GRCh38
rs560881374 Chr.19: 5994664-5994698 on Build GRCh38
rs939191255 Chr.19: 5994274-5994274 on Build GRCh38
rs746842122 Chr.19: 5994551-5994551 on Build GRCh38
rs937654640 Chr.19: 5994650-5994650 on Build GRCh38
rs571259040 Chr.19: 5994270-5994270 on Build GRCh38
rs79314920 Chr.19: 5994322-5994322 on Build GRCh38
rs553502759 Chr.19: 5994352-5994352 on Build GRCh38
rs758279273 Chr.19: 5994397-5994397 on Build GRCh38
rs941893440 Chr.19: 5994578-5994578 on Build GRCh38
rs901317374 Chr.19: 5994720-5994720 on Build GRCh38
rs868142727 Chr.19: 5994707-5994707 on Build GRCh38
rs552195624 Chr.19: 5994269-5994269 on Build GRCh38
rs753992646 Chr.19: 5994318-5994318 on Build GRCh38
rs960584194 Chr.19: 5994482-5994482 on Build GRCh38
rs1028759959 Chr.19: 5994538-5994538 on Build GRCh38
rs972379009 Chr.19: 5994483-5994483 on Build GRCh38
rs778790072 Chr.19: 5994346-5994346 on Build GRCh38
rs984177195 Chr.19: 5994573-5994573 on Build GRCh38
rs1049271026 Chr.19: 5994297-5994297 on Build GRCh38
rs142177290 Chr.19: 5994292-5994292 on Build GRCh38
rs999146404 Chr.19: 5994600-5994600 on Build GRCh38
rs970064703 Chr.19: 5994413-5994413 on Build GRCh38
rs1021551630 Chr.19: 5994709-5994709 on Build GRCh38
rs1032062723 Chr.19: 5994412-5994412 on Build GRCh38
rs1036176032 Chr.19: 5994280-5994280 on Build GRCh38
rs961344008 Chr.19: 5994716-5994716 on Build GRCh38
rs1023009018 Chr.19: 5994692-5994692 on Build GRCh38
rs111868938 Chr.19: 5994599-5994599 on Build GRCh38
rs957758628 Chr.19: 5994379-5994379 on Build GRCh38
rs544212307 Chr.19: 5994619-5994619 on Build GRCh38
rs910267677 Chr.19: 5994574-5994574 on Build GRCh38
rs776407216 Chr.19: 5994732-5994732 on Build GRCh38
rs893342463 Chr.19: 5994656-5994656 on Build GRCh38
rs375710691 Chr.19: 5994445-5994445 on Build GRCh38
rs369941620 Chr.19: 5994293-5994293 on Build GRCh38
rs370389256 Chr.19: 5994580-5994580 on Build GRCh38
rs976999674 Chr.19: 5994515-5994515 on Build GRCh38
rs745847952 Chr.19: 5994376-5994376 on Build GRCh38
rs927852218 Chr.19: 5994264-5994264 on Build GRCh38
rs146360636 Chr.19: 5994699-5994699 on Build GRCh38
rs566965901 Chr.19: 5994395-5994395 on Build GRCh38
rs932741242 Chr.19: 5994285-5994285 on Build GRCh38
rs1009734339 Chr.19: 5994478-5994478 on Build GRCh38
rs930471837 Chr.19: 5994525-5994525 on Build GRCh38
rs779942746 Chr.19: 5994398-5994398 on Build GRCh38

View Full Product Details