Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5995502-5996001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100128568
Entrez Gene ID
100128568
Gene Name
uncharacterized LOC100128568
Gene Aliases
-
UniGene
Hs.624525
Gene Chromosome Location
Chr.19: 5978403-6020363 on Build GRCh38
Transcripts
NR_046376.1 NM_000635.3 NM_134433.2 XM_011528171.2 XM_011528172.1 XM_017027107.1 XM_017027108.1

Gene Symbol
RFX2
Entrez Gene ID
5990
Gene Name
regulatory factor X2
Gene Aliases
-
UniGene
Hs.465709
Gene Chromosome Location
Chr.19: 5993164-6110653 on Build GRCh38
Transcripts
NR_046376.1 NM_000635.3 NM_134433.2 XM_011528171.2 XM_011528172.1 XM_017027107.1 XM_017027108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1613016 c.2026G>T p.A676S Chr.19: 5995631-5995631 on Build GRCh38
6430873 c.2053A>G p.K685E Chr.19: 5995604-5995604 on Build GRCh38
4475994 c.2032C>T p.L678L Chr.19: 5995625-5995625 on Build GRCh38
4438286 c.2042C>T p.T681M Chr.19: 5995615-5995615 on Build GRCh38
4654615 c.2019C>T p.N673N Chr.19: 5995638-5995638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918837720 Chr.19: 5995729-5995729 on Build GRCh38
rs753805552 Chr.19: 5995566-5995566 on Build GRCh38
rs201301974 Chr.19: 5995587-5995587 on Build GRCh38
rs548837988 Chr.19: 5995956-5995956 on Build GRCh38
rs568779361 Chr.19: 5995542-5995542 on Build GRCh38
rs370511208 Chr.19: 5995620-5995620 on Build GRCh38
rs377058340 Chr.19: 5995579-5995579 on Build GRCh38
rs35225284 Chr.19: 5995614-5995614 on Build GRCh38
rs942791702 Chr.19: 5995805-5995805 on Build GRCh38
rs922637056 Chr.19: 5995841-5995841 on Build GRCh38
rs993524663 Chr.19: 5995716-5995716 on Build GRCh38
rs1051684540 Chr.19: 5995610-5995610 on Build GRCh38
rs113671883 Chr.19: 5995952-5995952 on Build GRCh38
rs532529716 Chr.19: 5995564-5995564 on Build GRCh38
rs1011640148 Chr.19: 5995742-5995742 on Build GRCh38
rs904351212 Chr.19: 5995964-5995964 on Build GRCh38
rs1039793091 Chr.19: 5995810-5995810 on Build GRCh38
rs1018010273 Chr.19: 5995930-5995930 on Build GRCh38
rs893207619 Chr.19: 5995740-5995740 on Build GRCh38
rs937331347 Chr.19: 5995717-5995717 on Build GRCh38
rs560558106 Chr.19: 5995739-5995739 on Build GRCh38
rs57452891 Chr.19: 5995573-5995573 on Build GRCh38
rs757471649 Chr.19: 5995568-5995568 on Build GRCh38
rs909986044 Chr.19: 5995760-5995760 on Build GRCh38
rs371356318 Chr.19: 5995745-5995745 on Build GRCh38
rs149764443 Chr.19: 5995586-5995586 on Build GRCh38
rs1052126063 Chr.19: 5995901-5995901 on Build GRCh38
rs200826089 Chr.19: 5995592-5995592 on Build GRCh38
rs565959888 Chr.19: 5995628-5995628 on Build GRCh38
rs749024184 Chr.19: 5995786-5995786 on Build GRCh38
rs760770934 Chr.19: 5995559-5995559 on Build GRCh38
rs531936171 Chr.19: 5995920-5995920 on Build GRCh38
rs2302222 Chr.19: 5995571-5995571 on Build GRCh38
rs373222248 Chr.19: 5995694-5995695 on Build GRCh38
rs780136159 Chr.19: 5995569-5995569 on Build GRCh38
rs575841631 Chr.19: 5995607-5995607 on Build GRCh38
rs955108272 Chr.19: 5995883-5995883 on Build GRCh38
rs1010505943 Chr.19: 5995929-5995929 on Build GRCh38
rs765359795 Chr.19: 5995684-5995684 on Build GRCh38
rs540243563 Chr.19: 5995707-5995707 on Build GRCh38
rs373007421 Chr.19: 5995637-5995637 on Build GRCh38
rs772669666 Chr.19: 5995615-5995615 on Build GRCh38
rs546331340 Chr.19: 5995798-5995798 on Build GRCh38
rs891657409 Chr.19: 5995611-5995611 on Build GRCh38
rs999458503 Chr.19: 5995837-5995837 on Build GRCh38
rs997459706 Chr.19: 5995605-5995605 on Build GRCh38
rs1001171747 Chr.19: 5995699-5995699 on Build GRCh38
rs572339015 Chr.19: 5995670-5995670 on Build GRCh38
rs373952212 Chr.19: 5995576-5995576 on Build GRCh38
rs754181250 Chr.19: 5995669-5995669 on Build GRCh38
rs973307746 Chr.19: 5995539-5995539 on Build GRCh38
rs753131728 Chr.19: 5995555-5995555 on Build GRCh38
rs1034003618 Chr.19: 5995706-5995706 on Build GRCh38
rs781626584 Chr.19: 5995606-5995607 on Build GRCh38
rs563027883 Chr.19: 5995882-5995882 on Build GRCh38
rs1029987953 Chr.19: 5995843-5995843 on Build GRCh38
rs372282415 Chr.19: 5995565-5995565 on Build GRCh38
rs746939776 Chr.19: 5995808-5995808 on Build GRCh38
rs561826046 Chr.19: 5995631-5995631 on Build GRCh38
rs377349108 Chr.19: 5995697-5995697 on Build GRCh38
rs750504420 Chr.19: 5995665-5995666 on Build GRCh38

View Full Product Details