Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6495787-6496291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBB4A
Entrez Gene ID
10382
Gene Name
tubulin beta 4A class IVa
Gene Aliases
DYT4, TUBB4, beta-5
UniGene
Hs.110837
Gene Chromosome Location
Chr.19: 6494319-6502848 on Build GRCh38
Transcripts
NM_001289123.1 NM_001289127.1 NM_001289129.1 NM_001289130.1 NM_001289131.1 NM_006087.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5087842 c.484C>T p.R162C Chr.19: 6496015-6496015 on Build GRCh38
7301542 c.607G>A p.D203N Chr.19: 6495892-6495892 on Build GRCh38
6086192 c.279C>T p.G93G Chr.19: 6496220-6496220 on Build GRCh38
7213336 c.292G>A p.G98S Chr.19: 6496207-6496207 on Build GRCh38
2153371 c.624C>T p.Y208Y Chr.19: 6495875-6495875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199666595 Chr.19: 6495875-6495875 on Build GRCh38
rs765848572 Chr.19: 6495953-6495953 on Build GRCh38
rs560881201 Chr.19: 6496084-6496084 on Build GRCh38
rs587777468 Chr.19: 6495966-6495966 on Build GRCh38
rs371265470 Chr.19: 6496126-6496126 on Build GRCh38
rs760545489 Chr.19: 6495911-6495911 on Build GRCh38
rs745995021 Chr.19: 6496129-6496129 on Build GRCh38
rs781137307 Chr.19: 6496133-6496133 on Build GRCh38
rs753210565 Chr.19: 6496097-6496097 on Build GRCh38
rs797046078 Chr.19: 6495832-6495832 on Build GRCh38
rs878861264 Chr.19: 6496242-6496242 on Build GRCh38
rs772746260 Chr.19: 6496067-6496067 on Build GRCh38
rs1053263 Chr.19: 6495947-6495947 on Build GRCh38
rs772090741 Chr.19: 6496130-6496130 on Build GRCh38
rs769979649 Chr.19: 6495863-6495863 on Build GRCh38
rs780051412 Chr.19: 6496262-6496262 on Build GRCh38
rs747046930 Chr.19: 6496268-6496268 on Build GRCh38
rs770998209 Chr.19: 6495887-6495887 on Build GRCh38
rs778138515 Chr.19: 6496117-6496117 on Build GRCh38
rs778943389 Chr.19: 6496230-6496231 on Build GRCh38
rs748051542 Chr.19: 6496145-6496145 on Build GRCh38
rs776475401 Chr.19: 6495917-6495917 on Build GRCh38
rs777883701 Chr.19: 6496010-6496010 on Build GRCh38
rs375537381 Chr.19: 6496056-6496056 on Build GRCh38
rs755398669 Chr.19: 6495842-6495842 on Build GRCh38
rs576008094 Chr.19: 6496031-6496031 on Build GRCh38
rs536369814 Chr.19: 6495818-6495818 on Build GRCh38
rs748510977 Chr.19: 6495862-6495862 on Build GRCh38
rs779020457 Chr.19: 6496019-6496019 on Build GRCh38
rs146906606 Chr.19: 6495833-6495833 on Build GRCh38
rs764502394 Chr.19: 6496236-6496236 on Build GRCh38
rs61741669 Chr.19: 6496157-6496157 on Build GRCh38
rs767100230 Chr.19: 6496214-6496214 on Build GRCh38
rs770924510 Chr.19: 6496016-6496016 on Build GRCh38
rs752127268 Chr.19: 6495965-6495965 on Build GRCh38
rs781643888 Chr.19: 6495860-6495860 on Build GRCh38
rs768190406 Chr.19: 6496095-6496095 on Build GRCh38
rs759144466 Chr.19: 6496178-6496178 on Build GRCh38
rs759474575 Chr.19: 6496248-6496248 on Build GRCh38
rs766118356 Chr.19: 6496079-6496079 on Build GRCh38
rs774127285 Chr.19: 6496172-6496172 on Build GRCh38
rs768924966 Chr.19: 6496227-6496236 on Build GRCh38
rs758477616 Chr.19: 6496254-6496254 on Build GRCh38
rs141399672 Chr.19: 6495983-6495983 on Build GRCh38
rs200757174 Chr.19: 6496064-6496064 on Build GRCh38
rs748388564 Chr.19: 6496258-6496259 on Build GRCh38
rs757594393 Chr.19: 6496241-6496241 on Build GRCh38
rs761635539 Chr.19: 6495931-6495931 on Build GRCh38
rs139391205 Chr.19: 6495914-6495914 on Build GRCh38
rs769489513 Chr.19: 6496148-6496148 on Build GRCh38
rs775406352 Chr.19: 6495893-6495893 on Build GRCh38
rs1053262 Chr.19: 6496034-6496034 on Build GRCh38
rs886041008 Chr.19: 6495955-6495955 on Build GRCh38
rs371280184 Chr.19: 6495989-6495989 on Build GRCh38
rs754136059 Chr.19: 6496118-6496118 on Build GRCh38
rs374559176 Chr.19: 6496070-6496070 on Build GRCh38
rs761298089 Chr.19: 6496227-6496227 on Build GRCh38
rs773118700 Chr.19: 6496150-6496150 on Build GRCh38
rs143415955 Chr.19: 6495995-6495995 on Build GRCh38
rs1053260 Chr.19: 6496073-6496073 on Build GRCh38
rs902910254 Chr.19: 6496240-6496240 on Build GRCh38
rs147013122 Chr.19: 6496103-6496103 on Build GRCh38
rs749328256 Chr.19: 6495820-6495820 on Build GRCh38
rs750042422 Chr.19: 6495941-6495941 on Build GRCh38
rs796738621 Chr.19: 6496063-6496063 on Build GRCh38
rs1057517986 Chr.19: 6495964-6495964 on Build GRCh38
rs374382127 Chr.19: 6496246-6496246 on Build GRCh38
rs770749775 Chr.19: 6496169-6496169 on Build GRCh38
rs751182929 Chr.19: 6496081-6496081 on Build GRCh38
rs749457019 Chr.19: 6496014-6496014 on Build GRCh38
rs587777429 Chr.19: 6496032-6496032 on Build GRCh38
rs749381811 Chr.19: 6495878-6495878 on Build GRCh38
rs761535706 Chr.19: 6496052-6496052 on Build GRCh38
rs775871196 Chr.19: 6496222-6496222 on Build GRCh38
rs941410459 Chr.19: 6495959-6495959 on Build GRCh38
rs780447157 Chr.19: 6495831-6495831 on Build GRCh38
rs764847265 Chr.19: 6495934-6495934 on Build GRCh38

View Full Product Details