Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12349248-12349782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF442
Entrez Gene ID
79973
Gene Name
zinc finger protein 442
Gene Aliases
-
UniGene
Hs.253193
Gene Chromosome Location
Chr.19: 12349371-12372636 on Build GRCh38
Transcripts
NM_030824.2 XM_006722908.3 XM_017027316.1 XM_017027317.1 XM_017027318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6830269 c.1819A>G p.K607E Chr.19: 12349766-12349766 on Build GRCh38
991245 c.1859G>T p.R620M Chr.19: 12349726-12349726 on Build GRCh38
3362661 c.1831T>C p.S611P Chr.19: 12349754-12349754 on Build GRCh38
6830265 c.1825C>T p.L609L Chr.19: 12349760-12349760 on Build GRCh38
1234065 c.1871G>T p.R624I Chr.19: 12349714-12349714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372886629 Chr.19: 12349670-12349670 on Build GRCh38
rs112730723 Chr.19: 12349635-12349635 on Build GRCh38
rs753590052 Chr.19: 12349649-12349649 on Build GRCh38
rs373877693 Chr.19: 12349748-12349748 on Build GRCh38
rs1006805101 Chr.19: 12349515-12349515 on Build GRCh38
rs567251325 Chr.19: 12349291-12349291 on Build GRCh38
rs936524895 Chr.19: 12349312-12349312 on Build GRCh38
rs901617122 Chr.19: 12349725-12349725 on Build GRCh38
rs529105354 Chr.19: 12349721-12349721 on Build GRCh38
rs538355863 Chr.19: 12349561-12349561 on Build GRCh38
rs891445271 Chr.19: 12349465-12349465 on Build GRCh38
rs1029362772 Chr.19: 12349275-12349275 on Build GRCh38
rs575331444 Chr.19: 12349671-12349671 on Build GRCh38
rs973196916 Chr.19: 12349370-12349370 on Build GRCh38
rs185313320 Chr.19: 12349729-12349729 on Build GRCh38
rs761863288 Chr.19: 12349657-12349657 on Build GRCh38
rs951955626 Chr.19: 12349280-12349280 on Build GRCh38
rs868577874 Chr.19: 12349419-12349419 on Build GRCh38
rs760344434 Chr.19: 12349395-12349395 on Build GRCh38
rs992452064 Chr.19: 12349564-12349565 on Build GRCh38
rs901590565 Chr.19: 12349488-12349488 on Build GRCh38
rs984646701 Chr.19: 12349540-12349540 on Build GRCh38
rs181911412 Chr.19: 12349409-12349409 on Build GRCh38
rs564180828 Chr.19: 12349751-12349751 on Build GRCh38
rs562921627 Chr.19: 12349660-12349660 on Build GRCh38
rs190238577 Chr.19: 12349496-12349496 on Build GRCh38
rs375632226 Chr.19: 12349513-12349513 on Build GRCh38
rs917434757 Chr.19: 12349373-12349373 on Build GRCh38
rs1016901623 Chr.19: 12349566-12349566 on Build GRCh38
rs369005922 Chr.19: 12349659-12349659 on Build GRCh38
rs779984622 Chr.19: 12349653-12349653 on Build GRCh38
rs1010002801 Chr.19: 12349473-12349473 on Build GRCh38
rs1044568238 Chr.19: 12349389-12349389 on Build GRCh38
rs549604469 Chr.19: 12349454-12349454 on Build GRCh38
rs950157136 Chr.19: 12349623-12349623 on Build GRCh38
rs773079638 Chr.19: 12349363-12349363 on Build GRCh38
rs1041346556 Chr.19: 12349420-12349420 on Build GRCh38
rs1019070527 Chr.19: 12349480-12349480 on Build GRCh38
rs553519776 Chr.19: 12349584-12349584 on Build GRCh38
rs917362108 Chr.19: 12349614-12349614 on Build GRCh38
rs777041514 Chr.19: 12349756-12349756 on Build GRCh38
rs961327705 Chr.19: 12349582-12349582 on Build GRCh38
rs141143740 Chr.19: 12349703-12349703 on Build GRCh38
rs536123721 Chr.19: 12349647-12349647 on Build GRCh38
rs889719364 Chr.19: 12349323-12349324 on Build GRCh38
rs950226203 Chr.19: 12349382-12349382 on Build GRCh38
rs993455541 Chr.19: 12349486-12349486 on Build GRCh38
rs764423205 Chr.19: 12349750-12349750 on Build GRCh38
rs769627389 Chr.19: 12349704-12349704 on Build GRCh38
rs748760869 Chr.19: 12349713-12349713 on Build GRCh38
rs1050702990 Chr.19: 12349753-12349753 on Build GRCh38
rs770358035 Chr.19: 12349723-12349723 on Build GRCh38
rs1025212736 Chr.19: 12349570-12349570 on Build GRCh38
rs571923123 Chr.19: 12349640-12349640 on Build GRCh38
rs192090355 Chr.19: 12349315-12349315 on Build GRCh38
rs772844894 Chr.19: 12349708-12349708 on Build GRCh38
rs147498914 Chr.19: 12349535-12349535 on Build GRCh38
rs781174917 Chr.19: 12349697-12349697 on Build GRCh38
rs549733238 Chr.19: 12349283-12349283 on Build GRCh38
rs372370745 Chr.19: 12349601-12349601 on Build GRCh38
rs377156743 Chr.19: 12349732-12349732 on Build GRCh38
rs372912367 Chr.19: 12349441-12349441 on Build GRCh38

View Full Product Details