Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43949338-43949844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO13
Entrez Gene ID
9670
Gene Name
importin 13
Gene Aliases
IMP13, KAP13, LGL2, RANBP13
UniGene
Hs.158497
Gene Chromosome Location
Chr.1: 43946806-43968022 on Build GRCh38
Transcripts
NM_014652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
386051 c.205G>T p.D69Y Chr.1: 43949537-43949537 on Build GRCh38
7080150 c.318C>T p.F106F Chr.1: 43949650-43949650 on Build GRCh38
3865633 c.464C>T p.S155L Chr.1: 43949796-43949796 on Build GRCh38
249717 c.85-2A>T p.? Chr.1: 43949415-43949415 on Build GRCh38
5458457 c.391G>A p.A131T Chr.1: 43949723-43949723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562549124 Chr.1: 43949500-43949500 on Build GRCh38
rs763924857 Chr.1: 43949433-43949433 on Build GRCh38
rs551718604 Chr.1: 43949620-43949620 on Build GRCh38
rs761904095 Chr.1: 43949698-43949698 on Build GRCh38
rs199832608 Chr.1: 43949512-43949512 on Build GRCh38
rs200719466 Chr.1: 43949662-43949662 on Build GRCh38
rs757981340 Chr.1: 43949450-43949450 on Build GRCh38
rs746066696 Chr.1: 43949663-43949663 on Build GRCh38
rs765959818 Chr.1: 43949599-43949599 on Build GRCh38
rs769960153 Chr.1: 43949750-43949750 on Build GRCh38
rs200661833 Chr.1: 43949419-43949419 on Build GRCh38
rs114449275 Chr.1: 43949797-43949797 on Build GRCh38
rs368205975 Chr.1: 43949386-43949386 on Build GRCh38
rs754287104 Chr.1: 43949736-43949736 on Build GRCh38
rs931410472 Chr.1: 43949513-43949513 on Build GRCh38
rs758900435 Chr.1: 43949725-43949725 on Build GRCh38
rs2240447 Chr.1: 43949743-43949743 on Build GRCh38
rs189918025 Chr.1: 43949411-43949411 on Build GRCh38
rs776645448 Chr.1: 43949562-43949562 on Build GRCh38
rs41270385 Chr.1: 43949752-43949752 on Build GRCh38
rs775594295 Chr.1: 43949537-43949537 on Build GRCh38
rs150923394 Chr.1: 43949518-43949518 on Build GRCh38
rs769818305 Chr.1: 43949816-43949816 on Build GRCh38
rs1048914689 Chr.1: 43949598-43949598 on Build GRCh38
rs764926493 Chr.1: 43949438-43949438 on Build GRCh38
rs368161909 Chr.1: 43949436-43949436 on Build GRCh38
rs968187540 Chr.1: 43949388-43949388 on Build GRCh38
rs372324695 Chr.1: 43949661-43949661 on Build GRCh38
rs767588550 Chr.1: 43949413-43949413 on Build GRCh38
rs756681393 Chr.1: 43949591-43949591 on Build GRCh38
rs1004633348 Chr.1: 43949621-43949621 on Build GRCh38
rs750090898 Chr.1: 43949592-43949592 on Build GRCh38
rs768787553 Chr.1: 43949675-43949675 on Build GRCh38
rs150073613 Chr.1: 43949616-43949617 on Build GRCh38
rs760627652 Chr.1: 43949544-43949544 on Build GRCh38
rs555884453 Chr.1: 43949753-43949753 on Build GRCh38
rs34319792 Chr.1: 43949758-43949758 on Build GRCh38
rs979782410 Chr.1: 43949546-43949546 on Build GRCh38
rs753207880 Chr.1: 43949582-43949582 on Build GRCh38
rs768606187 Chr.1: 43949775-43949775 on Build GRCh38
rs201152991 Chr.1: 43949711-43949711 on Build GRCh38
rs762874919 Chr.1: 43949707-43949707 on Build GRCh38
rs759550114 Chr.1: 43949382-43949382 on Build GRCh38
rs988920669 Chr.1: 43949468-43949468 on Build GRCh38
rs754370368 Chr.1: 43949635-43949635 on Build GRCh38
rs769692960 Chr.1: 43949699-43949699 on Build GRCh38
rs774575745 Chr.1: 43949380-43949380 on Build GRCh38
rs779771871 Chr.1: 43949469-43949469 on Build GRCh38
rs888903190 Chr.1: 43949714-43949714 on Build GRCh38
rs377163183 Chr.1: 43949719-43949719 on Build GRCh38
rs779307238 Chr.1: 43949658-43949658 on Build GRCh38
rs750105127 Chr.1: 43949448-43949448 on Build GRCh38
rs750995906 Chr.1: 43949616-43949616 on Build GRCh38
rs772125439 Chr.1: 43949536-43949536 on Build GRCh38
rs771295587 Chr.1: 43949506-43949506 on Build GRCh38
rs527262092 Chr.1: 43949694-43949694 on Build GRCh38
rs778373516 Chr.1: 43949493-43949493 on Build GRCh38
rs35709314 Chr.1: 43949704-43949704 on Build GRCh38
rs763890912 Chr.1: 43949551-43949551 on Build GRCh38
rs772319600 Chr.1: 43949664-43949664 on Build GRCh38
rs1028641022 Chr.1: 43949705-43949705 on Build GRCh38
rs981217383 Chr.1: 43949799-43949799 on Build GRCh38
rs371807246 Chr.1: 43949761-43949761 on Build GRCh38
rs762747654 Chr.1: 43949594-43949594 on Build GRCh38
rs368253513 Chr.1: 43949581-43949581 on Build GRCh38
rs956956761 Chr.1: 43949455-43949455 on Build GRCh38
rs138977771 Chr.1: 43949726-43949726 on Build GRCh38
rs781453528 Chr.1: 43949418-43949418 on Build GRCh38
rs773307976 Chr.1: 43949817-43949817 on Build GRCh38
rs761449688 Chr.1: 43949573-43949573 on Build GRCh38
rs147398347 Chr.1: 43949774-43949774 on Build GRCh38
rs1029170506 Chr.1: 43949385-43949385 on Build GRCh38
rs757612978 Chr.1: 43949657-43949657 on Build GRCh38
rs748670508 Chr.1: 43949477-43949477 on Build GRCh38
rs757809156 Chr.1: 43949738-43949738 on Build GRCh38
rs1036255264 Chr.1: 43949627-43949627 on Build GRCh38
rs756621643 Chr.1: 43949491-43949491 on Build GRCh38
rs571227578 Chr.1: 43949668-43949668 on Build GRCh38
rs748071342 Chr.1: 43949803-43949803 on Build GRCh38

View Full Product Details