Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17751388-17751885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
UniGene
Hs.96485
Gene Chromosome Location
Chr.19: 17747718-17788568 on Build GRCh38
Transcripts
NM_001161357.1 NM_001161358.1 NM_001161359.1 NM_015122.2 XM_011527824.2 XM_011527825.2 XM_011527826.2 XM_011527828.2 XM_017026519.1 XM_017026520.1 XM_017026522.1 XM_017026523.1 XM_017026525.1 XM_017026526.1 XM_017026528.1

SNP Detail

SNP ID Chromosome Location
rs571520837 Chr.19: 17751465-17751465 on Build GRCh38
rs919952697 Chr.19: 17751647-17751647 on Build GRCh38
rs981241681 Chr.19: 17751642-17751643 on Build GRCh38
rs780317293 Chr.19: 17751780-17751780 on Build GRCh38
rs956985301 Chr.19: 17751778-17751779 on Build GRCh38
rs532657784 Chr.19: 17751844-17751844 on Build GRCh38
rs924032126 Chr.19: 17751749-17751749 on Build GRCh38
rs555880961 Chr.19: 17751628-17751628 on Build GRCh38
rs1050567409 Chr.19: 17751745-17751745 on Build GRCh38
rs923458177 Chr.19: 17751443-17751443 on Build GRCh38
rs561300269 Chr.19: 17751742-17751742 on Build GRCh38
rs964833151 Chr.19: 17751423-17751423 on Build GRCh38
rs942001183 Chr.19: 17751502-17751502 on Build GRCh38
rs893052142 Chr.19: 17751605-17751605 on Build GRCh38
rs371983934 Chr.19: 17751437-17751437 on Build GRCh38
rs963979390 Chr.19: 17751716-17751716 on Build GRCh38
rs992376879 Chr.19: 17751619-17751619 on Build GRCh38
rs768222116 Chr.19: 17751692-17751692 on Build GRCh38
rs575740975 Chr.19: 17751683-17751683 on Build GRCh38
rs552615306 Chr.19: 17751406-17751406 on Build GRCh38
rs541741293 Chr.19: 17751728-17751728 on Build GRCh38
rs1055542087 Chr.19: 17751515-17751515 on Build GRCh38
rs111855347 Chr.19: 17751706-17751706 on Build GRCh38
rs35089198 Chr.19: 17751459-17751459 on Build GRCh38
rs1030620460 Chr.19: 17751778-17751778 on Build GRCh38
rs907072122 Chr.19: 17751540-17751540 on Build GRCh38
rs112730218 Chr.19: 17751571-17751571 on Build GRCh38
rs563733641 Chr.19: 17751819-17751819 on Build GRCh38
rs983621631 Chr.19: 17751708-17751708 on Build GRCh38
rs750547185 Chr.19: 17751620-17751620 on Build GRCh38
rs1052930330 Chr.19: 17751564-17751564 on Build GRCh38
rs976224942 Chr.19: 17751442-17751442 on Build GRCh38
rs909431694 Chr.19: 17751712-17751712 on Build GRCh38
rs1011214535 Chr.19: 17751609-17751609 on Build GRCh38
rs375846928 Chr.19: 17751818-17751818 on Build GRCh38
rs929532014 Chr.19: 17751448-17751448 on Build GRCh38
rs952771372 Chr.19: 17751854-17751854 on Build GRCh38
rs188742362 Chr.19: 17751531-17751531 on Build GRCh38
rs1001083605 Chr.19: 17751610-17751610 on Build GRCh38
rs980950631 Chr.19: 17751834-17751834 on Build GRCh38
rs375765788 Chr.19: 17751484-17751484 on Build GRCh38
rs1011932509 Chr.19: 17751820-17751820 on Build GRCh38
rs1017322552 Chr.19: 17751422-17751422 on Build GRCh38
rs1017709671 Chr.19: 17751648-17751648 on Build GRCh38
rs749348215 Chr.19: 17751799-17751799 on Build GRCh38
rs185825867 Chr.19: 17751498-17751498 on Build GRCh38
rs567488795 Chr.19: 17751545-17751545 on Build GRCh38
rs916590572 Chr.19: 17751737-17751737 on Build GRCh38
rs544557189 Chr.19: 17751698-17751698 on Build GRCh38
rs983739806 Chr.19: 17751464-17751464 on Build GRCh38
rs192290646 Chr.19: 17751791-17751791 on Build GRCh38
rs1000510035 Chr.19: 17751754-17751754 on Build GRCh38
rs956933230 Chr.19: 17751616-17751616 on Build GRCh38
rs934463046 Chr.19: 17751775-17751775 on Build GRCh38
rs1014219562 Chr.19: 17751786-17751786 on Build GRCh38
rs964475555 Chr.19: 17751657-17751657 on Build GRCh38
rs997295842 Chr.19: 17751658-17751658 on Build GRCh38
rs950899290 Chr.19: 17751691-17751691 on Build GRCh38
rs1040216491 Chr.19: 17751740-17751740 on Build GRCh38
rs184173366 Chr.19: 17751817-17751817 on Build GRCh38
rs996296723 Chr.19: 17751514-17751514 on Build GRCh38
rs535961762 Chr.19: 17751608-17751608 on Build GRCh38
rs1045521646 Chr.19: 17751539-17751539 on Build GRCh38
rs1003857609 Chr.19: 17751447-17751447 on Build GRCh38
rs368485163 Chr.19: 17751743-17751743 on Build GRCh38
rs1048978841 Chr.19: 17751461-17751461 on Build GRCh38
rs757520274 Chr.19: 17751530-17751530 on Build GRCh38
rs796457553 Chr.19: 17751467-17751467 on Build GRCh38
rs111431923 Chr.19: 17751727-17751727 on Build GRCh38
rs1031241431 Chr.19: 17751615-17751615 on Build GRCh38
rs779087848 Chr.19: 17751713-17751714 on Build GRCh38
rs559487212 Chr.19: 17751561-17751561 on Build GRCh38
rs558150290 Chr.19: 17751715-17751715 on Build GRCh38

View Full Product Details