Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17770451-17770973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
UniGene
Hs.96485
Gene Chromosome Location
Chr.19: 17747718-17788568 on Build GRCh38
Transcripts
NM_001161357.1 NM_001161358.1 NM_001161359.1 NM_015122.2 XM_005259831.3 XM_006722701.3 XM_006722702.3 XM_011527824.2 XM_011527825.2 XM_011527826.2 XM_011527827.2 XM_011527828.2 XM_017026519.1 XM_017026520.1 XM_017026521.1 XM_017026522.1 XM_017026523.1 XM_017026524.1 XM_017026525.1 XM_017026526.1 XM_017026527.1 XM_017026528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
50980 c.588A>C p.S196S Chr.19: 17770890-17770890 on Build GRCh38
5056374 c.388G>A p.V130I Chr.19: 17770476-17770476 on Build GRCh38
7129193 c.455G>A p.R152Q Chr.19: 17770543-17770543 on Build GRCh38
3530557 c.410C>T p.S137F Chr.19: 17770498-17770498 on Build GRCh38
6524760 c.560C>A p.A187D Chr.19: 17770862-17770862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780905707 Chr.19: 17770548-17770548 on Build GRCh38
rs984495146 Chr.19: 17770632-17770632 on Build GRCh38
rs945660890 Chr.19: 17770732-17770732 on Build GRCh38
rs760562128 Chr.19: 17770853-17770853 on Build GRCh38
rs147599881 Chr.19: 17770859-17770859 on Build GRCh38
rs914852424 Chr.19: 17770592-17770592 on Build GRCh38
rs774056510 Chr.19: 17770872-17770872 on Build GRCh38
rs371544052 Chr.19: 17770600-17770600 on Build GRCh38
rs969759686 Chr.19: 17770705-17770705 on Build GRCh38
rs774772787 Chr.19: 17770755-17770755 on Build GRCh38
rs774364972 Chr.19: 17770756-17770756 on Build GRCh38
rs968870009 Chr.19: 17770913-17770913 on Build GRCh38
rs766676105 Chr.19: 17770610-17770610 on Build GRCh38
rs747932265 Chr.19: 17770530-17770530 on Build GRCh38
rs757233894 Chr.19: 17770518-17770518 on Build GRCh38
rs62121735 Chr.19: 17770937-17770937 on Build GRCh38
rs753362747 Chr.19: 17770934-17770934 on Build GRCh38
rs116103463 Chr.19: 17770597-17770597 on Build GRCh38
rs775884443 Chr.19: 17770582-17770582 on Build GRCh38
rs751689463 Chr.19: 17770508-17770508 on Build GRCh38
rs201514455 Chr.19: 17770621-17770621 on Build GRCh38
rs933354677 Chr.19: 17770790-17770790 on Build GRCh38
rs745587581 Chr.19: 17770550-17770550 on Build GRCh38
rs767242816 Chr.19: 17770629-17770634 on Build GRCh38
rs767557063 Chr.19: 17770595-17770595 on Build GRCh38
rs567199358 Chr.19: 17770593-17770593 on Build GRCh38
rs1003629486 Chr.19: 17770759-17770759 on Build GRCh38
rs765931639 Chr.19: 17770917-17770917 on Build GRCh38
rs956388123 Chr.19: 17770735-17770735 on Build GRCh38
rs3746197 Chr.19: 17770743-17770743 on Build GRCh38
rs561104956 Chr.19: 17770673-17770673 on Build GRCh38
rs535170386 Chr.19: 17770677-17770677 on Build GRCh38
rs759911593 Chr.19: 17770475-17770475 on Build GRCh38
rs369971263 Chr.19: 17770788-17770788 on Build GRCh38
rs777535869 Chr.19: 17770537-17770537 on Build GRCh38
rs750221294 Chr.19: 17770901-17770901 on Build GRCh38
rs769423107 Chr.19: 17770571-17770571 on Build GRCh38
rs1033725270 Chr.19: 17770618-17770618 on Build GRCh38
rs754641757 Chr.19: 17770815-17770815 on Build GRCh38
rs556530863 Chr.19: 17770620-17770620 on Build GRCh38
rs539199066 Chr.19: 17770651-17770651 on Build GRCh38
rs750292653 Chr.19: 17770892-17770892 on Build GRCh38
rs543802752 Chr.19: 17770775-17770775 on Build GRCh38
rs368907319 Chr.19: 17770591-17770591 on Build GRCh38
rs749006465 Chr.19: 17770814-17770814 on Build GRCh38
rs748511310 Chr.19: 17770829-17770829 on Build GRCh38
rs374155212 Chr.19: 17770861-17770861 on Build GRCh38
rs973762960 Chr.19: 17770525-17770525 on Build GRCh38
rs1015537405 Chr.19: 17770479-17770479 on Build GRCh38
rs970252335 Chr.19: 17770542-17770542 on Build GRCh38
rs185680019 Chr.19: 17770704-17770704 on Build GRCh38
rs1035465485 Chr.19: 17770730-17770730 on Build GRCh38
rs910321275 Chr.19: 17770879-17770879 on Build GRCh38
rs762814728 Chr.19: 17770787-17770787 on Build GRCh38
rs763511675 Chr.19: 17770501-17770501 on Build GRCh38
rs374906283 Chr.19: 17770612-17770612 on Build GRCh38
rs761838524 Chr.19: 17770764-17770764 on Build GRCh38
rs775819738 Chr.19: 17770694-17770694 on Build GRCh38
rs761831659 Chr.19: 17770591-17770591 on Build GRCh38
rs768835372 Chr.19: 17770747-17770747 on Build GRCh38
rs751421628 Chr.19: 17770590-17770591 on Build GRCh38
rs767453781 Chr.19: 17770884-17770884 on Build GRCh38
rs200502191 Chr.19: 17770794-17770794 on Build GRCh38
rs192634000 Chr.19: 17770502-17770502 on Build GRCh38
rs756039563 Chr.19: 17770910-17770910 on Build GRCh38
rs772606338 Chr.19: 17770769-17770769 on Build GRCh38
rs890930064 Chr.19: 17770662-17770662 on Build GRCh38
rs757215111 Chr.19: 17770615-17770615 on Build GRCh38
rs377605612 Chr.19: 17770828-17770828 on Build GRCh38
rs959405902 Chr.19: 17770627-17770627 on Build GRCh38
rs778395329 Chr.19: 17770942-17770942 on Build GRCh38
rs763087022 Chr.19: 17770702-17770702 on Build GRCh38
rs758083719 Chr.19: 17770613-17770613 on Build GRCh38
rs898336089 Chr.19: 17770827-17770827 on Build GRCh38
rs543572863 Chr.19: 17770798-17770798 on Build GRCh38
rs747102469 Chr.19: 17770836-17770836 on Build GRCh38
rs775827578 Chr.19: 17770480-17770480 on Build GRCh38
rs761956424 Chr.19: 17770594-17770594 on Build GRCh38
rs201033930 Chr.19: 17770824-17770824 on Build GRCh38
rs754058684 Chr.19: 17770785-17770785 on Build GRCh38
rs144793428 Chr.19: 17770793-17770793 on Build GRCh38
rs779685174 Chr.19: 17770812-17770812 on Build GRCh38
rs368137662 Chr.19: 17770614-17770614 on Build GRCh38
rs753554465 Chr.19: 17770875-17770875 on Build GRCh38
rs763709054 Chr.19: 17770524-17770524 on Build GRCh38
rs148568413 Chr.19: 17770796-17770796 on Build GRCh38
rs375019038 Chr.19: 17770589-17770589 on Build GRCh38
rs778905601 Chr.19: 17770522-17770522 on Build GRCh38
rs200074791 Chr.19: 17770831-17770831 on Build GRCh38
rs142922726 Chr.19: 17770832-17770832 on Build GRCh38
rs776102334 Chr.19: 17770472-17770472 on Build GRCh38
rs981572159 Chr.19: 17770719-17770719 on Build GRCh38
rs140181398 Chr.19: 17770476-17770476 on Build GRCh38
rs746554972 Chr.19: 17770541-17770541 on Build GRCh38
rs142764492 Chr.19: 17770666-17770666 on Build GRCh38
rs767793852 Chr.19: 17770519-17770519 on Build GRCh38
rs149422647 Chr.19: 17770500-17770500 on Build GRCh38
rs140361088 Chr.19: 17770864-17770864 on Build GRCh38
rs758447573 Chr.19: 17770531-17770531 on Build GRCh38
rs756397491 Chr.19: 17770520-17770520 on Build GRCh38
rs770292906 Chr.19: 17770543-17770543 on Build GRCh38
rs766733403 Chr.19: 17770783-17770783 on Build GRCh38
rs540892735 Chr.19: 17770616-17770616 on Build GRCh38
rs374940293 Chr.19: 17770770-17770770 on Build GRCh38

View Full Product Details