Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18161092-18161634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3R2
Entrez Gene ID
5296
Gene Name
phosphoinositide-3-kinase regulatory subunit 2
Gene Aliases
MPPH, MPPH1, P85B, p85, p85-BETA
UniGene
Hs.371344
Gene Chromosome Location
Chr.19: 18153178-18170533 on Build GRCh38
Transcripts
NM_005027.3 NR_073517.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6920699 c.572C>T p.S191L Chr.19: 18161159-18161159 on Build GRCh38
6956400 c.619_633del15 p.A208_P212delALEPP Chr.19: 18161299-18161313 on Build GRCh38
6973609 c.628G>A p.E210K Chr.19: 18161308-18161308 on Build GRCh38
7293615 c.540A>G p.A180A Chr.19: 18161127-18161127 on Build GRCh38
6943905 c.534_579del46 p.L179fs*147 Chr.19: 18161121-18161166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190904075 Chr.19: 18161558-18161558 on Build GRCh38
rs745489341 Chr.19: 18161172-18161172 on Build GRCh38
rs760953177 Chr.19: 18161374-18161374 on Build GRCh38
rs746544967 Chr.19: 18161135-18161135 on Build GRCh38
rs1005463624 Chr.19: 18161136-18161136 on Build GRCh38
rs993499628 Chr.19: 18161427-18161427 on Build GRCh38
rs775944558 Chr.19: 18161182-18161182 on Build GRCh38
rs945055008 Chr.19: 18161350-18161350 on Build GRCh38
rs536421132 Chr.19: 18161210-18161210 on Build GRCh38
rs964037353 Chr.19: 18161211-18161211 on Build GRCh38
rs749574353 Chr.19: 18161196-18161196 on Build GRCh38
rs770193876 Chr.19: 18161173-18161173 on Build GRCh38
rs183510863 Chr.19: 18161209-18161209 on Build GRCh38
rs1016301486 Chr.19: 18161140-18161140 on Build GRCh38
rs764152672 Chr.19: 18161396-18161396 on Build GRCh38
rs575718040 Chr.19: 18161360-18161360 on Build GRCh38
rs978198551 Chr.19: 18161584-18161584 on Build GRCh38
rs1038312942 Chr.19: 18161391-18161391 on Build GRCh38
rs548683774 Chr.19: 18161215-18161216 on Build GRCh38
rs1017764412 Chr.19: 18161250-18161250 on Build GRCh38
rs1022508653 Chr.19: 18161572-18161572 on Build GRCh38
rs556014963 Chr.19: 18161227-18161227 on Build GRCh38
rs780890044 Chr.19: 18161171-18161171 on Build GRCh38
rs1026825273 Chr.19: 18161206-18161206 on Build GRCh38
rs977691358 Chr.19: 18161352-18161352 on Build GRCh38
rs941281903 Chr.19: 18161386-18161386 on Build GRCh38
rs553996499 Chr.19: 18161146-18161146 on Build GRCh38
rs867576830 Chr.19: 18161335-18161335 on Build GRCh38
rs1008014277 Chr.19: 18161138-18161138 on Build GRCh38
rs1001791278 Chr.19: 18161571-18161571 on Build GRCh38
rs890571669 Chr.19: 18161494-18161494 on Build GRCh38
rs752073018 Chr.19: 18161572-18161572 on Build GRCh38
rs2241088 Chr.19: 18161380-18161380 on Build GRCh38
rs772272053 Chr.19: 18161337-18161337 on Build GRCh38
rs561592967 Chr.19: 18161550-18161550 on Build GRCh38
rs1018214708 Chr.19: 18161519-18161519 on Build GRCh38
rs756748269 Chr.19: 18161151-18161151 on Build GRCh38
rs571661207 Chr.19: 18161376-18161376 on Build GRCh38
rs1008971604 Chr.19: 18161500-18161504 on Build GRCh38
rs674231 Chr.19: 18161343-18161343 on Build GRCh38
rs777507577 Chr.19: 18161134-18161134 on Build GRCh38
rs897040629 Chr.19: 18161414-18161414 on Build GRCh38
rs544647015 Chr.19: 18161364-18161364 on Build GRCh38
rs966891706 Chr.19: 18161583-18161583 on Build GRCh38
rs558127851 Chr.19: 18161371-18161371 on Build GRCh38
rs758395628 Chr.19: 18161133-18161133 on Build GRCh38
rs965362349 Chr.19: 18161264-18161264 on Build GRCh38
rs769209017 Chr.19: 18161207-18161207 on Build GRCh38
rs752389430 Chr.19: 18161131-18161131 on Build GRCh38
rs762039260 Chr.19: 18161214-18161214 on Build GRCh38
rs1026312438 Chr.19: 18161434-18161434 on Build GRCh38
rs752545087 Chr.19: 18161381-18161381 on Build GRCh38
rs989674539 Chr.19: 18161324-18161324 on Build GRCh38
rs762600324 Chr.19: 18161388-18161388 on Build GRCh38
rs896846839 Chr.19: 18161152-18161152 on Build GRCh38
rs111848145 Chr.19: 18161114-18161114 on Build GRCh38

View Full Product Details