Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21184740-21185280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF431
Entrez Gene ID
170959
Gene Name
zinc finger protein 431
Gene Aliases
-
UniGene
Hs.656394
Gene Chromosome Location
Chr.19: 21142009-21191367 on Build GRCh38
Transcripts
NM_001319124.1 NM_001319126.1 NM_001319127.1 NM_133473.2 XM_011527750.2

SNP Detail

SNP ID Chromosome Location
rs967470556 Chr.19: 21184939-21184939 on Build GRCh38
rs929059283 Chr.19: 21185097-21185097 on Build GRCh38
rs543803211 Chr.19: 21185243-21185243 on Build GRCh38
rs778680808 Chr.19: 21184925-21184925 on Build GRCh38
rs573946890 Chr.19: 21185011-21185011 on Build GRCh38
rs577950065 Chr.19: 21184868-21184868 on Build GRCh38
rs915047450 Chr.19: 21185083-21185083 on Build GRCh38
rs767006742 Chr.19: 21185136-21185137 on Build GRCh38
rs544765879 Chr.19: 21185232-21185232 on Build GRCh38
rs1056165712 Chr.19: 21184865-21184865 on Build GRCh38
rs576211359 Chr.19: 21185065-21185065 on Build GRCh38
rs553767062 Chr.19: 21184899-21184899 on Build GRCh38
rs577052056 Chr.19: 21185194-21185195 on Build GRCh38
rs898666668 Chr.19: 21184941-21184941 on Build GRCh38
rs145011836 Chr.19: 21185037-21185037 on Build GRCh38
rs574427068 Chr.19: 21185181-21185181 on Build GRCh38
rs957608897 Chr.19: 21185050-21185050 on Build GRCh38
rs923667811 Chr.19: 21185117-21185117 on Build GRCh38
rs964577435 Chr.19: 21185057-21185057 on Build GRCh38
rs923721465 Chr.19: 21184826-21184826 on Build GRCh38
rs998659765 Chr.19: 21184884-21184884 on Build GRCh38
rs901777433 Chr.19: 21185184-21185184 on Build GRCh38
rs562624036 Chr.19: 21185044-21185044 on Build GRCh38
rs1048709766 Chr.19: 21185109-21185109 on Build GRCh38
rs141186657 Chr.19: 21184847-21184847 on Build GRCh38
rs748533075 Chr.19: 21184801-21184801 on Build GRCh38
rs916252288 Chr.19: 21185202-21185202 on Build GRCh38
rs537907979 Chr.19: 21184836-21184836 on Build GRCh38
rs950220092 Chr.19: 21185081-21185081 on Build GRCh38
rs569033090 Chr.19: 21184788-21184788 on Build GRCh38
rs555508173 Chr.19: 21184767-21184767 on Build GRCh38
rs918972309 Chr.19: 21185086-21185086 on Build GRCh38
rs955477383 Chr.19: 21185036-21185036 on Build GRCh38
rs968280243 Chr.19: 21185085-21185085 on Build GRCh38
rs975071437 Chr.19: 21185076-21185076 on Build GRCh38
rs997332489 Chr.19: 21185210-21185210 on Build GRCh38
rs916044264 Chr.19: 21185075-21185076 on Build GRCh38
rs887655954 Chr.19: 21184794-21184794 on Build GRCh38
rs1020810272 Chr.19: 21185237-21185237 on Build GRCh38
rs527272742 Chr.19: 21185249-21185249 on Build GRCh38
rs895387790 Chr.19: 21184871-21184871 on Build GRCh38
rs994380076 Chr.19: 21184983-21184983 on Build GRCh38
rs943268551 Chr.19: 21185148-21185148 on Build GRCh38
rs939112250 Chr.19: 21185166-21185166 on Build GRCh38
rs977991402 Chr.19: 21185104-21185104 on Build GRCh38
rs534122552 Chr.19: 21184875-21184875 on Build GRCh38
rs946449456 Chr.19: 21184792-21184792 on Build GRCh38
rs903231160 Chr.19: 21185238-21185238 on Build GRCh38

View Full Product Details