Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21970231-21970741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF208
Entrez Gene ID
7757
Gene Name
zinc finger protein 208
Gene Aliases
PMIDP, ZNF95
UniGene
Hs.732975
Gene Chromosome Location
Chr.19: 21966095-22010990 on Build GRCh38
Transcripts
NM_007153.3 XM_017027257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
326953 c.3466C>A p.H1156N Chr.19: 21970721-21970721 on Build GRCh38
6232567 c.3481G>A p.A1161T Chr.19: 21970706-21970706 on Build GRCh38
4000608 c.3494T>A p.L1165H Chr.19: 21970693-21970693 on Build GRCh38
1611933 c.3463A>T p.T1155S Chr.19: 21970724-21970724 on Build GRCh38
6231780 c.3504A>T p.*1168C Chr.19: 21970683-21970683 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1030751551 Chr.19: 21970268-21970268 on Build GRCh38
rs552941733 Chr.19: 21970627-21970627 on Build GRCh38
rs956519010 Chr.19: 21970424-21970424 on Build GRCh38
rs561248956 Chr.19: 21970257-21970257 on Build GRCh38
rs192743421 Chr.19: 21970521-21970521 on Build GRCh38
rs969232133 Chr.19: 21970517-21970517 on Build GRCh38
rs747744366 Chr.19: 21970644-21970644 on Build GRCh38
rs577761647 Chr.19: 21970636-21970636 on Build GRCh38
rs932252999 Chr.19: 21970460-21970460 on Build GRCh38
rs144759872 Chr.19: 21970325-21970325 on Build GRCh38
rs999168090 Chr.19: 21970396-21970396 on Build GRCh38
rs2359812 Chr.19: 21970693-21970693 on Build GRCh38
rs750803697 Chr.19: 21970440-21970440 on Build GRCh38
rs960009412 Chr.19: 21970485-21970488 on Build GRCh38
rs777221001 Chr.19: 21970371-21970371 on Build GRCh38
rs375759898 Chr.19: 21970637-21970637 on Build GRCh38
rs768333254 Chr.19: 21970673-21970673 on Build GRCh38
rs757781967 Chr.19: 21970658-21970658 on Build GRCh38
rs1051470626 Chr.19: 21970341-21970341 on Build GRCh38
rs1052188958 Chr.19: 21970502-21970502 on Build GRCh38
rs1031868135 Chr.19: 21970419-21970419 on Build GRCh38
rs931849029 Chr.19: 21970564-21970564 on Build GRCh38
rs370923661 Chr.19: 21970565-21970565 on Build GRCh38
rs1013439266 Chr.19: 21970509-21970509 on Build GRCh38
rs752873289 Chr.19: 21970706-21970706 on Build GRCh38
rs983534149 Chr.19: 21970384-21970384 on Build GRCh38
rs761570474 Chr.19: 21970692-21970692 on Build GRCh38
rs200355555 Chr.19: 21970666-21970666 on Build GRCh38
rs759650472 Chr.19: 21970701-21970701 on Build GRCh38
rs925521568 Chr.19: 21970679-21970679 on Build GRCh38
rs552011984 Chr.19: 21970472-21970472 on Build GRCh38
rs532714246 Chr.19: 21970441-21970441 on Build GRCh38
rs892046899 Chr.19: 21970583-21970583 on Build GRCh38
rs890094369 Chr.19: 21970388-21970388 on Build GRCh38
rs752280178 Chr.19: 21970576-21970576 on Build GRCh38
rs185187291 Chr.19: 21970660-21970660 on Build GRCh38
rs935009780 Chr.19: 21970684-21970684 on Build GRCh38
rs765431222 Chr.19: 21970703-21970703 on Build GRCh38
rs991953421 Chr.19: 21970279-21970279 on Build GRCh38
rs896141945 Chr.19: 21970702-21970702 on Build GRCh38
rs983376990 Chr.19: 21970588-21970588 on Build GRCh38
rs751910938 Chr.19: 21970651-21970651 on Build GRCh38
rs890228592 Chr.19: 21970572-21970572 on Build GRCh38
rs369950998 Chr.19: 21970643-21970643 on Build GRCh38
rs765746439 Chr.19: 21970404-21970404 on Build GRCh38
rs369562799 Chr.19: 21970709-21970709 on Build GRCh38
rs779629885 Chr.19: 21970671-21970671 on Build GRCh38
rs773758249 Chr.19: 21970635-21970635 on Build GRCh38
rs762189633 Chr.19: 21970373-21970373 on Build GRCh38
rs148539034 Chr.19: 21970479-21970479 on Build GRCh38
rs960539726 Chr.19: 21970269-21970269 on Build GRCh38
rs768802872 Chr.19: 21970349-21970349 on Build GRCh38
rs999136160 Chr.19: 21970573-21970573 on Build GRCh38
rs755642487 Chr.19: 21970619-21970619 on Build GRCh38
rs188115366 Chr.19: 21970478-21970478 on Build GRCh38
rs775847108 Chr.19: 21970334-21970334 on Build GRCh38
rs983446444 Chr.19: 21970686-21970686 on Build GRCh38
rs141876096 Chr.19: 21970507-21970507 on Build GRCh38
rs1026895406 Chr.19: 21970547-21970547 on Build GRCh38
rs1021886581 Chr.19: 21970608-21970608 on Build GRCh38
rs149405849 Chr.19: 21970285-21970285 on Build GRCh38
rs1987570 Chr.19: 21970335-21970335 on Build GRCh38
rs941979142 Chr.19: 21970525-21970525 on Build GRCh38
rs774191008 Chr.19: 21970683-21970683 on Build GRCh38
rs376979771 Chr.19: 21970630-21970630 on Build GRCh38
rs534695172 Chr.19: 21970603-21970603 on Build GRCh38
rs146087898 Chr.19: 21970515-21970515 on Build GRCh38
rs922199673 Chr.19: 21970458-21970458 on Build GRCh38
rs907930298 Chr.19: 21970401-21970401 on Build GRCh38
rs1037624364 Chr.19: 21970546-21970546 on Build GRCh38
rs921725515 Chr.19: 21970556-21970556 on Build GRCh38
rs748912528 Chr.19: 21970672-21970672 on Build GRCh38
rs907982344 Chr.19: 21970617-21970617 on Build GRCh38
rs1012203151 Chr.19: 21970598-21970598 on Build GRCh38
rs928723181 Chr.19: 21970480-21970480 on Build GRCh38
rs1043983024 Chr.19: 21970467-21970467 on Build GRCh38

View Full Product Details