Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 22180915-22181411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF676
Entrez Gene ID
163223
Gene Name
zinc finger protein 676
Gene Aliases
-
UniGene
Hs.55452
Gene Chromosome Location
Chr.19: 22179101-22196951 on Build GRCh38
Transcripts
NM_001001411.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4131566 c.567T>A p.S189R Chr.19: 22181150-22181150 on Build GRCh38
303649 c.700T>A p.F234I Chr.19: 22181017-22181017 on Build GRCh38
4832839 c.494G>C p.R165T Chr.19: 22181223-22181223 on Build GRCh38
3531658 c.754C>T p.P252S Chr.19: 22180963-22180963 on Build GRCh38
1750769 c.543C>G p.S181S Chr.19: 22181174-22181174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371079322 Chr.19: 22181014-22181014 on Build GRCh38
rs754722505 Chr.19: 22181357-22181357 on Build GRCh38
rs757292963 Chr.19: 22181005-22181015 on Build GRCh38
rs879193766 Chr.19: 22180983-22180983 on Build GRCh38
rs768097893 Chr.19: 22180943-22180943 on Build GRCh38
rs769716448 Chr.19: 22181104-22181104 on Build GRCh38
rs756843174 Chr.19: 22181164-22181164 on Build GRCh38
rs745695211 Chr.19: 22181066-22181066 on Build GRCh38
rs748147750 Chr.19: 22181263-22181263 on Build GRCh38
rs938116360 Chr.19: 22181374-22181374 on Build GRCh38
rs777400222 Chr.19: 22181156-22181158 on Build GRCh38
rs752604889 Chr.19: 22181367-22181367 on Build GRCh38
rs755960965 Chr.19: 22181185-22181185 on Build GRCh38
rs200737633 Chr.19: 22180947-22180947 on Build GRCh38
rs772791660 Chr.19: 22180975-22180975 on Build GRCh38
rs772808312 Chr.19: 22181327-22181327 on Build GRCh38
rs772169464 Chr.19: 22181118-22181118 on Build GRCh38
rs765582913 Chr.19: 22181135-22181135 on Build GRCh38
rs755899540 Chr.19: 22181113-22181113 on Build GRCh38
rs545560102 Chr.19: 22180982-22180982 on Build GRCh38
rs779786446 Chr.19: 22181252-22181252 on Build GRCh38
rs145271931 Chr.19: 22181216-22181216 on Build GRCh38
rs765070522 Chr.19: 22181363-22181363 on Build GRCh38
rs371456888 Chr.19: 22181157-22181157 on Build GRCh38
rs552127917 Chr.19: 22181127-22181127 on Build GRCh38
rs746645272 Chr.19: 22181095-22181095 on Build GRCh38
rs1027404980 Chr.19: 22181251-22181251 on Build GRCh38
rs142355819 Chr.19: 22180994-22180994 on Build GRCh38
rs781293893 Chr.19: 22181281-22181281 on Build GRCh38
rs776596666 Chr.19: 22181065-22181065 on Build GRCh38
rs531433977 Chr.19: 22181011-22181011 on Build GRCh38
rs543253815 Chr.19: 22181359-22181359 on Build GRCh38
rs755761897 Chr.19: 22181249-22181249 on Build GRCh38
rs386807906 Chr.19: 22181156-22181161 on Build GRCh38
rs61745988 Chr.19: 22181375-22181375 on Build GRCh38
rs759826017 Chr.19: 22181129-22181129 on Build GRCh38
rs570225845 Chr.19: 22181150-22181150 on Build GRCh38
rs772857713 Chr.19: 22181316-22181320 on Build GRCh38
rs753525785 Chr.19: 22181254-22181254 on Build GRCh38
rs767194516 Chr.19: 22181240-22181240 on Build GRCh38
rs765764630 Chr.19: 22181089-22181089 on Build GRCh38
rs911738136 Chr.19: 22181316-22181316 on Build GRCh38
rs879253472 Chr.19: 22181303-22181303 on Build GRCh38
rs780605725 Chr.19: 22181079-22181080 on Build GRCh38
rs7253392 Chr.19: 22181059-22181059 on Build GRCh38
rs763400454 Chr.19: 22181224-22181224 on Build GRCh38
rs768436729 Chr.19: 22181191-22181191 on Build GRCh38
rs759564847 Chr.19: 22181010-22181010 on Build GRCh38
rs371219264 Chr.19: 22181340-22181340 on Build GRCh38
rs199880241 Chr.19: 22181044-22181044 on Build GRCh38
rs767498279 Chr.19: 22181072-22181072 on Build GRCh38
rs777166606 Chr.19: 22181094-22181094 on Build GRCh38
rs754813419 Chr.19: 22181256-22181256 on Build GRCh38
rs370343274 Chr.19: 22181247-22181247 on Build GRCh38
rs749217105 Chr.19: 22181189-22181189 on Build GRCh38
rs771727885 Chr.19: 22180967-22180967 on Build GRCh38
rs762460001 Chr.19: 22181320-22181320 on Build GRCh38
rs192725054 Chr.19: 22181287-22181287 on Build GRCh38
rs769830637 Chr.19: 22181067-22181067 on Build GRCh38
rs7252397 Chr.19: 22181085-22181085 on Build GRCh38
rs367961273 Chr.19: 22181026-22181026 on Build GRCh38
rs112721128 Chr.19: 22181144-22181144 on Build GRCh38
rs770981051 Chr.19: 22181269-22181269 on Build GRCh38
rs145180034 Chr.19: 22181045-22181045 on Build GRCh38
rs769862523 Chr.19: 22181289-22181289 on Build GRCh38
rs746650384 Chr.19: 22181047-22181047 on Build GRCh38
rs376158454 Chr.19: 22181356-22181356 on Build GRCh38
rs776648123 Chr.19: 22180997-22180997 on Build GRCh38
rs576145652 Chr.19: 22181325-22181325 on Build GRCh38
rs200144586 Chr.19: 22181109-22181109 on Build GRCh38
rs537289839 Chr.19: 22181151-22181151 on Build GRCh38
rs563924087 Chr.19: 22180989-22180989 on Build GRCh38
rs746324688 Chr.19: 22181046-22181056 on Build GRCh38
rs781016866 Chr.19: 22181099-22181099 on Build GRCh38
rs7253403 Chr.19: 22181077-22181077 on Build GRCh38
rs369532914 Chr.19: 22181211-22181211 on Build GRCh38
rs201929387 Chr.19: 22181019-22181019 on Build GRCh38
rs367617888 Chr.19: 22181119-22181119 on Build GRCh38
rs768917229 Chr.19: 22181159-22181160 on Build GRCh38
rs749241989 Chr.19: 22181114-22181114 on Build GRCh38
rs780927079 Chr.19: 22181179-22181179 on Build GRCh38
rs761405736 Chr.19: 22181239-22181239 on Build GRCh38
rs770255323 Chr.19: 22181067-22181068 on Build GRCh38
rs755387397 Chr.19: 22181087-22181087 on Build GRCh38
rs200880635 Chr.19: 22181186-22181186 on Build GRCh38
rs879017683 Chr.19: 22181009-22181009 on Build GRCh38
rs767765362 Chr.19: 22181013-22181013 on Build GRCh38
rs769133073 Chr.19: 22181317-22181317 on Build GRCh38
rs72479456 Chr.19: 22181023-22181024 on Build GRCh38
rs115951008 Chr.19: 22181229-22181229 on Build GRCh38
rs371296106 Chr.19: 22181012-22181013 on Build GRCh38
rs111749597 Chr.19: 22181102-22181102 on Build GRCh38
rs753215116 Chr.19: 22181091-22181091 on Build GRCh38
rs879038542 Chr.19: 22181015-22181015 on Build GRCh38
rs756933126 Chr.19: 22181096-22181096 on Build GRCh38
rs7252593 Chr.19: 22181024-22181024 on Build GRCh38
rs372676798 Chr.19: 22181259-22181259 on Build GRCh38
rs961714317 Chr.19: 22181235-22181235 on Build GRCh38
rs763413284 Chr.19: 22181156-22181156 on Build GRCh38
rs747675263 Chr.19: 22181035-22181035 on Build GRCh38
rs763163054 Chr.19: 22181300-22181300 on Build GRCh38
rs755511645 Chr.19: 22181027-22181027 on Build GRCh38
rs370500138 Chr.19: 22181143-22181143 on Build GRCh38
rs201994000 Chr.19: 22181068-22181068 on Build GRCh38
rs774986241 Chr.19: 22181319-22181319 on Build GRCh38
rs757863408 Chr.19: 22181092-22181092 on Build GRCh38
rs773425783 Chr.19: 22181073-22181073 on Build GRCh38
rs988032831 Chr.19: 22181321-22181321 on Build GRCh38
rs749800250 Chr.19: 22180940-22180940 on Build GRCh38
rs1002365548 Chr.19: 22181181-22181181 on Build GRCh38
rs111727602 Chr.19: 22181108-22181108 on Build GRCh38
rs754173504 Chr.19: 22181086-22181086 on Build GRCh38
rs781280530 Chr.19: 22181024-22181024 on Build GRCh38
rs557677916 Chr.19: 22181299-22181299 on Build GRCh38
rs770921118 Chr.19: 22180995-22180995 on Build GRCh38
rs765887947 Chr.19: 22181343-22181343 on Build GRCh38
rs759807597 Chr.19: 22181208-22181208 on Build GRCh38
rs201045239 Chr.19: 22181199-22181199 on Build GRCh38
rs533650030 Chr.19: 22181121-22181121 on Build GRCh38
rs189222594 Chr.19: 22181074-22181074 on Build GRCh38
rs376692957 Chr.19: 22181180-22181180 on Build GRCh38
rs747716880 Chr.19: 22180953-22180953 on Build GRCh38
rs878878539 Chr.19: 22180988-22180988 on Build GRCh38
rs778929725 Chr.19: 22181196-22181196 on Build GRCh38
rs766537711 Chr.19: 22181125-22181125 on Build GRCh38
rs7252604 Chr.19: 22181042-22181042 on Build GRCh38
rs751068129 Chr.19: 22181163-22181163 on Build GRCh38
rs764453399 Chr.19: 22181225-22181225 on Build GRCh38
rs747031273 Chr.19: 22181197-22181197 on Build GRCh38
rs776826213 Chr.19: 22181204-22181204 on Build GRCh38
rs761947904 Chr.19: 22181071-22181071 on Build GRCh38
rs770168459 Chr.19: 22181213-22181213 on Build GRCh38
rs529239131 Chr.19: 22181033-22181033 on Build GRCh38
rs1052368661 Chr.19: 22181136-22181136 on Build GRCh38
rs775797149 Chr.19: 22181298-22181298 on Build GRCh38

View Full Product Details