Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40699836-40700333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ8B
Entrez Gene ID
79934
Gene Name
coenzyme Q8B
Gene Aliases
ADCK4, NPHS9
UniGene
Hs.130712
Gene Chromosome Location
Chr.19: 40691529-40716885 on Build GRCh38
Transcripts
NM_001142555.2 NM_024876.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7201388 c.1061G>A p.C354Y Chr.19: 40700149-40700149 on Build GRCh38
1730310 c.1110G>T p.W370C Chr.19: 40700100-40700100 on Build GRCh38
6865043 c.1096A>C p.T366P Chr.19: 40700114-40700114 on Build GRCh38
1712319 c.1115A>G p.N372S Chr.19: 40700095-40700095 on Build GRCh38
1304613 c.1056G>A p.T352T Chr.19: 40700154-40700154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753687112 Chr.19: 40700119-40700119 on Build GRCh38
rs202196406 Chr.19: 40700297-40700297 on Build GRCh38
rs376533654 Chr.19: 40700036-40700036 on Build GRCh38
rs775232727 Chr.19: 40700176-40700176 on Build GRCh38
rs886763853 Chr.19: 40700032-40700032 on Build GRCh38
rs531989896 Chr.19: 40700144-40700144 on Build GRCh38
rs2246095 Chr.19: 40700303-40700303 on Build GRCh38
rs760950087 Chr.19: 40700307-40700308 on Build GRCh38
rs760280495 Chr.19: 40700299-40700299 on Build GRCh38
rs761736348 Chr.19: 40700197-40700197 on Build GRCh38
rs376627440 Chr.19: 40700302-40700302 on Build GRCh38
rs36012476 Chr.19: 40700155-40700155 on Build GRCh38
rs747299492 Chr.19: 40700029-40700029 on Build GRCh38
rs763887289 Chr.19: 40700254-40700254 on Build GRCh38
rs758422079 Chr.19: 40700218-40700218 on Build GRCh38
rs760192949 Chr.19: 40700189-40700189 on Build GRCh38
rs762699398 Chr.19: 40700205-40700205 on Build GRCh38
rs370946869 Chr.19: 40700156-40700156 on Build GRCh38
rs1000981334 Chr.19: 40700101-40700101 on Build GRCh38
rs753488008 Chr.19: 40700196-40700196 on Build GRCh38
rs973640042 Chr.19: 40699999-40699999 on Build GRCh38
rs763849360 Chr.19: 40700194-40700194 on Build GRCh38
rs868835135 Chr.19: 40700245-40700245 on Build GRCh38
rs548120543 Chr.19: 40700253-40700253 on Build GRCh38
rs772145293 Chr.19: 40700024-40700024 on Build GRCh38
rs368182892 Chr.19: 40700307-40700307 on Build GRCh38
rs754994798 Chr.19: 40700264-40700264 on Build GRCh38
rs531330340 Chr.19: 40699966-40699966 on Build GRCh38
rs751645543 Chr.19: 40700263-40700263 on Build GRCh38
rs750338593 Chr.19: 40700132-40700132 on Build GRCh38
rs376101078 Chr.19: 40700306-40700306 on Build GRCh38
rs536536694 Chr.19: 40700249-40700249 on Build GRCh38
rs770553076 Chr.19: 40700081-40700081 on Build GRCh38
rs773290357 Chr.19: 40700201-40700201 on Build GRCh38
rs749608985 Chr.19: 40700277-40700277 on Build GRCh38
rs775609626 Chr.19: 40700025-40700025 on Build GRCh38
rs767092972 Chr.19: 40700078-40700078 on Build GRCh38
rs757057069 Chr.19: 40700120-40700120 on Build GRCh38
rs775130346 Chr.19: 40700184-40700184 on Build GRCh38
rs779854319 Chr.19: 40700221-40700221 on Build GRCh38
rs771470507 Chr.19: 40700295-40700295 on Build GRCh38
rs769884860 Chr.19: 40700270-40700270 on Build GRCh38
rs899437407 Chr.19: 40699967-40699967 on Build GRCh38
rs765739053 Chr.19: 40700050-40700050 on Build GRCh38
rs762162639 Chr.19: 40700047-40700047 on Build GRCh38
rs758999160 Chr.19: 40700074-40700074 on Build GRCh38
rs765124326 Chr.19: 40700200-40700200 on Build GRCh38
rs879013998 Chr.19: 40700185-40700185 on Build GRCh38
rs759199445 Chr.19: 40700143-40700143 on Build GRCh38
rs943544164 Chr.19: 40699956-40699956 on Build GRCh38
rs771948613 Chr.19: 40700191-40700192 on Build GRCh38
rs778256979 Chr.19: 40700275-40700275 on Build GRCh38
rs78185540 Chr.19: 40700085-40700085 on Build GRCh38
rs1039763398 Chr.19: 40700088-40700088 on Build GRCh38
rs773416977 Chr.19: 40700164-40700164 on Build GRCh38
rs993690033 Chr.19: 40700002-40700002 on Build GRCh38
rs56056214 Chr.19: 40700091-40700091 on Build GRCh38
rs375860406 Chr.19: 40700167-40700167 on Build GRCh38
rs373220747 Chr.19: 40700016-40700016 on Build GRCh38
rs146616224 Chr.19: 40700126-40700126 on Build GRCh38
rs777039616 Chr.19: 40700041-40700041 on Build GRCh38
rs747080363 Chr.19: 40700145-40700145 on Build GRCh38
rs760458734 Chr.19: 40700090-40700090 on Build GRCh38
rs561515341 Chr.19: 40699880-40699880 on Build GRCh38
rs1052272822 Chr.19: 40699910-40699910 on Build GRCh38
rs747867828 Chr.19: 40700132-40700133 on Build GRCh38
rs758241049 Chr.19: 40700136-40700136 on Build GRCh38
rs768934453 Chr.19: 40700154-40700154 on Build GRCh38
rs200061712 Chr.19: 40700177-40700177 on Build GRCh38
rs1002049078 Chr.19: 40700125-40700125 on Build GRCh38
rs773711662 Chr.19: 40700063-40700063 on Build GRCh38
rs139747226 Chr.19: 40700265-40700265 on Build GRCh38
rs770154433 Chr.19: 40700162-40700162 on Build GRCh38
rs778943760 Chr.19: 40700066-40700066 on Build GRCh38
rs374668619 Chr.19: 40700266-40700266 on Build GRCh38
rs910762306 Chr.19: 40699946-40699946 on Build GRCh38

View Full Product Details