Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44148686-44149147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF234
Entrez Gene ID
10780
Gene Name
zinc finger protein 234
Gene Aliases
HZF4, ZNF269
UniGene
Hs.235992
Gene Chromosome Location
Chr.19: 44141530-44160309 on Build GRCh38
Transcripts
NM_001144824.1 NM_006630.2 XM_006722974.3 XM_017026149.1 XM_017026150.1 XM_017026151.1 XM_017026152.1 XM_017026153.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6289839 c.93T>G p.N31K Chr.19: 44148848-44148848 on Build GRCh38
3289913 c.130C>A p.L44M Chr.19: 44148885-44148885 on Build GRCh38
377187 c.55G>T p.E19* Chr.19: 44148810-44148810 on Build GRCh38
997826 c.72G>A p.L24L Chr.19: 44148827-44148827 on Build GRCh38
5905110 c.46G>A p.V16I Chr.19: 44148801-44148801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938199855 Chr.19: 44149118-44149118 on Build GRCh38
rs113757822 Chr.19: 44148931-44148931 on Build GRCh38
rs781575396 Chr.19: 44148728-44148728 on Build GRCh38
rs926796732 Chr.19: 44149083-44149083 on Build GRCh38
rs770268889 Chr.19: 44148744-44148744 on Build GRCh38
rs749771393 Chr.19: 44148924-44148924 on Build GRCh38
rs765406192 Chr.19: 44148716-44148720 on Build GRCh38
rs2293587 Chr.19: 44148801-44148801 on Build GRCh38
rs761213944 Chr.19: 44148792-44148792 on Build GRCh38
rs182403374 Chr.19: 44148962-44148962 on Build GRCh38
rs781415073 Chr.19: 44148893-44148893 on Build GRCh38
rs547154147 Chr.19: 44148885-44148885 on Build GRCh38
rs763833829 Chr.19: 44148961-44148961 on Build GRCh38
rs370341936 Chr.19: 44148779-44148779 on Build GRCh38
rs759705049 Chr.19: 44148785-44148785 on Build GRCh38
rs748488913 Chr.19: 44148726-44148726 on Build GRCh38
rs998087135 Chr.19: 44148767-44148767 on Build GRCh38
rs1023979093 Chr.19: 44149011-44149011 on Build GRCh38
rs368405532 Chr.19: 44148824-44148824 on Build GRCh38
rs993050251 Chr.19: 44148864-44148864 on Build GRCh38
rs867486654 Chr.19: 44148799-44148820 on Build GRCh38
rs868305225 Chr.19: 44149006-44149006 on Build GRCh38
rs759613290 Chr.19: 44148835-44148835 on Build GRCh38
rs568599489 Chr.19: 44148990-44148990 on Build GRCh38
rs1052334345 Chr.19: 44149109-44149109 on Build GRCh38
rs933780131 Chr.19: 44149081-44149081 on Build GRCh38
rs762594566 Chr.19: 44148851-44148851 on Build GRCh38
rs765242513 Chr.19: 44148979-44148979 on Build GRCh38
rs548818218 Chr.19: 44148932-44148932 on Build GRCh38
rs201162038 Chr.19: 44148827-44148827 on Build GRCh38
rs776754760 Chr.19: 44148833-44148833 on Build GRCh38
rs982785192 Chr.19: 44149072-44149072 on Build GRCh38
rs893787773 Chr.19: 44148808-44148808 on Build GRCh38
rs777728486 Chr.19: 44148750-44148750 on Build GRCh38
rs938080392 Chr.19: 44148797-44148797 on Build GRCh38
rs778053224 Chr.19: 44148850-44148851 on Build GRCh38
rs773732241 Chr.19: 44148788-44148788 on Build GRCh38
rs753048194 Chr.19: 44148905-44148905 on Build GRCh38
rs754623417 Chr.19: 44148890-44148890 on Build GRCh38
rs1031751305 Chr.19: 44148843-44148843 on Build GRCh38
rs76244995 Chr.19: 44148799-44148799 on Build GRCh38
rs368959493 Chr.19: 44148714-44148714 on Build GRCh38
rs751661791 Chr.19: 44148877-44148878 on Build GRCh38
rs752894687 Chr.19: 44148736-44148736 on Build GRCh38
rs539444358 Chr.19: 44148743-44148743 on Build GRCh38
rs772348773 Chr.19: 44148781-44148781 on Build GRCh38
rs749429903 Chr.19: 44148774-44148774 on Build GRCh38
rs188064720 Chr.19: 44148991-44148991 on Build GRCh38
rs371472588 Chr.19: 44148925-44148925 on Build GRCh38
rs970980298 Chr.19: 44149018-44149018 on Build GRCh38
rs537656924 Chr.19: 44148965-44148965 on Build GRCh38
rs3215367 Chr.19: 44148720-44148724 on Build GRCh38
rs778321383 Chr.19: 44148920-44148920 on Build GRCh38
rs748638862 Chr.19: 44148742-44148742 on Build GRCh38
rs756423016 Chr.19: 44148906-44148906 on Build GRCh38
rs1017411564 Chr.19: 44148960-44148960 on Build GRCh38
rs1027445727 Chr.19: 44148976-44148976 on Build GRCh38
rs373117172 Chr.19: 44148844-44148844 on Build GRCh38
rs774602605 Chr.19: 44148782-44148782 on Build GRCh38
rs751283016 Chr.19: 44148887-44148887 on Build GRCh38
rs758417647 Chr.19: 44148846-44148847 on Build GRCh38
rs751227947 Chr.19: 44148717-44148717 on Build GRCh38
rs750378916 Chr.19: 44148830-44148830 on Build GRCh38

View Full Product Details