Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45151327-45151825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKPD1
Entrez Gene ID
284353
Gene Name
NTPase, KAP family P-loop domain containing 1
Gene Aliases
-
UniGene
Hs.299256
Gene Chromosome Location
Chr.19: 45149750-45162831 on Build GRCh38
Transcripts
NM_198478.3 XM_011526799.2 XM_011526802.2 XM_011526804.2 XM_011526805.2

SNP Detail

SNP ID Chromosome Location
rs1057384826 Chr.19: 45151766-45151766 on Build GRCh38
rs867625253 Chr.19: 45151560-45151560 on Build GRCh38
rs4330014 Chr.19: 45151629-45151629 on Build GRCh38
rs913377975 Chr.19: 45151641-45151641 on Build GRCh38
rs908815884 Chr.19: 45151574-45151574 on Build GRCh38
rs192224730 Chr.19: 45151762-45151762 on Build GRCh38
rs117181329 Chr.19: 45151598-45151598 on Build GRCh38
rs116106679 Chr.19: 45151631-45151631 on Build GRCh38
rs902994227 Chr.19: 45151706-45151706 on Build GRCh38
rs903487774 Chr.19: 45151418-45151418 on Build GRCh38
rs531670017 Chr.19: 45151456-45151456 on Build GRCh38
rs940155037 Chr.19: 45151785-45151785 on Build GRCh38
rs565662923 Chr.19: 45151666-45151666 on Build GRCh38
rs765271065 Chr.19: 45151466-45151466 on Build GRCh38
rs913639449 Chr.19: 45151401-45151401 on Build GRCh38
rs752659658 Chr.19: 45151572-45151572 on Build GRCh38
rs758292248 Chr.19: 45151623-45151623 on Build GRCh38
rs1047832684 Chr.19: 45151504-45151504 on Build GRCh38
rs915955041 Chr.19: 45151554-45151554 on Build GRCh38
rs763941672 Chr.19: 45151746-45151746 on Build GRCh38
rs71352254 Chr.19: 45151513-45151513 on Build GRCh38
rs938828766 Chr.19: 45151376-45151376 on Build GRCh38
rs533293282 Chr.19: 45151611-45151611 on Build GRCh38
rs957341214 Chr.19: 45151535-45151535 on Build GRCh38
rs977418040 Chr.19: 45151497-45151497 on Build GRCh38
rs376849417 Chr.19: 45151727-45151727 on Build GRCh38
rs563805868 Chr.19: 45151760-45151760 on Build GRCh38
rs563001304 Chr.19: 45151416-45151416 on Build GRCh38
rs923179062 Chr.19: 45151761-45151761 on Build GRCh38
rs1036016251 Chr.19: 45151383-45151385 on Build GRCh38
rs542822678 Chr.19: 45151400-45151400 on Build GRCh38
rs945046627 Chr.19: 45151402-45151402 on Build GRCh38
rs115379353 Chr.19: 45151632-45151632 on Build GRCh38
rs994465958 Chr.19: 45151618-45151618 on Build GRCh38
rs943048988 Chr.19: 45151691-45151691 on Build GRCh38
rs940266277 Chr.19: 45151583-45151583 on Build GRCh38
rs867240442 Chr.19: 45151359-45151359 on Build GRCh38
rs1039399110 Chr.19: 45151708-45151708 on Build GRCh38
rs759655335 Chr.19: 45151405-45151405 on Build GRCh38
rs1006269807 Chr.19: 45151788-45151788 on Build GRCh38
rs898938074 Chr.19: 45151399-45151399 on Build GRCh38
rs943286576 Chr.19: 45151426-45151426 on Build GRCh38
rs991942073 Chr.19: 45151388-45151388 on Build GRCh38
rs950302402 Chr.19: 45151568-45151568 on Build GRCh38
rs867292553 Chr.19: 45151743-45151743 on Build GRCh38
rs966461669 Chr.19: 45151647-45151647 on Build GRCh38
rs187319433 Chr.19: 45151367-45151367 on Build GRCh38
rs940108268 Chr.19: 45151530-45151530 on Build GRCh38
rs117540719 Chr.19: 45151683-45151683 on Build GRCh38
rs1037752531 Chr.19: 45151796-45151796 on Build GRCh38
rs1047573094 Chr.19: 45151772-45151772 on Build GRCh38
rs957367391 Chr.19: 45151755-45151755 on Build GRCh38
rs998743714 Chr.19: 45151707-45151707 on Build GRCh38
rs991539812 Chr.19: 45151546-45151546 on Build GRCh38
rs984046297 Chr.19: 45151523-45151523 on Build GRCh38
rs908647950 Chr.19: 45151527-45151527 on Build GRCh38
rs928804582 Chr.19: 45151375-45151375 on Build GRCh38
rs908728035 Chr.19: 45151782-45151782 on Build GRCh38
rs1016691794 Chr.19: 45151563-45151563 on Build GRCh38
rs529107934 Chr.19: 45151680-45151680 on Build GRCh38
rs1019314784 Chr.19: 45151462-45151462 on Build GRCh38
rs886245722 Chr.19: 45151775-45151775 on Build GRCh38
rs369182454 Chr.19: 45151469-45151469 on Build GRCh38
rs748346982 Chr.19: 45151654-45151654 on Build GRCh38
rs1045892312 Chr.19: 45151573-45151573 on Build GRCh38
rs953337999 Chr.19: 45151443-45151443 on Build GRCh38
rs974324750 Chr.19: 45151795-45151795 on Build GRCh38
rs979168477 Chr.19: 45151679-45151679 on Build GRCh38
rs886509135 Chr.19: 45151552-45151552 on Build GRCh38

View Full Product Details