Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48123083-48123591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIG1
Entrez Gene ID
3978
Gene Name
DNA ligase 1
Gene Aliases
-
UniGene
Hs.1770
Gene Chromosome Location
Chr.19: 48115445-48170603 on Build GRCh38
Transcripts
NM_000234.2 NM_001289063.1 NM_001289064.1 NM_001320970.1 NM_001320971.1 NR_110296.1 NR_135497.1 NR_135498.1 NR_135499.1 NR_135500.1 NR_135501.1 XM_017026809.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
390800 c.2076G>T p.E692D Chr.19: 48123247-48123247 on Build GRCh38
4434763 c.2045T>C p.L682P Chr.19: 48123278-48123278 on Build GRCh38
4079800 c.2112C>T p.I704I Chr.19: 48123211-48123211 on Build GRCh38
998867 c.2052G>T p.E684D Chr.19: 48123271-48123271 on Build GRCh38
6841884 c.2125G>A p.E709K Chr.19: 48123198-48123198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55950593 Chr.19: 48123309-48123309 on Build GRCh38
rs969486790 Chr.19: 48123242-48123242 on Build GRCh38
rs747098797 Chr.19: 48123300-48123300 on Build GRCh38
rs868452926 Chr.19: 48123180-48123180 on Build GRCh38
rs765248612 Chr.19: 48123344-48123344 on Build GRCh38
rs553406550 Chr.19: 48123310-48123310 on Build GRCh38
rs763704726 Chr.19: 48123407-48123407 on Build GRCh38
rs148955551 Chr.19: 48123524-48123524 on Build GRCh38
rs754729163 Chr.19: 48123214-48123214 on Build GRCh38
rs578075828 Chr.19: 48123375-48123375 on Build GRCh38
rs369389106 Chr.19: 48123232-48123232 on Build GRCh38
rs118108412 Chr.19: 48123137-48123137 on Build GRCh38
rs767523202 Chr.19: 48123163-48123163 on Build GRCh38
rs959539203 Chr.19: 48123428-48123428 on Build GRCh38
rs540584531 Chr.19: 48123404-48123404 on Build GRCh38
rs202203416 Chr.19: 48123329-48123329 on Build GRCh38
rs756981021 Chr.19: 48123362-48123362 on Build GRCh38
rs760954030 Chr.19: 48123191-48123191 on Build GRCh38
rs142066936 Chr.19: 48123241-48123241 on Build GRCh38
rs753653757 Chr.19: 48123358-48123358 on Build GRCh38
rs773159176 Chr.19: 48123160-48123160 on Build GRCh38
rs914160739 Chr.19: 48123130-48123130 on Build GRCh38
rs949817453 Chr.19: 48123533-48123533 on Build GRCh38
rs755830886 Chr.19: 48123349-48123349 on Build GRCh38
rs759535301 Chr.19: 48123162-48123162 on Build GRCh38
rs769895197 Chr.19: 48123211-48123211 on Build GRCh38
rs914172971 Chr.19: 48123307-48123307 on Build GRCh38
rs960813552 Chr.19: 48123534-48123534 on Build GRCh38
rs748470299 Chr.19: 48123202-48123202 on Build GRCh38
rs759018012 Chr.19: 48123275-48123275 on Build GRCh38
rs377540560 Chr.19: 48123213-48123213 on Build GRCh38
rs377682304 Chr.19: 48123238-48123238 on Build GRCh38
rs762097925 Chr.19: 48123265-48123265 on Build GRCh38
rs767020971 Chr.19: 48123328-48123328 on Build GRCh38
rs568934454 Chr.19: 48123283-48123283 on Build GRCh38
rs558235828 Chr.19: 48123365-48123365 on Build GRCh38
rs56165744 Chr.19: 48123199-48123199 on Build GRCh38
rs542539719 Chr.19: 48123475-48123475 on Build GRCh38
rs140821248 Chr.19: 48123210-48123210 on Build GRCh38
rs138776333 Chr.19: 48123427-48123427 on Build GRCh38
rs779865980 Chr.19: 48123123-48123123 on Build GRCh38
rs78651054 Chr.19: 48123400-48123400 on Build GRCh38
rs750752539 Chr.19: 48123270-48123270 on Build GRCh38
rs200644453 Chr.19: 48123250-48123250 on Build GRCh38
rs527311159 Chr.19: 48123531-48123531 on Build GRCh38
rs549131231 Chr.19: 48123268-48123268 on Build GRCh38
rs917012169 Chr.19: 48123518-48123518 on Build GRCh38
rs373324917 Chr.19: 48123237-48123237 on Build GRCh38
rs3731009 Chr.19: 48123132-48123132 on Build GRCh38
rs1016798424 Chr.19: 48123487-48123487 on Build GRCh38
rs200049505 Chr.19: 48123308-48123308 on Build GRCh38
rs553586573 Chr.19: 48123115-48123115 on Build GRCh38
rs531348489 Chr.19: 48123288-48123288 on Build GRCh38
rs575899041 Chr.19: 48123333-48123333 on Build GRCh38
rs752760338 Chr.19: 48123167-48123167 on Build GRCh38
rs3731007 Chr.19: 48123347-48123347 on Build GRCh38
rs754078742 Chr.19: 48123156-48123159 on Build GRCh38
rs747923235 Chr.19: 48123225-48123225 on Build GRCh38
rs763364446 Chr.19: 48123326-48123326 on Build GRCh38
rs755164186 Chr.19: 48123334-48123335 on Build GRCh38
rs764267948 Chr.19: 48123198-48123198 on Build GRCh38
rs769639094 Chr.19: 48123139-48123139 on Build GRCh38
rs908229399 Chr.19: 48123561-48123561 on Build GRCh38
rs573367714 Chr.19: 48123116-48123116 on Build GRCh38
rs140087554 Chr.19: 48123201-48123201 on Build GRCh38
rs776848611 Chr.19: 48123251-48123251 on Build GRCh38
rs780941248 Chr.19: 48123218-48123218 on Build GRCh38
rs575836346 Chr.19: 48123133-48123133 on Build GRCh38
rs988574489 Chr.19: 48123556-48123556 on Build GRCh38
rs201550323 Chr.19: 48123177-48123177 on Build GRCh38
rs774233229 Chr.19: 48123296-48123296 on Build GRCh38
rs745863395 Chr.19: 48123369-48123369 on Build GRCh38
rs979623338 Chr.19: 48123566-48123566 on Build GRCh38
rs768664854 Chr.19: 48123302-48123302 on Build GRCh38
rs988328914 Chr.19: 48123128-48123128 on Build GRCh38
rs201353645 Chr.19: 48123316-48123316 on Build GRCh38
rs1020935689 Chr.19: 48123557-48123557 on Build GRCh38
rs369814356 Chr.19: 48123228-48123228 on Build GRCh38
rs760719822 Chr.19: 48123249-48123249 on Build GRCh38
rs772196964 Chr.19: 48123472-48123472 on Build GRCh38
rs769562354 Chr.19: 48123229-48123229 on Build GRCh38
rs748346381 Chr.19: 48123444-48123445 on Build GRCh38
rs562392879 Chr.19: 48123489-48123489 on Build GRCh38
rs1046725630 Chr.19: 48123535-48123535 on Build GRCh38
rs773335614 Chr.19: 48123486-48123486 on Build GRCh38
rs368993529 Chr.19: 48123515-48123515 on Build GRCh38
rs900838413 Chr.19: 48123392-48123392 on Build GRCh38
rs753278583 Chr.19: 48123431-48123431 on Build GRCh38
rs777266972 Chr.19: 48123357-48123357 on Build GRCh38
rs542376014 Chr.19: 48123124-48123124 on Build GRCh38
rs187905940 Chr.19: 48123565-48123565 on Build GRCh38
rs751139851 Chr.19: 48123276-48123276 on Build GRCh38
rs3731008 Chr.19: 48123293-48123293 on Build GRCh38
rs754494250 Chr.19: 48123280-48123280 on Build GRCh38
rs551073867 Chr.19: 48123517-48123517 on Build GRCh38
rs777689854 Chr.19: 48123290-48123290 on Build GRCh38
rs555962151 Chr.19: 48123330-48123330 on Build GRCh38
rs942464396 Chr.19: 48123359-48123359 on Build GRCh38
rs747774796 Chr.19: 48123287-48123287 on Build GRCh38
rs768423303 Chr.19: 48123126-48123126 on Build GRCh38
rs914082051 Chr.19: 48123108-48123108 on Build GRCh38
rs770901517 Chr.19: 48123294-48123294 on Build GRCh38

View Full Product Details