Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48724638-48725135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASIP1
Entrez Gene ID
54922
Gene Name
Ras interacting protein 1
Gene Aliases
RAIN
UniGene
Hs.233955
Gene Chromosome Location
Chr.19: 48720585-48740833 on Build GRCh38
Transcripts
NM_017805.2 XM_011527053.2 XM_011527054.2 XM_011527055.2 XM_017026914.1 XM_017026915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
333318 c.2271G>T p.L757L Chr.19: 48724817-48724817 on Build GRCh38
3422982 c.2284G>A p.E762K Chr.19: 48724804-48724804 on Build GRCh38
5962228 c.2185G>T p.E729* Chr.19: 48724903-48724903 on Build GRCh38
3107864 c.2138C>G p.S713* Chr.19: 48724950-48724950 on Build GRCh38
5961540 c.2323T>C p.L775L Chr.19: 48724765-48724765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199721800 Chr.19: 48724699-48724699 on Build GRCh38
rs917480088 Chr.19: 48725085-48725085 on Build GRCh38
rs761318658 Chr.19: 48724799-48724799 on Build GRCh38
rs201454840 Chr.19: 48724861-48724861 on Build GRCh38
rs756762672 Chr.19: 48724722-48724722 on Build GRCh38
rs867062678 Chr.19: 48725021-48725021 on Build GRCh38
rs534906467 Chr.19: 48724731-48724731 on Build GRCh38
rs533104945 Chr.19: 48725042-48725042 on Build GRCh38
rs774618405 Chr.19: 48724872-48724872 on Build GRCh38
rs76284216 Chr.19: 48724955-48724955 on Build GRCh38
rs373380374 Chr.19: 48724854-48724854 on Build GRCh38
rs899828449 Chr.19: 48724909-48724909 on Build GRCh38
rs2287921 Chr.19: 48725015-48725015 on Build GRCh38
rs374850746 Chr.19: 48724973-48724973 on Build GRCh38
rs776430624 Chr.19: 48724773-48724773 on Build GRCh38
rs754313593 Chr.19: 48724827-48724827 on Build GRCh38
rs144968151 Chr.19: 48725106-48725106 on Build GRCh38
rs753452522 Chr.19: 48724721-48724721 on Build GRCh38
rs777029102 Chr.19: 48725007-48725007 on Build GRCh38
rs768020611 Chr.19: 48724677-48724677 on Build GRCh38
rs372046630 Chr.19: 48724730-48724730 on Build GRCh38
rs758096963 Chr.19: 48724848-48724848 on Build GRCh38
rs34354498 Chr.19: 48724802-48724802 on Build GRCh38
rs540884748 Chr.19: 48724992-48724992 on Build GRCh38
rs1027710127 Chr.19: 48724841-48724841 on Build GRCh38
rs756689051 Chr.19: 48724710-48724710 on Build GRCh38
rs1033016551 Chr.19: 48724662-48724662 on Build GRCh38
rs758005264 Chr.19: 48724747-48724747 on Build GRCh38
rs1028854984 Chr.19: 48724937-48724937 on Build GRCh38
rs890386231 Chr.19: 48724951-48724951 on Build GRCh38
rs958383314 Chr.19: 48725044-48725044 on Build GRCh38
rs917532114 Chr.19: 48724851-48724851 on Build GRCh38
rs760940466 Chr.19: 48724806-48724806 on Build GRCh38
rs780985377 Chr.19: 48724865-48724865 on Build GRCh38
rs764758039 Chr.19: 48724839-48724839 on Build GRCh38
rs111269191 Chr.19: 48724978-48724978 on Build GRCh38
rs777014384 Chr.19: 48724894-48724894 on Build GRCh38
rs762346774 Chr.19: 48724895-48724895 on Build GRCh38
rs748122600 Chr.19: 48724867-48724867 on Build GRCh38
rs950166393 Chr.19: 48724860-48724860 on Build GRCh38
rs751311940 Chr.19: 48724869-48724869 on Build GRCh38
rs370144366 Chr.19: 48724929-48724929 on Build GRCh38
rs377327537 Chr.19: 48724980-48724980 on Build GRCh38
rs796296101 Chr.19: 48724682-48724682 on Build GRCh38
rs146800732 Chr.19: 48724787-48724787 on Build GRCh38
rs150685376 Chr.19: 48724940-48724940 on Build GRCh38
rs531483937 Chr.19: 48724906-48724906 on Build GRCh38
rs924873195 Chr.19: 48724905-48724905 on Build GRCh38
rs868287322 Chr.19: 48724812-48724812 on Build GRCh38
rs764386217 Chr.19: 48724886-48724886 on Build GRCh38
rs937039834 Chr.19: 48724811-48724811 on Build GRCh38
rs754613166 Chr.19: 48724947-48724947 on Build GRCh38
rs768699666 Chr.19: 48724772-48724772 on Build GRCh38
rs765712432 Chr.19: 48724896-48724896 on Build GRCh38
rs762846127 Chr.19: 48724820-48724820 on Build GRCh38
rs775977001 Chr.19: 48724805-48724805 on Build GRCh38
rs202046985 Chr.19: 48724885-48724885 on Build GRCh38
rs778335654 Chr.19: 48724727-48724727 on Build GRCh38
rs978108517 Chr.19: 48724904-48724904 on Build GRCh38
rs34460988 Chr.19: 48724870-48724870 on Build GRCh38
rs114251138 Chr.19: 48724748-48724748 on Build GRCh38
rs772054805 Chr.19: 48724976-48724976 on Build GRCh38
rs936229937 Chr.19: 48724908-48724908 on Build GRCh38
rs746890403 Chr.19: 48724856-48724856 on Build GRCh38
rs563385587 Chr.19: 48725096-48725096 on Build GRCh38
rs34043113 Chr.19: 48724883-48724883 on Build GRCh38
rs771150276 Chr.19: 48724790-48724790 on Build GRCh38
rs764492529 Chr.19: 48724719-48724719 on Build GRCh38
rs375784788 Chr.19: 48724914-48724914 on Build GRCh38
rs148024901 Chr.19: 48724946-48724946 on Build GRCh38
rs952983590 Chr.19: 48725018-48725018 on Build GRCh38
rs754849879 Chr.19: 48724665-48724665 on Build GRCh38
rs773187933 Chr.19: 48724804-48724804 on Build GRCh38
rs539389939 Chr.19: 48724796-48724796 on Build GRCh38
rs757797723 Chr.19: 48725040-48725041 on Build GRCh38
rs544988157 Chr.19: 48725047-48725047 on Build GRCh38
rs973441996 Chr.19: 48724794-48724794 on Build GRCh38
rs894449382 Chr.19: 48724934-48724934 on Build GRCh38
rs201472500 Chr.19: 48724766-48724766 on Build GRCh38
rs991164539 Chr.19: 48725065-48725065 on Build GRCh38
rs1018696228 Chr.19: 48725093-48725093 on Build GRCh38
rs749903629 Chr.19: 48724847-48724847 on Build GRCh38
rs764449110 Chr.19: 48724816-48724816 on Build GRCh38
rs549015921 Chr.19: 48725107-48725107 on Build GRCh38
rs376943064 Chr.19: 48724830-48724830 on Build GRCh38
rs759778449 Chr.19: 48724670-48724670 on Build GRCh38
rs745926016 Chr.19: 48724873-48724873 on Build GRCh38
rs141007241 Chr.19: 48724974-48724974 on Build GRCh38
rs756174903 Chr.19: 48724959-48724959 on Build GRCh38

View Full Product Details