Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50878669-50879162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLK2
Entrez Gene ID
3817
Gene Name
kallikrein related peptidase 2
Gene Aliases
KLK2A2, hGK-1, hK2
UniGene
Hs.515560
Gene Chromosome Location
Chr.19: 50873433-50880567 on Build GRCh38
Transcripts
NM_001002231.2 NM_001256080.1 NM_005551.4 NR_045762.1 NR_045763.1

SNP Detail

SNP ID Chromosome Location
rs1015941318 Chr.19: 50878911-50878911 on Build GRCh38
rs926542905 Chr.19: 50878806-50878806 on Build GRCh38
rs566068251 Chr.19: 50879036-50879036 on Build GRCh38
rs984469547 Chr.19: 50878739-50878739 on Build GRCh38
rs1019742182 Chr.19: 50878693-50878693 on Build GRCh38
rs1028559383 Chr.19: 50878730-50878730 on Build GRCh38
rs113358789 Chr.19: 50878699-50878699 on Build GRCh38
rs968589009 Chr.19: 50878951-50878951 on Build GRCh38
rs1029029419 Chr.19: 50878879-50878879 on Build GRCh38
rs932845493 Chr.19: 50879115-50879115 on Build GRCh38
rs751822538 Chr.19: 50879122-50879122 on Build GRCh38
rs564658423 Chr.19: 50878791-50878791 on Build GRCh38
rs533797092 Chr.19: 50879066-50879066 on Build GRCh38
rs1044271433 Chr.19: 50878815-50878815 on Build GRCh38
rs773912267 Chr.19: 50878875-50878875 on Build GRCh38
rs537678710 Chr.19: 50879134-50879134 on Build GRCh38
rs1041132078 Chr.19: 50879080-50879080 on Build GRCh38
rs1053707722 Chr.19: 50878828-50878828 on Build GRCh38
rs978713132 Chr.19: 50878763-50878763 on Build GRCh38
rs924310901 Chr.19: 50878764-50878764 on Build GRCh38
rs9884 Chr.19: 50879042-50879042 on Build GRCh38
rs749259468 Chr.19: 50878841-50878841 on Build GRCh38
rs777671817 Chr.19: 50878800-50878800 on Build GRCh38
rs901366822 Chr.19: 50878870-50878870 on Build GRCh38
rs961684417 Chr.19: 50878913-50878913 on Build GRCh38
rs992596884 Chr.19: 50878788-50878788 on Build GRCh38
rs1041078960 Chr.19: 50878848-50878848 on Build GRCh38
rs1059711 Chr.19: 50878801-50878801 on Build GRCh38
rs183710386 Chr.19: 50878787-50878787 on Build GRCh38
rs936609825 Chr.19: 50879029-50879029 on Build GRCh38
rs960929683 Chr.19: 50879011-50879011 on Build GRCh38
rs970991077 Chr.19: 50878997-50878997 on Build GRCh38
rs562073596 Chr.19: 50878844-50878844 on Build GRCh38
rs915385008 Chr.19: 50878744-50878744 on Build GRCh38
rs1037326411 Chr.19: 50878852-50878852 on Build GRCh38
rs11090 Chr.19: 50878960-50878960 on Build GRCh38
rs916912512 Chr.19: 50879065-50879065 on Build GRCh38
rs541371645 Chr.19: 50878829-50878829 on Build GRCh38
rs1049932358 Chr.19: 50879125-50879125 on Build GRCh38
rs978382876 Chr.19: 50879002-50879002 on Build GRCh38
rs954109024 Chr.19: 50878729-50878729 on Build GRCh38
rs952982873 Chr.19: 50878898-50878898 on Build GRCh38
rs889900567 Chr.19: 50878923-50878923 on Build GRCh38
rs745433266 Chr.19: 50878949-50878949 on Build GRCh38
rs144992183 Chr.19: 50878706-50878706 on Build GRCh38
rs529403038 Chr.19: 50878933-50878933 on Build GRCh38
rs747230481 Chr.19: 50878751-50878751 on Build GRCh38
rs897180258 Chr.19: 50878860-50878860 on Build GRCh38
rs531987005 Chr.19: 50878825-50878825 on Build GRCh38
rs901253519 Chr.19: 50879113-50879113 on Build GRCh38
rs574417004 Chr.19: 50879138-50879138 on Build GRCh38
rs570311646 Chr.19: 50879120-50879120 on Build GRCh38
rs971821805 Chr.19: 50878914-50878914 on Build GRCh38
rs527965028 Chr.19: 50878755-50878755 on Build GRCh38
rs980224693 Chr.19: 50879136-50879136 on Build GRCh38
rs4846 Chr.19: 50879007-50879007 on Build GRCh38
rs145052361 Chr.19: 50879137-50879137 on Build GRCh38
rs77177726 Chr.19: 50878731-50878731 on Build GRCh38
rs1029603603 Chr.19: 50878903-50878903 on Build GRCh38
rs968370579 Chr.19: 50878753-50878753 on Build GRCh38
rs949220843 Chr.19: 50878799-50878799 on Build GRCh38
rs971769510 Chr.19: 50878790-50878790 on Build GRCh38
rs141318172 Chr.19: 50879095-50879095 on Build GRCh38
rs369146825 Chr.19: 50878928-50878928 on Build GRCh38
rs1005771296 Chr.19: 50878909-50878909 on Build GRCh38

View Full Product Details