Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9405525-9406048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP2
Entrez Gene ID
8853
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1
UniGene
Hs.555902
Gene Chromosome Location
Chr.2: 9206765-9405683 on Build GRCh38
Transcripts
NM_001135191.1 NM_003887.2 XM_006711898.1 XM_006711901.3 XM_006711902.1 XM_011510403.2 XM_011510404.2 XM_011510405.2 XM_011510406.2 XM_011510407.2 XM_011510409.1 NM_001319066.1 NM_001319067.1 NM_001319068.1 NM_001319069.1 NM_001319070.1 NM_004763.4 NM_022334.4 NR_134951.1 NR_134952.1 NR_134953.1 NR_134954.1 NR_134955.1 NR_134956.1 NR_134957.1

Gene Symbol
ITGB1BP1
Entrez Gene ID
9270
Gene Name
integrin subunit beta 1 binding protein 1
Gene Aliases
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B
UniGene
Hs.467662
Gene Chromosome Location
Chr.2: 9405681-9423569 on Build GRCh38
Transcripts
NM_001135191.1 NM_003887.2 XM_006711898.1 XM_006711901.3 XM_006711902.1 XM_011510403.2 XM_011510404.2 XM_011510405.2 XM_011510406.2 XM_011510407.2 XM_011510409.1 NM_001319066.1 NM_001319067.1 NM_001319068.1 NM_001319069.1 NM_001319070.1 NM_004763.4 NM_022334.4 NR_134951.1 NR_134952.1 NR_134953.1 NR_134954.1 NR_134955.1 NR_134956.1 NR_134957.1

SNP Detail

SNP ID Chromosome Location
rs921125262 Chr.2: 9405756-9405756 on Build GRCh38
rs947710347 Chr.2: 9405760-9405760 on Build GRCh38
rs1025604960 Chr.2: 9405638-9405638 on Build GRCh38
rs113446247 Chr.2: 9406005-9406005 on Build GRCh38
rs1054046000 Chr.2: 9405922-9405922 on Build GRCh38
rs1013748712 Chr.2: 9405632-9405632 on Build GRCh38
rs894094099 Chr.2: 9405925-9405925 on Build GRCh38
rs1018808626 Chr.2: 9405906-9405906 on Build GRCh38
rs369449557 Chr.2: 9405754-9405755 on Build GRCh38
rs984644090 Chr.2: 9405624-9405624 on Build GRCh38
rs1018258820 Chr.2: 9405641-9405641 on Build GRCh38
rs551299130 Chr.2: 9405955-9405955 on Build GRCh38
rs749174484 Chr.2: 9405909-9405909 on Build GRCh38
rs909795998 Chr.2: 9406013-9406013 on Build GRCh38
rs976721180 Chr.2: 9405913-9405913 on Build GRCh38
rs568699625 Chr.2: 9405948-9405948 on Build GRCh38
rs1021039630 Chr.2: 9405709-9405709 on Build GRCh38
rs923656579 Chr.2: 9405743-9405743 on Build GRCh38
rs946177371 Chr.2: 9405698-9405698 on Build GRCh38
rs112694656 Chr.2: 9406010-9406010 on Build GRCh38
rs964054986 Chr.2: 9405674-9405674 on Build GRCh38
rs187834036 Chr.2: 9405791-9405791 on Build GRCh38
rs936420605 Chr.2: 9405747-9405747 on Build GRCh38
rs916337485 Chr.2: 9406021-9406021 on Build GRCh38
rs111470184 Chr.2: 9405843-9405843 on Build GRCh38
rs10593 Chr.2: 9405901-9405901 on Build GRCh38
rs376591339 Chr.2: 9405871-9405871 on Build GRCh38
rs917005745 Chr.2: 9405877-9405877 on Build GRCh38
rs758324503 Chr.2: 9405600-9405600 on Build GRCh38
rs778112684 Chr.2: 9405915-9405916 on Build GRCh38
rs906467558 Chr.2: 9405858-9405858 on Build GRCh38
rs1032318598 Chr.2: 9405676-9405676 on Build GRCh38
rs564214089 Chr.2: 9405691-9405691 on Build GRCh38
rs897955303 Chr.2: 9405604-9405604 on Build GRCh38
rs539850840 Chr.2: 9405803-9405803 on Build GRCh38
rs878857962 Chr.2: 9406008-9406008 on Build GRCh38
rs183107032 Chr.2: 9405932-9405932 on Build GRCh38
rs375868643 Chr.2: 9405949-9405949 on Build GRCh38
rs905428125 Chr.2: 9405660-9405660 on Build GRCh38
rs1026125593 Chr.2: 9405562-9405565 on Build GRCh38
rs1028135508 Chr.2: 9405951-9405951 on Build GRCh38
rs1002899784 Chr.2: 9405665-9405665 on Build GRCh38
rs954016686 Chr.2: 9405956-9405956 on Build GRCh38
rs187400497 Chr.2: 9405940-9405940 on Build GRCh38
rs939697973 Chr.2: 9405824-9405824 on Build GRCh38
rs1002415733 Chr.2: 9405916-9405916 on Build GRCh38
rs965885302 Chr.2: 9405715-9405715 on Build GRCh38
rs748377124 Chr.2: 9405558-9405558 on Build GRCh38
rs958026344 Chr.2: 9405678-9405678 on Build GRCh38
rs865914061 Chr.2: 9405670-9405670 on Build GRCh38
rs192727720 Chr.2: 9405958-9405958 on Build GRCh38
rs953957824 Chr.2: 9405775-9405775 on Build GRCh38
rs976593980 Chr.2: 9405748-9405748 on Build GRCh38
rs909514500 Chr.2: 9405815-9405815 on Build GRCh38
rs1020990315 Chr.2: 9405947-9405947 on Build GRCh38
rs41264171 Chr.2: 9405631-9405631 on Build GRCh38
rs770897020 Chr.2: 9405974-9405974 on Build GRCh38
rs112829430 Chr.2: 9405883-9405884 on Build GRCh38
rs905375291 Chr.2: 9405911-9405911 on Build GRCh38
rs772082699 Chr.2: 9405680-9405680 on Build GRCh38
rs147582792 Chr.2: 9405749-9405750 on Build GRCh38
rs192272981 Chr.2: 9405799-9405799 on Build GRCh38

View Full Product Details