Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31931939-31932434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEMO1
Entrez Gene ID
51072
Gene Name
mediator of cell motility 1
Gene Aliases
C2orf4, CGI-27, MEMO, NS5ATP7
UniGene
Hs.444969
Gene Chromosome Location
Chr.2: 31867188-32011052 on Build GRCh38
Transcripts
NM_001137602.2 NM_001301833.1 NM_001301852.1 NM_015955.3 NR_126032.1 NR_126034.1 XM_011532892.2 XM_011532896.2 XM_017004255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6452726 c.212C>T p.T71I Chr.2: 31932067-31932067 on Build GRCh38
6325055 c.187G>A p.A63T Chr.2: 31932101-31932101 on Build GRCh38
4873103 c.161G>T p.G54V Chr.2: 31932127-31932127 on Build GRCh38
1020148 c.152G>T p.G51V Chr.2: 31932127-31932127 on Build GRCh38
6401534 c.149C>T p.A50V Chr.2: 31932130-31932130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970338542 Chr.2: 31932099-31932099 on Build GRCh38
rs771537562 Chr.2: 31932152-31932152 on Build GRCh38
rs755117715 Chr.2: 31932057-31932057 on Build GRCh38
rs977598540 Chr.2: 31932005-31932005 on Build GRCh38
rs375610194 Chr.2: 31932009-31932009 on Build GRCh38
rs763592061 Chr.2: 31932040-31932040 on Build GRCh38
rs774098214 Chr.2: 31932349-31932349 on Build GRCh38
rs779001021 Chr.2: 31932161-31932162 on Build GRCh38
rs763836880 Chr.2: 31932173-31932173 on Build GRCh38
rs988316651 Chr.2: 31932277-31932277 on Build GRCh38
rs752354630 Chr.2: 31932028-31932028 on Build GRCh38
rs948942270 Chr.2: 31932066-31932066 on Build GRCh38
rs1007459974 Chr.2: 31932307-31932307 on Build GRCh38
rs745955873 Chr.2: 31931983-31931983 on Build GRCh38
rs375894934 Chr.2: 31932120-31932120 on Build GRCh38
rs767019086 Chr.2: 31932023-31932023 on Build GRCh38
rs913704527 Chr.2: 31932300-31932300 on Build GRCh38
rs747491821 Chr.2: 31932054-31932054 on Build GRCh38
rs933133659 Chr.2: 31932306-31932306 on Build GRCh38
rs924950907 Chr.2: 31932162-31932162 on Build GRCh38
rs201060162 Chr.2: 31932071-31932071 on Build GRCh38
rs759538079 Chr.2: 31932148-31932148 on Build GRCh38
rs940188445 Chr.2: 31932138-31932138 on Build GRCh38
rs371259294 Chr.2: 31932171-31932171 on Build GRCh38
rs749426274 Chr.2: 31932057-31932058 on Build GRCh38
rs759169335 Chr.2: 31932018-31932018 on Build GRCh38
rs753439407 Chr.2: 31932179-31932179 on Build GRCh38
rs76862178 Chr.2: 31932346-31932346 on Build GRCh38
rs190447823 Chr.2: 31931974-31931974 on Build GRCh38
rs200462854 Chr.2: 31932111-31932111 on Build GRCh38
rs993996893 Chr.2: 31931984-31931984 on Build GRCh38
rs554639191 Chr.2: 31932051-31932051 on Build GRCh38
rs534879746 Chr.2: 31932048-31932048 on Build GRCh38
rs992414674 Chr.2: 31932031-31932031 on Build GRCh38
rs774466982 Chr.2: 31932137-31932137 on Build GRCh38
rs775076970 Chr.2: 31932155-31932155 on Build GRCh38
rs755775655 Chr.2: 31932039-31932039 on Build GRCh38
rs750790214 Chr.2: 31932043-31932043 on Build GRCh38
rs748120049 Chr.2: 31932073-31932073 on Build GRCh38
rs957774297 Chr.2: 31932275-31932275 on Build GRCh38
rs199741022 Chr.2: 31932154-31932154 on Build GRCh38
rs140215147 Chr.2: 31932075-31932075 on Build GRCh38
rs757311448 Chr.2: 31932145-31932145 on Build GRCh38
rs576834284 Chr.2: 31932358-31932358 on Build GRCh38
rs1021524709 Chr.2: 31932314-31932314 on Build GRCh38
rs865827659 Chr.2: 31932312-31932312 on Build GRCh38
rs763111118 Chr.2: 31932188-31932188 on Build GRCh38
rs546087048 Chr.2: 31932393-31932393 on Build GRCh38

View Full Product Details