Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70453139-70453634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFA
Entrez Gene ID
7039
Gene Name
transforming growth factor alpha
Gene Aliases
TFGA
UniGene
Hs.170009
Gene Chromosome Location
Chr.2: 70447280-70554015 on Build GRCh38
Transcripts
NM_001099691.2 NM_001308158.1 NM_001308159.1 NM_003236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3582796 c.436C>T p.L146F Chr.2: 70453257-70453257 on Build GRCh38
5745905 c.433G>A p.A145T Chr.2: 70453260-70453260 on Build GRCh38
4733342 c.429C>T p.P143P Chr.2: 70453264-70453264 on Build GRCh38
1685516 c.366-1_376delGCTGCTGCCAGG p.? Chr.2: 70453317-70453328 on Build GRCh38
1267706 c.425A>C p.K142T Chr.2: 70453268-70453268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781896307 Chr.2: 70453317-70453317 on Build GRCh38
rs372158742 Chr.2: 70453292-70453292 on Build GRCh38
rs62149639 Chr.2: 70453437-70453437 on Build GRCh38
rs782808061 Chr.2: 70453259-70453259 on Build GRCh38
rs1000972903 Chr.2: 70453577-70453577 on Build GRCh38
rs146962706 Chr.2: 70453313-70453313 on Build GRCh38
rs185312436 Chr.2: 70453467-70453467 on Build GRCh38
rs781954461 Chr.2: 70453361-70453362 on Build GRCh38
rs572811235 Chr.2: 70453476-70453476 on Build GRCh38
rs368248694 Chr.2: 70453273-70453273 on Build GRCh38
rs782095962 Chr.2: 70453239-70453239 on Build GRCh38
rs140828225 Chr.2: 70453249-70453249 on Build GRCh38
rs781849385 Chr.2: 70453181-70453181 on Build GRCh38
rs782058394 Chr.2: 70453261-70453261 on Build GRCh38
rs530453246 Chr.2: 70453555-70453555 on Build GRCh38
rs782774951 Chr.2: 70453180-70453180 on Build GRCh38
rs782001599 Chr.2: 70453244-70453244 on Build GRCh38
rs782034148 Chr.2: 70453344-70453344 on Build GRCh38
rs782356331 Chr.2: 70453304-70453304 on Build GRCh38
rs11466264 Chr.2: 70453479-70453479 on Build GRCh38
rs781837347 Chr.2: 70453178-70453178 on Build GRCh38
rs782486455 Chr.2: 70453195-70453195 on Build GRCh38
rs782558378 Chr.2: 70453182-70453182 on Build GRCh38
rs573763983 Chr.2: 70453442-70453442 on Build GRCh38
rs535119179 Chr.2: 70453240-70453240 on Build GRCh38
rs887900658 Chr.2: 70453439-70453439 on Build GRCh38
rs782115973 Chr.2: 70453247-70453247 on Build GRCh38
rs943417996 Chr.2: 70453456-70453456 on Build GRCh38
rs200451222 Chr.2: 70453286-70453286 on Build GRCh38
rs916059931 Chr.2: 70453290-70453290 on Build GRCh38
rs782472588 Chr.2: 70453179-70453179 on Build GRCh38
rs782394183 Chr.2: 70453257-70453257 on Build GRCh38
rs782701320 Chr.2: 70453171-70453171 on Build GRCh38
rs368643430 Chr.2: 70453212-70453212 on Build GRCh38
rs782326575 Chr.2: 70453238-70453238 on Build GRCh38
rs199594512 Chr.2: 70453234-70453234 on Build GRCh38
rs782267879 Chr.2: 70453205-70453205 on Build GRCh38
rs782700834 Chr.2: 70453271-70453271 on Build GRCh38
rs147241222 Chr.2: 70453278-70453278 on Build GRCh38
rs782687296 Chr.2: 70453226-70453226 on Build GRCh38
rs782586279 Chr.2: 70453293-70453293 on Build GRCh38
rs782293622 Chr.2: 70453492-70453492 on Build GRCh38
rs371359481 Chr.2: 70453433-70453433 on Build GRCh38
rs375187935 Chr.2: 70453337-70453337 on Build GRCh38
rs782179752 Chr.2: 70453302-70453302 on Build GRCh38
rs781911960 Chr.2: 70453282-70453282 on Build GRCh38
rs782043633 Chr.2: 70453198-70453198 on Build GRCh38
rs190572527 Chr.2: 70453473-70453473 on Build GRCh38
rs370068770 Chr.2: 70453170-70453170 on Build GRCh38
rs781826380 Chr.2: 70453272-70453272 on Build GRCh38
rs566085609 Chr.2: 70453198-70453198 on Build GRCh38
rs75511448 Chr.2: 70453481-70453481 on Build GRCh38
rs782185921 Chr.2: 70453194-70453194 on Build GRCh38
rs782253342 Chr.2: 70453314-70453314 on Build GRCh38
rs554966762 Chr.2: 70453277-70453277 on Build GRCh38
rs1020842318 Chr.2: 70453270-70453270 on Build GRCh38
rs782674265 Chr.2: 70453370-70453370 on Build GRCh38
rs965977294 Chr.2: 70453241-70453241 on Build GRCh38
rs782650087 Chr.2: 70453189-70453189 on Build GRCh38
rs535108529 Chr.2: 70453260-70453260 on Build GRCh38
rs578081517 Chr.2: 70453291-70453291 on Build GRCh38
rs782325140 Chr.2: 70453361-70453361 on Build GRCh38

View Full Product Details